Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C1orf226 (Chromosome 1 Open Reading Frame 226) is a Protein Coding gene. Diseases associated with C1orf226 include Phosphoglycerate Dehydrogenase Deficiency. An important paralog of this gene is DAB2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C1orf226
Gene Aliases: C1orf226
UniProt ID: (Human) A1L170
Entrez Gene ID: (Human) 400793
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support