Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GAIGGRGLPII QRGLPRGGLR GGRATRTLLR GGMSLRGQNL LRGGRAVAPR MGLRRGGVRG RGGPGRGGLG RGAMGRGG
Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.
CHTOP is a small nuclear protein that is characterized by an arginine and glycine rich region. This protein may have an important role in the regulation of fetal globin gene expression and in the activation of estrogen-responsive genes. A recent study reported that this protein binds 5-hydroxymethylcytosine (5hmC) and associates with an arginine methyltransferase complex (methylosome), which promotes methylation of arginine 3 of histone H4 (H4R3) and activation of genes involved in glioblastomagenesis. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chromatin target of PRMT1 protein; DKFZP547E1010; Friend of Prmt1; Friend of PRMT1 protein; RP1-178F15.2; Small arginine- and glycine-rich protein; small protein rich in arginine and glycine; SRAG
Gene Aliases: C1orf77; CHTOP; FL-SRAG; FOP; HT031; PP7704; SRAG; SRAG-3; SRAG-5
UniProt ID: (Human) Q9Y3Y2
Entrez Gene ID: (Human) 26097
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support