Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VADRLLQFSTI QQNDWLRALS DNA^M
^M
Highest antigen sequence identity to the following orthologs - mouse 75%, rat 79%.
TMEM267 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein 267
Gene Aliases: C5orf28; TMEM267
UniProt ID: (Human) Q0VDI3
Entrez Gene ID: (Human) 64417
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support