Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FAM65B Antibodies

          Abnova

          C6orf32 Monoclonal Antibody (2E6-1B10)

          View all (6) FAM65B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite C6orf32 Monoclonal Antibody (2E6-1B10)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          C6orf32 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          C6orf32 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          C6orf32 Antibody (H00009750-M01) in ELISA

          Detection limit for recombinant GST tagged FAM65B is 0.1 ng/mL as a capture antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          C6orf32 Antibody in ELISA (ELISA)
          C6orf32 Monoclonal Antibody (2E6-1B10)

          Product Details

          H00009750-M01

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2E6-1B10

          Immunogen

          C6orf32 (AAH01232.1, 1 a.a. approximately 591 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Sandwich ELISA (Recombinant protein).

          Immunogen sequence: MLVGSQSFSP GGPNGIIRSQ SFAGFSGLQE RRSRCNSFIE NSSALKKPQA KLKKMHNLGH KNNNPPKEPQ PKRVEEVYRA LKNGLDEYLE VHQTELDKLT AQLKDMKRNS RLGVLYDLDK QIKTIERYMR RLEFHISKVD ELYEAYCIQR RLQDGASKMK QAFATSPASK AARESLTEIN RSFKEYTENM CTIEVELENL LGEFSIKMKG LAGFARLCPG DQYEIFMKYG RQRWKLKGKI EVNGKQSWDG EETVFLPLIV GFISIKVTEL KGLATHILVG SVTCETKELF AARPQVVAVD INDLGTIKLN LEITWYPFDV EDMTASSGAG NKAAALQRRM SMYSQGTPET PTFKDHSFFS NLPDDIFENG KAAEEKMPLS LSFSDLPNGD CALTSHSTGS PSNSTNPEIT ITPAEFNLSS LASQNEGMDD TSSASSRNSL GEGQEPKSHL KEEDPEEPRK PASAPSEACR RQSSGAGAEH LFLENDVAEA LLQESEEASE LKPVELDTSE GNITKQLVKR LTSAEVPMAT DRLLSEGSVG GESEGCRSFL DGSLEDAFNG LLLALEPHKE QYKEFQDLNQ EVMNLDDILK K

          Target Information

          Required for hearing (PubMed:24958875). Involved in skeletal muscle development (PubMed:24687993).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: family with sequence similarity 65 member B; KIAA0386; myogenesis-related and NCAM-associated protein homolog; protein FAM65B; Rho family-interacting cell polarization regulator 2; unnamed protein product

          View more View less

          Gene Aliases: C6orf32; DFNA21; DFNB104; DIFF40; DIFF48; FAM65B; KIAA0386; MYONAP; PL48; RIPOR2

          View more View less

          UniProt ID: (Human) Q9Y4F9

          View more View less

          Entrez Gene ID: (Human) 9750

          View more View less

          Function(s)
          protein binding 14-3-3 protein binding
          Process(es)
          chemotaxis cell adhesion negative regulation of cell adhesion muscle organ development sensory perception of sound negative regulation of signal transduction cell differentiation negative regulation of Rho protein signal transduction negative regulation of T cell proliferation positive regulation of myoblast differentiation positive regulation of filopodium assembly regulation of cell cycle positive regulation of neutrophil chemotaxis regulation of mitotic spindle assembly positive regulation of myoblast fusion negative regulation of establishment of T cell polarity negative regulation of protein localization to cell leading edge cellular response to chemokine regulation of establishment of cell polarity positive regulation of neutrophil extravasation negative regulation of T cell migration negative regulation of Rho guanyl-nucleotide exchange factor activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-2x6nv:80/100.66.78.26:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline