Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Inhibits PPP1CA phosphatase activity. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein phosphatase 1 regulatory subunit 35; protein phosphatase 1, regulatory subunit 35; UPF0683 protein C7orf47; UPF0683 protein C7orf47 homolog
Gene Aliases: C7orf47; PPP1R35
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support