Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TEX47 (Testis Expressed 47) is a Protein Coding gene. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Testis-expressed protein 47
Gene Aliases: C7orf62; TEX47
UniProt ID: (Human) Q8TBZ9
Entrez Gene ID: (Human) 219557
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support