Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Hgh1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain protein 16; brain protein 16-like; family with sequence similarity 203, member A; family with sequence similarity 203, member B; protein FAM203A; protein FAM203B; Protein HGH1 homolog
Gene Aliases: BRP16; BRP16L; C8orf30A; C8orf30B; D15Ertd741e; FAM203A; FAM203B; HGH1; MNCb-5873
UniProt ID: (Rat) Q6AY79, (Human) Q9BTY7, (Mouse) Q9JJF4
Entrez Gene ID: (Rat) 315094, (Human) 51236, (Mouse) 59053
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support