Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAICPELAQ TDKSALANLS DETETLKNFT DEVQTSSSFS SSGGRQSSPL TSGSKLEREK QTPSLEQGDT QSELLDYKNY EKKLSKKWIN YLKLKDSNFE RHQPDTKLPT EITRVSDEEL NALQSYCTMK INLIHRRGDS KKKTSSRHKK LHLGLDVEAS ERDAFSCTVP DELLNRIYFK NMRTTPKQEA AAKQHISYQC PYCNRKRAEL ALSAFLKQKK TLLESFLLQE RIDEHLHTKD FLTRIGEAHQ DFPRLSDDPR IIWKRLTEKS HIRYSGFERS ETEQKMQRDG NSACHLPFSL PFLKRLTLIK PELVIVNDNV (1-319 aa encoded by BC031245)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C8orf48
Gene Aliases: C8orf48
UniProt ID: (Human) Q96LL4
Entrez Gene ID: (Human) 157773
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support