Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
C9orf114 is a protein coding gene. Required for association of the centrosomes with the poles of the bipolar mitotic spindle during metaphase. Also involved in chromosome alignment. May promote centrosome maturation probably by recruiting A-kinase anchor protein AKAP9 to centrosomes in early mitosis. Binds specifically to miRNA MIR145 hairpin, regulates MIR145 expression at a postranscriptional level.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centromere protein 32; kinetochore-associated protein; methyltransferase C9orf114; methyltransferase C9orf114 homolog; putative methyltransferase C9orf114; putative methyltransferase C9orf114 homolog; ribosomal RNA methyltransferase SPOUT1; uncharacterized protein C9orf114 homolog; unnamed protein product
Gene Aliases: 9930028P05; C9orf114; CENP-32; CENP32; D2Wsu81e; HSPC109; NEDGSB
UniProt ID: (Human) Q5T280, (Mouse) Q3UHX9
Entrez Gene ID: (Human) 51490, (Mouse) 227695, (Rat) 499770
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support