Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTSTSPAA MLLRRLRRLS WGSTAVQLFI LTVVTFGLLA PLACHRLLHS YFYLRHWHLN QMSQEFLQQS LKEGEAALHY FEELPSANGS VPIVWQATPR PWLVITIITV DRQPGFHYVL QVVSQFHRLL QQCGPQCEGH QLFLCNVERS VSHFDAKLLS KYVPVANRYE GTEDDYGDDP STNSFEKEKQ DYVYCLESSL QTYNPDYVLM VEDDAVPEEQ IFPVLEHLLR ARFSEPHLRD ALYLKLYHPE RLQHYINPEP M (1-260 aa encoded by BC006115)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Post-GPI attachment to proteins factor 4; Post-GPI attachment to proteins GalNAc transferase 4; Transmembrane protein 246; transmembrane protein C9orf125
Gene Aliases: C9orf125; PGAP4; TMEM246
UniProt ID: (Human) Q9BRR3
Entrez Gene ID: (Human) 84302
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support