Predicted to react with Mouse samples.
This gene belongs to a subfamily of calcium binding proteins that share similarity to calmodulin. Like calmodulin, these family members can likely stimulate calmodulin-dependent kinase II and the protein phosphatase calcineurin. Calcium binding proteins are an important component of calcium mediated cellular signal transduction. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jan 2016].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CaBP2; Calcium-binding protein 2
Gene Aliases: CABP2; DFNB93
UniProt ID: (Human) Q9NPB3, (Mouse) Q9JLK4
Entrez Gene ID: (Human) 51475, (Mouse) 29866
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support