Sequence of this protein is as follows: MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKVKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAK
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calmodulin 2 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; LP7057 protein; phosphorylase kinase delta; phosphorylase kinase subunit delta; prepro-calmodulin 2
Gene Aliases: CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMII; CAMIII; LQT15; PHKD; PHKD2
UniProt ID: (Human) P0DP24
Entrez Gene ID: (Human) 805
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support