Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | - |
Immunohistochemistry (IHC) |
1:100-1:500 | - |
Immunoprecipitation (IP) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Between 1180 and 1230 |
Conjugate |
Unconjugated |
Form |
liquid |
Concentration |
1 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
TBS, pH 7.0 to 8.0 |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
CAND1 enhances transcription from various types of promoters. It is a regulatory protein that interferes with the assembly of the SCF (SKP1-CUL1-F-box protein) ubiquitin ligase complex and thereby down-regulates ubiquitination of target proteins. CAND1 prevents neddylation of CUL1 by physically blocking access to the neddylation site. CAND1 disrupts interactions between CUL1 and SKP1A and between CUL1 and F-box proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cullin-associated and neddylation-dissociated 1; Cullin-associated and neddylation-dissociated protein 1; Cullin-associated NEDD8-dissociated protein 1; DKFZp434M1414; FLJ10114; FLJ10929; FLJ38691; FLJ90441; p120 CAND1; TBP interacting protein; TBP-interacting protein; TBP-interacting protein 120A; TBP-interacting protein of 120 kDa A
Gene Aliases: 2310038O07Rik; 6330512O03Rik; AI195005; AI846556; CAND1; D10Ertd516e; KIAA0829; mKIAA0829; TIP120; TIP120A; Tp120a
UniProt ID: (Human) Q86VP6, (Mouse) Q6ZQ38
Entrez Gene ID: (Human) 55832, (Mouse) 71902
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support