Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1 µg/mL | View 1 publication 1 publication |
Immunocytochemistry (ICC/IF) |
8-25 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Published species |
Not Applicable |
Host/Isotype |
Rat / IgG2b |
Class |
Monoclonal |
Type |
Antibody |
Clone |
756613 |
Immunogen |
E. coli-derived recombinant mouse CCL3/MIP-1 alpha Ala24-Ala92 |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.5 mg/mL |
Purification |
Protein A/G |
Storage buffer |
PBS with 5% trehalose |
Contains |
No Preservative |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2609517 |
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Both MIP-1alpha and MIP-1beta are structurally and functionally related CC chemokines. They participate in host response to invading bacterial, viral, parasite and fungal pathogens by regulating the trafficking and activation state of selected subgroups of inflammatory cells (e.g. macrophages, lymphocytes and NK cells). While both MIP-1alpha and MIP-1beta exert similar effects on monocytes, their effect on lymphocytes differ; with MIP-1alpha selectively attracting CD8+ lymphocytes, and MIP-1beta selectively attracting CD4+ lymphocytes. Additionally, MIP-1alpha and MIP-1beta have also been shown to be potent chemoattractants for B cells, eosinophils and dendritic cells. Both human and murine MIP-1alpha and MIP-1beta are active on human and murine hematopoietic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C motif chemokine 3; H-MIP-1-alpha; Heparin-binding chemotaxis protein; L2G25B; LD78-alpha; Macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1alpha; MIP-1 alpha; MIP-1-alpha; MIP1 (a); mip1 alpha; RP23-320E6.7; SIS-alpha; small inducible cytokine A3; Small-inducible cytokine A3; TY-5
Gene Aliases: AI323804; Ccl3; G0S19-1; LD78alpha; MIP-1alpha; MIP1-(a); MIP1-alpha; Mip1a; Scya3
UniProt ID: (Mouse) P10855
Entrez Gene ID: (Mouse) 20302
Molecular Function:
cytokine
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support