Invitrogen
Catalog # 12-1152-81
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | 0.06 µg/test |
PUBLISHED APPLICATIONS | |
---|---|
Flow Cytometry (Flow) | See 37 publications below |
Immunofluorescence (IF) | See 1 publications below |
Western Blot (WB) | See 1 publications below |
Species reactivity | Mouse |
Published species | Mouse |
Host / Isotype | Rat / IgG2a, kappa |
Recommended Isotype Control | Rat IgG2a kappa Isotype Control, PE, eBioscience™ |
Class | Monoclonal |
Type | Antibody |
Clone | AFS98 |
Conjugate |
PE
|
Excitation/Emission Profile | View spectra |
Form | Liquid |
Concentration | 0.2 mg/mL |
Purification | Affinity chromatography |
Storage buffer | PBS, pH 7.2, with 0.1% gelatin |
Contains | 0.09% sodium azide |
Storage conditions | 4° C, store in dark, DO NOT FREEZE! |
RRID | AB_465807 |
Description: The AFS98 monoclonal antibody reacts with the mouse CD115 molecule, a receptor for macrophage colony stimulating factor (M-CSF) or colony stimulating factor-1 (CSF-1). CD115 is expressed by monocyte, macrophage, osteoclast, and some epithelial cells. It is a 150 kDa c-fms gene product and belongs to immunoglobulin family. CSF-1 signaling through CSF-1R regulates the proliferation and differentiation of cells in the monocytic lineage.
Applications Reported: The AFS98 antibody has been reported for use in flow cytometric analysis.
Applications Tested: The AFS98 antibody has been tested by flow cytometric analysis of peritoneal exudate cells. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
The protein encoded by this gene is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most if not all of the biological effects of this cytokine. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in this gene have been associated with a predisposition to myeloid malignancy. The first intron of this gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-fms; CD115; CSF-1 receptor; CSF-1-R; Macrophage colony-stimulating factor 1 receptor; Proto-oncogene c-Fms; proto-oncogene fms
Gene Aliases: AI323359; CD115; CSF-1R; Csf1r; Csfmr; Fim-2; Fms; M-CSF-R; M-CSFR
UniProt ID: (Mouse) P09581
Entrez Gene ID: (Mouse) 12978
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.