Product References
Metabolic improvement and liver regeneration by inhibiting CXXC5 function for non-alcoholic steatohepatitis treatment.
Experimental & molecular medicine
Seo SH,Kim E,Yoon M,Lee SH,Park BH,Choi KY
14-1331-82 was used in Immunohistochemistry to conclude that the pronounced metabolic improvements with the stimulation of liver regeneration by interfering with the CXXC5-Dvl interaction provide a therapeutic approach for NASH.
Thu Sep 01 00:00:00 EDT 2022
Prominin 1 and Notch regulate ciliary length and dynamics in multiciliated cells of the airway epithelium.
iScience
Serra CFH,Liu H,Qian J,Mori M,Lu J,Cardoso WV
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Fri Aug 19 00:00:00 EDT 2022
Prominin 1 and Notch regulate ciliary length and dynamics in multiciliated cells of the airway epithelium.
iScience
Serra CFH,Liu H,Qian J,Mori M,Lu J,Cardoso WV
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Fri Aug 19 00:00:00 EDT 2022
Analyzing mouse neural stem cell and progenitor cell proliferation using EdU incorporation and multicolor flow cytometry.
STAR protocols
Velloso FJ,Kumari E,Buono KD,Frondelli MJ,Levison SW
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Fri Mar 18 00:00:00 EDT 2022
C/EBPB-dependent adaptation to palmitic acid promotes tumor formation in hormone receptor negative breast cancer.
Nature communications
Liu XZ,Rulina A,Choi MH,Pedersen L,Lepland J,Takle ST,Madeleine N,Peters SD,Wogsland CE,Grøndal SM,Lorens JB,Goodarzi H,Lønning PE,Knappskog S,Molven A,Halberg N
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Mon Jan 10 00:00:00 EST 2022
RBP-J deficiency promoted the proliferation and differentiation of CD133-positive cells in both in vitro and in vivo studies.
The European journal of neuroscience
Ye X,Li M,Bian W,Wu A,Zhang T,Li J,Zhou P,Cui H,Ding YQ,Liao M,Sun C
14-1331-82 was used in Immunohistochemistry to study the effect of Notch signalling pathway on the neurogenesis of CD133-positive cells.
Fri Jul 01 00:00:00 EDT 2022
Cell population analysis of the adult murine subependymal neurogenic lineage by flow cytometry.
STAR protocols
Belenguer G,Duart-Abadia P,Domingo-Muelas A,Morante-Redolat JM,Fariñas I
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Fri Jun 18 00:00:00 EDT 2021
Green Synthesized Honokiol Transfersomes Relieve the Immunosuppressive and Stem-Like Cell Characteristics of the Aggressive B16F10 Melanoma.
International journal of nanomedicine
Ezzeldeen Y,Swidan S,ElMeshad A,Sebak A
14-1331-82 was used in Western Blot to fabricate Honokiol transfersomes for topical treatment of melanoma.
Wed Nov 24 00:00:00 EST 2021
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Wed Mar 11 00:00:00 EDT 2020
Moderate alcohol intake promotes pancreatic ductal adenocarcinoma development in mice expressing oncogenic Kras.
American journal of physiology. Gastrointestinal and liver physiology
Asahina K,Balog S,Hwang E,Moon E,Wan E,Skrypek K,Chen Y,Fernandez J,Romo J,Yang Q,Lai K,French SW,Tsukamoto H
14-1331-82 was used in Immunohistochemistry (Frozen) to test whether a combination of risk factors or moderate alcohol intake induces pancreatic ductal adenocarcinoma development in mice.
Sat Feb 01 00:00:00 EST 2020
Musashi-1 promotes cancer stem cell properties of glioblastoma cells via upregulation of YTHDF1.
Cancer cell international
Yarmishyn AA,Yang YP,Lu KH,Chen YC,Chien Y,Chou SJ,Tsai PH,Ma HI,Chien CS,Chen MT,Wang ML
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Western Blot
Mon Dec 14 00:00:00 EST 2020
Musashi-1 promotes cancer stem cell properties of glioblastoma cells via upregulation of YTHDF1.
Cancer cell international
Yarmishyn AA,Yang YP,Lu KH,Chen YC,Chien Y,Chou SJ,Tsai PH,Ma HI,Chien CS,Chen MT,Wang ML
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Western Blot
Mon Dec 14 00:00:00 EST 2020
Heterogeneity and dynamics of active Kras-induced dysplastic lineages from mouse corpus stomach.
Nature communications
Min J,Vega PN,Engevik AC,Williams JA,Yang Q,Patterson LM,Simmons AJ,Bliton RJ,Betts JW,Lau KS,Magness ST,Goldenring JR,Choi E
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Thu Dec 05 00:00:00 EST 2019
Heterogeneity and dynamics of active Kras-induced dysplastic lineages from mouse corpus stomach.
Nature communications
Min J,Vega PN,Engevik AC,Williams JA,Yang Q,Patterson LM,Simmons AJ,Bliton RJ,Betts JW,Lau KS,Magness ST,Goldenring JR,Choi E
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Thu Dec 05 00:00:00 EST 2019
Yin Yang 1 sustains biosynthetic demands during brain development in a stage-specific manner.
Nature communications
Zurkirchen L,Varum S,Giger S,Klug A,Häusel J,Bossart R,Zemke M,Cantù C,Atak ZK,Zamboni N,Basler K,Sommer L
14-1331-82 was used in Flow Cytometry to demonstrate transcription factor Yin Yang 1 as a stage-dependent regulator of brain development and that biosynthetic demands of neural progenitor cells dynamically change throughout development.
Thu May 16 00:00:00 EDT 2019
Identification of CD24 as a marker of Patched1 deleted medulloblastoma-initiating neural progenitor cells.
PloS one
Robson JP,Remke M,Kool M,Julian E,Korshunov A,Pfister SM,Osborne GW,Taylor MD,Wainwright B,Reynolds BA
14-1331 was used in Flow cytometry/Cell sorting to identify and isolate tumour-initiating cell populations in Patched1 deleted medulloblastoma, and determine if these antibodies can help classify the identity of human medulloblastoma subgroups.
Fri Oct 18 00:00:00 EDT 2019
Mutant ataxin1 disrupts cerebellar development in spinocerebellar ataxia type 1.
The Journal of clinical investigation
Edamakanti CR,Do J,Didonna A,Martina M,Opal P
14-1331 was used in Immunocytochemistry to study whether cerebellar development is altered by mutant ATXN1.
Fri Jun 01 00:00:00 EDT 2018
Mutant ataxin1 disrupts cerebellar development in spinocerebellar ataxia type 1.
The Journal of clinical investigation
Edamakanti CR,Do J,Didonna A,Martina M,Opal P
14-1331 was used in Immunocytochemistry to study whether cerebellar development is altered by mutant ATXN1.
Fri Jun 01 00:00:00 EDT 2018
Mutant ataxin1 disrupts cerebellar development in spinocerebellar ataxia type 1.
The Journal of clinical investigation
Edamakanti CR,Do J,Didonna A,Martina M,Opal P
14-1331 was used in Immunocytochemistry to study whether cerebellar development is altered by mutant ATXN1.
Fri Jun 01 00:00:00 EDT 2018
Mosaic Analysis with Double Markers Reveals Distinct Sequential Functions of Lgl1 in Neural Stem Cells.
Neuron
Beattie R,Postiglione MP,Burnett LE,Laukoter S,Streicher C,Pauler FM,Xiao G,Klezovitch O,Vasioukhin V,Ghashghaei TH,Hippenmeyer S
14-1331 was used in Immunohistochemistry to suggest that NSC-mediated neuron and glia production is regulated through the interplay of sequential Lgl1-dependent global and cell intrinsic mechanisms.
Wed May 03 00:00:00 EDT 2017
Epigenome profiling and editing of neocortical progenitor cells during development.
The EMBO journal
Albert M,Kalebic N,Florio M,Lakshmanaperumal N,Haffner C,Brandl H,Henry I,Huttner WB
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Fri Sep 01 00:00:00 EDT 2017
The C-terminus domain of the hepatitis B virus x protein stimulates the proliferation of mouse foetal hepatic progenitor cells, although it is not required for the formation of spheroids.
International journal of molecular medicine
Yu L,Chen S,Luo N,He S
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Tue Aug 01 00:00:00 EDT 2017
Y-box Protein-1 Regulates the Expression of Collagen I in Hepatic Progenitor Cells via PDGFR-β/ERK/p90RSK Signalling.
Stem cells international
Li F,Ma Z,Liu H,Zhang Q,Cai X,Qu Y,Xu M,Lu L
14-1331 was used in Immunohistochemistry on paraffin embedded tissues-immunofluorescence to identify that Y-box protein-1 participates in the biological adhesion process and extracellular matrix-receptor interactions in hepatic progenitor cells.
Thu Oct 01 00:00:00 EDT 2020
Rescue of mutant rhodopsin traffic by metformin-induced AMPK activation accelerates photoreceptor degeneration.
Human molecular genetics
Athanasiou D,Aguila M,Opefi CA,South K,Bellingham J,Bevilacqua D,Munro PM,Kanuga N,Mackenzie FE,Dubis AM,Georgiadis A,Graca AB,Pearson RA,Ali RR,Sakami S,Palczewski K,Sherman MY,Reeves PJ,Cheetham ME
14133182 was used in immunohistochemistry - frozen section to examine whether the AMPK activator metformin could affect the P23H rhodopsin synthesis and folding to cause retinitis pigmentosa
Sun Jan 15 00:00:00 EST 2017
Isolation of cancer progenitor cells from cancer stem cells in gastric cancer.
Molecular medicine reports
Shang Z,Xu Y,Liang W,Liang K,Hu X,Wang L,Zou Z,Ma Y
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Lymphatic Contribution to the Cellular Niche in Heterotopic Ossification.
Annals of surgery
Loder S,Agarwal S,Sorkin M,Breuler C,Li J,Peterson J,Gardenier J,Hsieh HH,Wang SC,Mehrara BJ,Levi B
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Stem Cells Antigen-1 Enriches for a Cancer Stem Cell-Like Subpopulation in Mouse Gastric Cancer.
Stem cells (Dayton, Ohio)
Park JW,Park JM,Park DM,Kim DY,Kim HK
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Sun May 01 00:00:00 EDT 2016
Characterization of the expression of the pro-metastatic Mena(INV) isoform during breast tumor progression.
Clinical & experimental metastasis
Oudin MJ,Hughes SK,Rohani N,Moufarrej MN,Jones JG,Condeelis JS,Lauffenburger DA,Gertler FB
14-1331 was used in Immunohistochemistry to characterise the expression of the pro-metastatic Mena(INV) isoform during breast tumour progression.
Tue Mar 01 00:00:00 EST 2016
ECM-Dependence of Endothelial Progenitor Cell Features.
Journal of cellular biochemistry
Siavashi V,Nassiri SM,Rahbarghazi R,Vafaei R,Sariri R
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Role of CD133+ cells in tongue squamous carcinomas: Characteristics of 'stemness' in vivo and in vitro.
Oncology letters
Wang K,Zhou XK,Wu M,Kang FW,Wang ZL,Zhu Y
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Mon Aug 01 00:00:00 EDT 2016
Macrophages form functional vascular mimicry channels in vivo.
Scientific reports
Barnett FH,Rosenfeld M,Wood M,Kiosses WB,Usui Y,Marchetti V,Aguilar E,Friedlander M
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Fri Nov 11 00:00:00 EST 2016
Identification of a CD133-CD55- population functions as a fetal common skeletal progenitor.
Scientific reports
Weng L,Hu X,Kumar B,Garcia M,Todorov I,Jung X,Marcucci G,Forman SJ,Chen CC
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Thu Dec 08 00:00:00 EST 2016
IGF-1 contributes to the expansion of melanoma-initiating cells through an epithelial-mesenchymal transition process.
Oncotarget
Le Coz V,Zhu C,Devocelle A,Vazquez A,Boucheix C,Azzi S,Gallerne C,Eid P,Lecourt S,Giron-Michel J
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Tue Dec 13 00:00:00 EST 2016
LIG4 mediates Wnt signalling-induced radioresistance.
Nature communications
Jun S,Jung YS,Suh HN,Wang W,Kim MJ,Oh YS,Lien EM,Shen X,Matsumoto Y,McCrea PD,Li L,Chen J,Park JI
14-1331 was used in Immunohistochemistry to study the molecular mechanism of Wnt signalling-induced radioresistance in CRC and ISCs.
Thu Mar 24 00:00:00 EDT 2016
Single-cell transcriptome analyses reveal signals to activate dormant neural stem cells.
Cell
Luo Y,Coskun V,Liang A,Yu J,Cheng L,Ge W,Shi Z,Zhang K,Li C,Cui Y,Lin H,Luo D,Wang J,Lin C,Dai Z,Zhu H,Zhang J,Liu J,Liu H,deVellis J,Horvath S,Sun YE,Li S
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Thu May 21 00:00:00 EDT 2015
Lovastatin-Induced Phosphatidylinositol-4-Phosphate 5-Kinase Diffusion from Microvilli Stimulates ROMK Channels.
Journal of the American Society of Nephrology : JASN
Liu BC,Yang LL,Lu XY,Song X,Li XC,Chen G,Li Y,Yao X,Humphrey DR,Eaton DC,Shen BZ,Ma HP
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunocytochemistry
Wed Jul 01 00:00:00 EDT 2015
Fate of Prominin-1 Expressing Dermal Papilla Cells during Homeostasis, Wound Healing and Wnt Activation.
The Journal of investigative dermatology
Kaushal GS,Rognoni E,Lichtenberger BM,Driskell RR,Kretzschmar K,Hoste E,Watt FM
14-1331 was used in Immunofluorescence to conclude that Prom1+ dermal papilla cells do not contribute to dermal repair but are capable of regulating dermal papilla size.
Tue Dec 01 00:00:00 EST 2015
Lin41/Trim71 is essential for mouse development and specifically expressed in postnatal ependymal cells of the brain.
Frontiers in cell and developmental biology
Cuevas E,Rybak-Wolf A,Rohde AM,Nguyen DT,Wulczyn FG
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Fri Apr 17 00:00:00 EDT 2015
Glycosylation of rhodopsin is necessary for its stability and incorporation into photoreceptor outer segment discs.
Human molecular genetics
Murray AR,Vuong L,Brobst D,Fliesler SJ,Peachey NS,Gorbatyuk MS,Naash MI,Al-Ubaidi MR
14-1331 was used in Immunohistochemistry to study the role of rhodopsin's glycosylation in vivo.
Fri May 15 00:00:00 EDT 2015
Simvastatin exhibits antiproliferative effects on spheres derived from canine mammary carcinoma cells.
Oncology reports
Torres CG,Olivares A,Stoore C
14-1331 was used in Flow cytometry/Cell sorting to suggest that simvastatin targets canine mammary cancer stem-like cells, supporting its therapeutical application for treating canine mammary cancer.
Fri May 01 00:00:00 EDT 2015
Notch receptor regulation of intestinal stem cell homeostasis and crypt regeneration.
Developmental biology
Carulli AJ,Keeley TM,Demitrack ES,Chung J,Maillard I,Samuelson LC
14-1331 was used in Immunohistochemistry to define the roles of the Notch1 and Notch2 receptors on stem cell function, cell proliferation/differentiation, and repair following injury.
Mon Jun 01 00:00:00 EDT 2015
Impact of diabetes type II and chronic inflammation on pancreatic cancer.
BMC cancer
Zechner D,Radecke T,Amme J,Bürtin F,Albert AC,Partecke LI,Vollmar B
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Western Blot
Fri Feb 13 00:00:00 EST 2015
Impact of diabetes type II and chronic inflammation on pancreatic cancer.
BMC cancer
Zechner D,Radecke T,Amme J,Bürtin F,Albert AC,Partecke LI,Vollmar B
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Western Blot
Fri Feb 13 00:00:00 EST 2015
Mesenchymal-endothelial transition contributes to cardiac neovascularization.
Nature
Ubil E,Duan J,Pillai IC,Rosa-Garrido M,Wu Y,Bargiacchi F,Lu Y,Stanbouly S,Huang J,Rojas M,Vondriska TM,Stefani E,Deb A
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Thu Oct 30 00:00:00 EDT 2014
Mesodermal mesenchymal cells give rise to myofibroblasts, but not epithelial cells, in mouse liver injury.
Hepatology (Baltimore, Md.)
Lua I,James D,Wang J,Wang KS,Asahina K
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Tue Jul 01 00:00:00 EDT 2014
Rim formation is not a prerequisite for distribution of cone photoreceptor outer segment proteins.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Conley SM,Al-Ubaidi MR,Han Z,Naash MI
14-1331 was used in Immunofluorescence to suggest that the unique folded lamella architecture of the cone OS may maximize density or proximity of phototransduction proteins.
Fri Aug 01 00:00:00 EDT 2014
Aberrant Proliferation of Differentiating Alveolar Cells Induces Hyperplasia in Resting Mammary Glands of SV40-TAg Transgenic Mice.
Frontiers in oncology
Quante T,Wegwitz F,Abe J,Rossi A,Deppert W,Bohn W
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Mon Jul 14 00:00:00 EDT 2014
Lgr5 Marks Post-Mitotic, Lineage Restricted Cerebellar Granule Neurons during Postnatal Development.
PloS one
Miller TE,Wang J,Sukhdeo K,Horbinski C,Tesar PJ,Wechsler-Reya RJ,Rich JN
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Fri Oct 13 00:00:00 EDT 2017
Prominin-1 (CD133) defines both stem and non-stem cell populations in CNS development and gliomas.
PloS one
Holmberg Olausson K,Maire CL,Haidar S,Ling J,Learner E,Nistér M,Ligon KL
14-1331 was used in Immunohistochemistry on frozen tissues to explore the normal and cancer expression pattern and lineage associations of Prominin-1 in human and mouse brain.
Thu May 14 00:00:00 EDT 2015
Prominin-1 (CD133) defines both stem and non-stem cell populations in CNS development and gliomas.
PloS one
Holmberg Olausson K,Maire CL,Haidar S,Ling J,Learner E,Nistér M,Ligon KL
14-1331 was used in Immunohistochemistry on frozen tissues to explore the normal and cancer expression pattern and lineage associations of Prominin-1 in human and mouse brain.
Thu May 14 00:00:00 EDT 2015
Cardiac mouse lymphatics: developmental and anatomical update.
Anatomical record (Hoboken, N.J. : 2007)
Flaht-Zabost A,Gula G,Ciszek B,Czarnowska E,Jankowska-Steifer E,Madej M,Niderla-Bielińska J,Radomska-Leśniewska D,Ratajska A
14-1331-82 was used in Immunohistochemistry to report that murine cardiac lymphatic plexus development that is limited prenatally only to the subepicardial area, postnatally proceeds from the subepicardium toward the myocardial wall with the base-to-apex gradient; this plexus eventually reaches the outer half of the myocardium with a predominant location around branches of coronary arteries and veins.
Sun Jun 01 00:00:00 EDT 2014
Usp16 contributes to somatic stem-cell defects in Down's syndrome.
Nature
Adorno M,Sikandar S,Mitra SS,Kuo A,Nicolis Di Robilant B,Haro-Acosta V,Ouadah Y,Quarta M,Rodriguez J,Qian D,Reddy VM,Cheshier S,Garner CC,Clarke MF
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Thu Sep 19 00:00:00 EDT 2013
Sox2 modulates the function of two distinct cell lineages in mouse skin.
Developmental biology
Lesko MH,Driskell RR,Kretzschmar K,Goldie SJ,Watt FM
14-1331 was used in Flow cytometry/Cell sorting to investigate how Sox2 modulates the function of two distinct cell lineages in mouse skin.
Tue Oct 01 00:00:00 EDT 2013
Notch signaling maintains neural rosette polarity.
PloS one
Main H,Radenkovic J,Jin SB,Lendahl U,Andersson ER
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Tue Dec 17 00:00:00 EST 2013
Notch signaling maintains neural rosette polarity.
PloS one
Main H,Radenkovic J,Jin SB,Lendahl U,Andersson ER
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Tue Dec 17 00:00:00 EST 2013
Notch signaling maintains neural rosette polarity.
PloS one
Main H,Radenkovic J,Jin SB,Lendahl U,Andersson ER
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Tue Dec 17 00:00:00 EST 2013
Notch signaling maintains neural rosette polarity.
PloS one
Main H,Radenkovic J,Jin SB,Lendahl U,Andersson ER
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Tue Dec 17 00:00:00 EST 2013
In vitro transformation of mouse testis cells by oncogene transfection.
Biology of reproduction
Morimoto H,Lee J,Tanaka T,Ishii K,Toyokuni S,Kanatsu-Shinohara M,Shinohara T
14-1331 was used in Flow cytometry/Cell sorting to develop a novel strategy to transform mouse testis cells in vitro for the study of germ cell tumours.
Tue May 01 00:00:00 EDT 2012
Partial characterization of the Sox2+ cell population in an adult murine model of digit amputation.
Tissue engineering. Part A
Agrawal V,Siu BF,Chao H,Hirschi KK,Raborn E,Johnson SA,Tottey S,Hurley KB,Medberry CJ,Badylak SF
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2012
Biciliated ependymal cell proliferation contributes to spinal cord growth.
The Journal of comparative neurology
Alfaro-Cervello C,Soriano-Navarro M,Mirzadeh Z,Alvarez-Buylla A,Garcia-Verdugo JM
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Mon Oct 15 00:00:00 EDT 2012
Wnt/beta-catenin signaling in embryonic stem cell converted tumor cells.
Journal of translational medicine
Peng X,Liu T,Wang Y,Yan Q,Jin H,Li L,Qian Q,Wu M
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Thu Sep 20 00:00:00 EDT 2012
Chorionic gonadotropin and its receptor are both expressed in human retina, possible implications in normal and pathological conditions.
PloS one
Dukic-Stefanovic S,Walther J,Wosch S,Zimmermann G,Wiedemann P,Alexander H,Claudepierre T
14-1331 was used in Immunocytochemistry to demonstrate that human retina is producing this gonadotropin that acts as a neuroactive molecule.
Tue Jun 11 00:00:00 EDT 2013
Midbody accumulation through evasion of autophagy contributes to cellular reprogramming and tumorigenicity.
Nature cell biology
Kuo TC,Chen CT,Baron D,Onder TT,Loewer S,Almeida S,Weismann CM,Xu P,Houghton JM,Gao FB,Daley GQ,Doxsey S
14-1331 was used in Immunofluorescence to investigate the fate of midbodies.
Sun Sep 11 00:00:00 EDT 2011
The Notch ligand Jagged2 promotes lung adenocarcinoma metastasis through a miR-200-dependent pathway in mice.
The Journal of clinical investigation
Yang Y,Ahn YH,Gibbons DL,Zang Y,Lin W,Thilaganathan N,Alvarez CA,Moreira DC,Creighton CJ,Gregory PA,Goodall GJ,Kurie JM
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Western Blot
Fri Apr 01 00:00:00 EDT 2011
The Notch ligand Jagged2 promotes lung adenocarcinoma metastasis through a miR-200-dependent pathway in mice.
The Journal of clinical investigation
Yang Y,Ahn YH,Gibbons DL,Zang Y,Lin W,Thilaganathan N,Alvarez CA,Moreira DC,Creighton CJ,Gregory PA,Goodall GJ,Kurie JM
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Western Blot
Fri Apr 01 00:00:00 EDT 2011
Increased re-entry into cell cycle mitigates age-related neurogenic decline in the murine subventricular zone.
Stem cells (Dayton, Ohio)
Stoll EA,Habibi BA,Mikheev AM,Lasiene J,Massey SC,Swanson KR,Rostomily RC,Horner PJ
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Thu Dec 01 00:00:00 EST 2011
Stem cell marker prominin-1 regulates branching morphogenesis, but not regenerative capacity, in the mammary gland.
Developmental dynamics : an official publication of the American Association of Anatomists
Anderson LH,Boulanger CA,Smith GH,Carmeliet P,Watson CJ
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Tue Mar 01 00:00:00 EST 2011
Stem cell marker prominin-1 regulates branching morphogenesis, but not regenerative capacity, in the mammary gland.
Developmental dynamics : an official publication of the American Association of Anatomists
Anderson LH,Boulanger CA,Smith GH,Carmeliet P,Watson CJ
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Tue Mar 01 00:00:00 EST 2011
CXCR4 signaling regulates metastasis of chemoresistant melanoma cells by a lymphatic metastatic niche.
Cancer research
Kim M,Koh YJ,Kim KE,Koh BI,Nam DH,Alitalo K,Kim I,Koh GY
14-1331 was used in Flow cytometry/Cell sorting to investigate the mechanisms responsible for the aggressive characteristics of malignant melanomas, showing that CXCR4 signaling regulates metastasis.
Wed Dec 15 00:00:00 EST 2010
Mouse lung contains endothelial progenitors with high capacity to form blood and lymphatic vessels.
BMC cell biology
Schniedermann J,Rennecke M,Buttler K,Richter G,Städtler AM,Norgall S,Badar M,Barleon B,May T,Wilting J,Weich HA
14-1331 was used in Flow cytometry/Cell sorting to evaluate the differentiation capacity of resident lung endothelial progenitor cells from mice.
Thu Jul 01 00:00:00 EDT 2010
Mouse tissues express multiple splice variants of prominin-1.
PloS one
Kemper K,Tol MJ,Medema JP
14-1331 was used in Western Blotting to analyze the expression of the prominin-1 splice variants on mRNA level in several mouse tissues and found a broad tissue expression of the majority of SVs.
Fri Aug 20 00:00:00 EDT 2010
Prominin 1 marks intestinal stem cells that are susceptible to neoplastic transformation.
Nature
Zhu L,Gibson P,Currle DS,Tong Y,Richardson RJ,Bayazitov IT,Poppleton H,Zakharenko S,Ellison DW,Gilbertson RJ
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunohistochemistry
Thu Jan 29 00:00:00 EST 2009
Prominin-1/CD133 marks stem cells and early progenitors in mouse small intestine.
Gastroenterology
Snippert HJ,van Es JH,van den Born M,Begthel H,Stange DE,Barker N,Clevers H
14-1331 was used in Immunofluorescence to show that prom-1 marks intestinal stem cells, and transit-amplifying progenitors, thus is not a specific marker for Lgr5(+) intestinal stem cells.
Mon Jun 01 00:00:00 EDT 2009
FoxJ1-dependent gene expression is required for differentiation of radial glia into ependymal cells and a subset of astrocytes in the postnatal brain.
Development (Cambridge, England)
Jacquet BV,Salinas-Mondragon R,Liang H,Therit B,Buie JD,Dykstra M,Campbell K,Ostrowski LE,Brody SL,Ghashghaei HT
14-1331 was used in Blocking experiments to suggest that time and cell-specific FoxJ1 expression in the brain regulates the differentiation of ependymal cells and a subset of astrocytes.
Tue Dec 01 00:00:00 EST 2009
Loss of the cholesterol-binding protein prominin-1/CD133 causes disk dysmorphogenesis and photoreceptor degeneration.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Zacchigna S,Oh H,Wilsch-Bräuninger M,Missol-Kolka E,Jászai J,Jansen S,Tanimoto N,Tonagel F,Seeliger M,Huttner WB,Corbeil D,Dewerchin M,Vinckier S,Moons L,Carmeliet P
14-1331 was used in Western Blotting to show that loss of Prom-1 results in progressive degeneration of mature photoreceptors with complete loss of vision.
Wed Feb 18 00:00:00 EST 2009
Loss of the cholesterol-binding protein prominin-1/CD133 causes disk dysmorphogenesis and photoreceptor degeneration.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Zacchigna S,Oh H,Wilsch-Bräuninger M,Missol-Kolka E,Jászai J,Jansen S,Tanimoto N,Tonagel F,Seeliger M,Huttner WB,Corbeil D,Dewerchin M,Vinckier S,Moons L,Carmeliet P
14-1331 was used in Western Blotting to show that loss of Prom-1 results in progressive degeneration of mature photoreceptors with complete loss of vision.
Wed Feb 18 00:00:00 EST 2009
CD133 (Prominin) negative human neural stem cells are clonogenic and tripotent.
PloS one
Sun Y,Kong W,Falk A,Hu J,Zhou L,Pollard S,Smith A
14-1331 was used in Immunofluorescence to assess the assumption that CD133 is expressed constitutively in neural precursor cells.
Fri Sep 25 00:00:00 EDT 2009
Neural stem cells confer unique pinwheel architecture to the ventricular surface in neurogenic regions of the adult brain.
Cell stem cell
Mirzadeh Z,Merkle FT,Soriano-Navarro M,Garcia-Verdugo JM,Alvarez-Buylla A
14-1331 was used in Immunohistochemistry to reveal that adult neural stem cells retain fundamental epithelial properties, including apical and basal compartmentalization.
Thu Sep 11 00:00:00 EDT 2008
CD133 expression is not restricted to stem cells, and both CD133+ and CD133- metastatic colon cancer cells initiate tumors.
The Journal of clinical investigation
Shmelkov SV,Butler JM,Hooper AT,Hormigo A,Kushner J,Milde T,St Clair R,Baljevic M,White I,Jin DK,Chadburn A,Murphy AJ,Valenzuela DM,Gale NW,Thurston G,Yancopoulos GD,D'Angelica M,Kemeny N,Lyden D,Rafii S
14-1331 was used in Immunohistochemistry on frozen tissues to temporally and spatially track the expression of CD133 in adult mice and during tumourigenesis using a knockin lacZ reporter mouse.
Sun Jun 01 00:00:00 EDT 2008
Spinal cord injury reveals multilineage differentiation of ependymal cells.
PLoS biology
Meletis K,Barnabé-Heider F,Carlén M,Evergren E,Tomilin N,Shupliakov O,Frisén J
14-1331 was used in Immunohistochemistry to investigate the potential for an alternative treatment to cell transplantation for promoting functional recovery following spinal cord injury.
Tue Jul 22 00:00:00 EDT 2008
A genome-wide expression analysis identifies a network of EpCAM-induced cell cycle regulators.
British journal of cancer
Maaser K,Borlak J
14-1331 was used in Western Blotting to suggest that EpCAM is involved in signal transduction triggering several intracellular signalling pathways.
Tue Nov 18 00:00:00 EST 2008
CD133 is not present on neurogenic astrocytes in the adult subventricular zone, but on embryonic neural stem cells, ependymal cells, and glioblastoma cells.
Cancer research
Pfenninger CV,Roschupkina T,Hertwig F,Kottwitz D,Englund E,Bengzon J,Jacobsen SE,Nuber UA
Fri Jun 15 00:00:00 EDT 2007
Conserved markers of fetal pancreatic epithelium permit prospective isolation of islet progenitor cells by FACS.
Proceedings of the National Academy of Sciences of the United States of America
Sugiyama T,Rodriguez RT,McLean GW,Kim SK
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Tue Jan 02 00:00:00 EST 2007
Conserved markers of fetal pancreatic epithelium permit prospective isolation of islet progenitor cells by FACS.
Proceedings of the National Academy of Sciences of the United States of America
Sugiyama T,Rodriguez RT,McLean GW,Kim SK
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Tue Jan 02 00:00:00 EST 2007
Conserved markers of fetal pancreatic epithelium permit prospective isolation of islet progenitor cells by FACS.
Proceedings of the National Academy of Sciences of the United States of America
Sugiyama T,Rodriguez RT,McLean GW,Kim SK
Published figure using CD133 (Prominin-1) monoclonal antibody (Product # 14-1331-82) in Immunofluorescence
Tue Jan 02 00:00:00 EST 2007
Isolation of murine hair-inducing cells using the cell surface marker prominin-1/CD133.
The Journal of investigative dermatology
Ito Y,Hamazaki TS,Ohnuma K,Tamaki K,Asashima M,Okochi H
14-1331 was used in Flow cytometry/Cell sorting to propose that the CD133 is a novel surface marker useful for collecting DP cells in the anagen stage.
Tue May 01 00:00:00 EDT 2007