Product References
Decoding the pathogenesis of Diamond-Blackfan anemia using single-cell RNA-seq.
Cell discovery
Wang B,Wang C,Wan Y,Gao J,Ma Y,Zhang Y,Tong J,Zhang Y,Liu J,Chang L,Xu C,Shen B,Chen Y,Jiang E,Kurita R,Nakamura Y,Lim KC,Engel JD,Zhou J,Cheng T,Zhu X,Zhu P,Shi L
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue May 10 00:00:00 EDT 2022
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Thu Mar 03 00:00:00 EST 2022
Increased TNF-α Initiates Cytoplasmic Vacuolization in Whole Blood Coculture with Dengue Virus.
Journal of immunology research
Satria RD,Huang TW,Jhan MK,Shen TJ,Tseng PC,Wang YT,Yang ZY,Hsing CH,Lin CF
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Mon Dec 06 00:00:00 EST 2021
SARS-CoV-2 infection paralyzes cytotoxic and metabolic functions of the immune cells.
Heliyon
Singh Y,Trautwein C,Fendel R,Krickeberg N,Berezhnoy G,Bissinger R,Ossowski S,Salker MS,Casadei N,Riess O
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Th2 Biased Immunity With Altered B Cell Profiles in Circulation of Patients With Sporotrichosis Caused by Sporothrix globosa.
Frontiers in immunology
Zu J,Yao L,Song Y,Cui Y,Guan M,Chen R,Zhen Y,Li S
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Mon May 31 00:00:00 EDT 2021
Human T Cells Expressing a CD19 CAR-T Receptor Provide Insights into Mechanisms of Human CD19-Positive β Cell Destruction.
Cell reports. Medicine
Ma H,Jeppesen JF,Jaenisch R
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue Sep 22 00:00:00 EDT 2020
Peli1 negatively regulates noncanonical NF-κB signaling to restrain systemic lupus erythematosus.
Nature communications
Liu J,Huang X,Hao S,Wang Y,Liu M,Xu J,Zhang X,Yu T,Gan S,Dai D,Luo X,Lu Q,Mao C,Zhang Y,Shen N,Li B,Huang M,Zhu X,Jin J,Cheng X,Sun SC,Xiao Y
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Mon Mar 19 00:00:00 EDT 2018
A Human IPS Model Implicates Embryonic B-Myeloid Fate Restriction as Developmental Susceptibility to B Acute Lymphoblastic Leukemia-Associated ETV6-RUNX1.
Developmental cell
Böiers C,Richardson SE,Laycock E,Zriwil A,Turati VA,Brown J,Wray JP,Wang D,James C,Herrero J,Sitnicka E,Karlsson S,Smith AJH,Jacobsen SEW,Enver T
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Mon Feb 05 00:00:00 EST 2018
Human Secretory IgM Emerges from Plasma Cells Clonally Related to Gut Memory B Cells and Targets Highly Diverse Commensals.
Immunity
Magri G,Comerma L,Pybus M,Sintes J,Lligé D,Segura-Garzón D,Bascones S,Yeste A,Grasset EK,Gutzeit C,Uzzan M,Ramanujam M,van Zelm MC,Albero-González R,Vazquez I,Iglesias M,Serrano S,Márquez L,Mercade E,Mehandru S,Cerutti A
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue Jul 18 00:00:00 EDT 2017
Antibodies to P-selectin glycoprotein ligand-1 block dendritic cell-mediated enterovirus 71 transmission and prevent virus-induced cells death.
Virulence
Ren XX,Li C,Xiong SD,Huang Z,Wang JH,Wang HB
12-0199 was used in Flow cytometry/Cell sorting to demonstrate that PSGL-1 present on the cell surfaces of monocyte-derived dendritic cells mediates enterovirus 71 binding and facilitates further infection.
Thu Oct 06 00:00:00 EDT 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
CD19 CAR immune pressure induces B-precursor acute lymphoblastic leukaemia lineage switch exposing inherent leukaemic plasticity.
Nature communications
Jacoby E,Nguyen SM,Fountaine TJ,Welp K,Gryder B,Qin H,Yang Y,Chien CD,Seif AE,Lei H,Song YK,Khan J,Lee DW,Mackall CL,Gardner RA,Jensen MC,Shern JF,Fry TJ
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Wed Jul 27 00:00:00 EDT 2016
Medial HOXA genes demarcate haematopoietic stem cell fate during human development.
Nature cell biology
Dou DR,Calvanese V,Sierra MI,Nguyen AT,Minasian A,Saarikoski P,Sasidharan R,Ramirez CM,Zack JA,Crooks GM,Galic Z,Mikkola HK
12019980 was used in flow cytometry to study how retinoic acid-induced HOXA gene expression controls hematopoietic stem/progenitor cells fate, identify, and function
Wed Jun 01 00:00:00 EDT 2016
Development of depressive symptoms post hip fracture is associated with altered immunosuppressive phenotype in regulatory T and B lymphocytes.
Biogerontology
Duggal NA,Upton J,Phillips AC,Lord JM
12-0199 was used in Flow cytometry/Cell sorting to investigate whether hip fracture and depressive symptoms had additive effects upon the aged immune system.
Mon Feb 01 00:00:00 EST 2016
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue Jan 19 00:00:00 EST 2016
The BAFF receptor TACI controls IL-10 production by regulatory B cells and CLL B cells.
Leukemia
Saulep-Easton D,Vincent FB,Quah PS,Wei A,Ting SB,Croce CM,Tam C,Mackay F
12-0199 was used in Flow cytometry/Cell sorting to uncover a major targetable pathway important for the generation of regulatory B cells that is detrimental to immunity in chronic lymphocytic leukemia.
Fri Jan 01 00:00:00 EST 2016
Phenotypic and functional characterization of macrophages with therapeutic potential generated from human cirrhotic monocytes in a cohort study.
Cytotherapy
Moore JK,Mackinnon AC,Wojtacha D,Pope C,Fraser AR,Burgoyne P,Bailey L,Pass C,Atkinson A,Mcgowan NW,Manson L,Turner ML,Campbell JD,Forbes SJ
12-0199 was used in Flow cytometry/Cell sorting to determine whether chronic inflammation affects precursor number or the phenotype of monocyte-derived macrophages.
Sun Nov 01 00:00:00 EDT 2015
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue Sep 15 00:00:00 EDT 2015
Interleukin-22 is increased in multiple sclerosis patients and targets astrocytes.
Journal of neuroinflammation
Perriard G,Mathias A,Enz L,Canales M,Schluep M,Gentner M,Schaeren-Wiemers N,Du Pasquier RA
12-0199 was used in Flow cytometry/Cell sorting to characterise the role of IL-22 in the context of multiple sclerosis.
Tue Jun 16 00:00:00 EDT 2015
Induction of type I and type III interferons by Borrelia burgdorferi correlates with pathogenesis and requires linear plasmid 36.
PloS one
Krupna-Gaylord MA,Liveris D,Love AC,Wormser GP,Schwartz I,Petzke MM
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Thu Mar 12 00:00:00 EDT 2015
Reprogramming human endothelial cells to haematopoietic cells requires vascular induction.
Nature
Sandler VM,Lis R,Liu Y,Kedem A,James D,Elemento O,Butler JM,Scandura JM,Rafii S
12-0199 was used in Flow cytometry/Cell sorting to investigate the role of inductive cues from the vascular niche in coordinating and sustaining haematopoietic specification.
Thu Jul 17 00:00:00 EDT 2014
Reprogramming human endothelial cells to haematopoietic cells requires vascular induction.
Nature
Sandler VM,Lis R,Liu Y,Kedem A,James D,Elemento O,Butler JM,Scandura JM,Rafii S
12-0199 was used in Flow cytometry/Cell sorting to investigate the role of inductive cues from the vascular niche in coordinating and sustaining haematopoietic specification.
Thu Jul 17 00:00:00 EDT 2014
Aberrant expression of SALL4 in acute B cell lymphoblastic leukemia: mechanism, function, and implication for a potential novel therapeutic target.
Experimental hematology
Ueno S,Lu J,He J,Li A,Zhang X,Ritz J,Silberstein LE,Chai L
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue Apr 01 00:00:00 EDT 2014
Comparative study of the cytokine/chemokine response in children with differing disease severity in enterovirus 71-induced hand, foot, and mouth disease.
PloS one
Zhang Y,Liu H,Wang L,Yang F,Hu Y,Ren X,Li G,Yang Y,Yu Y,Sun S,Li Y,Chen X,Li X,Jin Q
Published figure using CD19 monoclonal antibody (Product # 12-0199-80) in Flow Cytometry
Tue Jan 21 00:00:00 EST 2014
A pathobiological role of the insulin receptor in chronic lymphocytic leukemia.
Clinical cancer research : an official journal of the American Association for Cancer Research
Saiya-Cork K,Collins R,Parkin B,Ouillette P,Kuizon E,Kujawski L,Erba H,Campagnaro E,Shedden K,Kaminski M,Malek SN
12-0199 was used in Flow cytometry/Cell sorting to characterise the genes that are disrupted in chronic lymphocytic leukemia in conjunction with the disease modifying chromosomal deletion 11q, showing that insulin receptor expression is increased in a large proportion of the cases.
Sun May 01 00:00:00 EDT 2011
Effects of NOD-like receptors in human B lymphocytes and crosstalk between NOD1/NOD2 and Toll-like receptors.
Journal of leukocyte biology
Petterson T,Jendholm J,Månsson A,Bjartell A,Riesbeck K,Cardell LO
12-0199 was used in Flow cytometry/Cell sorting to investigate how NLRs represent a new pathway for B cell activation and a potentially important host defense system against bacterial infections.
Tue Feb 01 00:00:00 EST 2011
Differential IL-7 responses in developing human thymocytes.
Human immunology
Marino JH,Tan C,Taylor AA,Bentley C,Van De Wiele CJ,Ranne R,Paliotta M,Broughan TA,Teague TK
12-0199 was used in Flow cytometry/Cell sorting to assess IL-7 receptor expression and IL-7 signalling in human thymocytes.
Thu Apr 01 00:00:00 EDT 2010
Dendritic cells derived from hemozoin-loaded monocytes display a partial maturation phenotype that promotes HIV-1 trans-infection of CD4+ T cells and virus replication.
Journal of immunology (Baltimore, Md. : 1950)
Diou J,Tardif MR,Barat C,Tremblay MJ
12-0199 was used in Flow cytometry/Cell sorting to investigate the effect of malarial pigment hemozoin on HIV-1 transmission, showing that hemozoid-loaded monocytes promote HIV-1 trans-infection and replication.
Mon Mar 15 00:00:00 EDT 2010
B cells and monocytes from patients with active multiple sclerosis exhibit increased surface expression of both HERV-H Env and HERV-W Env, accompanied by increased seroreactivity.
Retrovirology
Brudek T,Christensen T,Aagaard L,Petersen T,Hansen HJ,Møller-Larsen A
12-0199 was used in Flow cytometry/Cell sorting to study the increased surface expression of both HERV-H Env and HERV-W Env, accompanied by increased seroreactivity, in B cells and monocytes from patients with active multiple sclerosis.
Mon Nov 16 00:00:00 EST 2009
Sepsis-induced human lymphocyte apoptosis and cytokine production in "humanized" mice.
Journal of leukocyte biology
Unsinger J,McDonough JS,Shultz LD,Ferguson TA,Hotchkiss RS
12-0199 was used in Flow cytometry/Cell sorting to study sepsis-induced human lymphocyte apoptosis and cytokine production in "humanized" mice.
Sat Aug 01 00:00:00 EDT 2009