Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Description: The J10MBS monoclonal antibody reacts with human interleukin 17 receptor A (IL-17RA, IL-17R). IL-17RA is a type I membrane protein of approximately 96 kDa (866 amino acids) and is involved in inflammatory responses to IL-17A, IL-17F and IL-17AF heterodimer. It forms heterodimeric complexes with several other members of its receptor family, including IL-17RC for IL-17A and IL-17F signaling and IL-17RC for IL-17E (IL-25) signaling. IL-17RA is ubiquitously expressed, but in leukocytes its expression is elevated on granulocytes and monocytes.
Applications Reported: This J10MBS antibody has been reported for use in flow cytometric analysis.
Applications Tested: This J10MBS antibody has been pre-titrated and tested by flow cytometric analysis of human lysed whole blood. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Interleukin 17A (IL-17A) is a proinflammatory cytokine secreted by activated T-lymphocytes. It is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils. IL-17RA is a ubiquitous type I membrane glycoprotein that binds with low affinity to IL-17A and IL-17F. IL-17A and its receptor play a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. Defects in IL-17RA are the cause of familial candidiasis type 5 (CANDF5) which is a rare disorder with altered immune responses and impaired clearance of fungal infections.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CD217; CDw217; IL-17 receptor A; Interleukin-17 receptor A; MGC10262
Gene Aliases: CANDF5; CD217; CDw217; hIL-17R; IL-17RA; IL17R; IL17RA
UniProt ID: (Human) Q96F46
Entrez Gene ID: (Human) 23765
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support