Product References
The immunoregulatory function of peripheral blood CD71+ erythroid cells in systemic-onset juvenile idiopathic arthritis.
Scientific reports
Kanemasa H,Ishimura M,Eguchi K,Tanaka T,Nanishi E,Shiraishi A,Goto M,Motomura Y,Ohga S
17-9987-42 was used in Flow Cytometry to conclude that circulating CECs may control excessive inflammation via the immunoregulatory pathways, partly involving arginase-2.
Tue Jul 13 00:00:00 EDT 2021
Polycomb Factor PHF19 Controls Cell Growth and Differentiation Toward Erythroid Pathway in Chronic Myeloid Leukemia Cells.
Frontiers in cell and developmental biology
García-Montolio M,Ballaré C,Blanco E,Gutiérrez A,Aranda S,Gómez A,Kok CH,Yeung DT,Hughes TP,Vizán P,Di Croce L
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Tue May 18 00:00:00 EDT 2021
Extracellular vesicles are associated with C-reactive protein in sepsis.
Scientific reports
Fendl B,Weiss R,Eichhorn T,Linsberger I,Afonyushkin T,Puhm F,Binder CJ,Fischer MB,Weber V
17-9987 was used in Flow cytometry/Cell sorting to provide evidence that CRP+ EVs exhibit pro-inflammatory characteristics and can contribute to the spreading of inflammation throughout the circulation on top of their pro-coagulant activity.
Fri Mar 26 00:00:00 EDT 2021
Iron control of erythroid microtubule cytoskeleton as a potential target in treatment of iron-restricted anemia.
Nature communications
Goldfarb AN,Freeman KC,Sahu RK,Elagib KE,Holy M,Arneja A,Polanowska-Grabowska R,Gru AA,White Z,Khalil S,Kerins MJ,Ooi A,Leitinger N,Luckey CJ,Delehanty LL
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Fri Mar 12 00:00:00 EST 2021
Common variants in signaling transcription-factor-binding sites drive phenotypic variability in red blood cell traits.
Nature genetics
Choudhuri A,Trompouki E,Abraham BJ,Colli LM,Kock KH,Mallard W,Yang ML,Vinjamur DS,Ghamari A,Sporrij A,Hoi K,Hummel B,Boatman S,Chan V,Tseng S,Nandakumar SK,Yang S,Lichtig A,Superdock M,Grimes SN,Bowman TV,Zhou Y,Takahashi S,Joehanes R,Cantor AB,Bauer DE,Ganesh SK,Rinn J,Albert PS,Bulyk ML,Chanock SJ,Young RA,Zon LI
17-9987 was used in Flow cytometry/Cell sorting to propose that the majority of the RBC-trait-associated variants that reside on transcription-factor-binding sequences fall in STF target sequences, suggesting that the phenotypic variation of RBC traits could stem from altered responsiveness to extracellular stimuli.
Tue Dec 01 00:00:00 EST 2020
The antifibrotic adipose-derived stromal cell: Grafted fat enriched with CD74+ adipose-derived stromal cells reduces chronic radiation-induced skin fibrosis.
Stem cells translational medicine
Borrelli MR,Patel RA,Adem S,Diaz Deleon NM,Shen AH,Sokol J,Yen S,Chang EY,Nazerali R,Nguyen D,Momeni A,Wang KC,Longaker MT,Wan DC
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2020
Genome-Wide Transcriptional Regulation of the Long Non-coding RNA Steroid Receptor RNA Activator in Human Erythroblasts.
Frontiers in genetics
Sawaengdee W,Cui K,Zhao K,Hongeng S,Fucharoen S,Wongtrakoongate P
17-9987 was used in Flow cytometry/Cell sorting to establish that the lncRNA SRA occupies chromatin, and promotes transcription of erythroid genes, therefore facilitating human erythroid transcriptional program.
Mon Sep 28 00:00:00 EDT 2020
Proteosomal degradation of NSD2 by BRCA1 promotes leukemia cell differentiation.
Communications biology
Park JW,Kang JY,Hahm JY,Kim HJ,Seo SB
17-9987 was used in Flow cytometry/Cell sorting to study the regulation of NSD2 stability by BRCA1-mediated ubiquitination as a potential therapeutic target process in multiple myeloma.
Fri Aug 21 00:00:00 EDT 2020
Transcriptional States and Chromatin Accessibility Underlying Human Erythropoiesis.
Cell reports
Ludwig LS,Lareau CA,Bao EL,Nandakumar SK,Muus C,Ulirsch JC,Chowdhary K,Buenrostro JD,Mohandas N,An X,Aryee MJ,Regev A,Sankaran VG
17-9987-42 was used in Flow Cytometry to reveal stage-specific transcriptional states and chromatin accessibility during various stages of erythropoiesis.
Tue Jun 11 00:00:00 EDT 2019
Gene-centric functional dissection of human genetic variation uncovers regulators of hematopoiesis.
eLife
Nandakumar SK,McFarland SK,Mateyka LM,Lareau CA,Ulirsch JC,Ludwig LS,Agarwal G,Engreitz JM,Przychodzen B,McConkey M,Cowley GS,Doench JG,Maciejewski JP,Ebert BL,Root DE,Sankaran VG
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Thu May 09 00:00:00 EDT 2019
EDAG promotes the expansion and survival of human CD34+ cells.
PloS one
Zhao K,Zheng WW,Dong XM,Yin RH,Gao R,Li X,Liu JF,Zhan YQ,Yu M,Chen H,Ge CH,Ning HM,Yang XM,Li CY
17-9987 was used in Flow cytometry/Cell sorting to provide novel insight into EDAG in regulation of expansion and survival of human hematopoietic stem/progenitor cells.
Tue Feb 06 00:00:00 EST 2018
Functional Selectivity in Cytokine Signaling Revealed Through a Pathogenic EPO Mutation.
Cell
Kim AR,Ulirsch JC,Wilmes S,Unal E,Moraga I,Karakukcu M,Yuan D,Kazerounian S,Abdulhay NJ,King DS,Gupta N,Gabriel SB,Lander ES,Patiroglu T,Ozcan A,Ozdemir MA,Garcia KC,Piehler J,Gazda HT,Klein DE,Sankaran VG
17-9987 was used in Flow cytometry/Cell sorting to demonstrate how variation in a single cytokine can lead to biased downstream signaling and can thereby cause human disease.
Thu Mar 09 00:00:00 EST 2017
The Impact of Lipoprotein-Associated Oxidative Stress on Cell-Specific Microvesicle Release in Patients with Familial Hypercholesterolemia.
Oxidative medicine and cellular longevity
Nielsen MH,Irvine H,Vedel S,Raungaard B,Beck-Nielsen H,Handberg A
17-9987 was used in Flow cytometry/Cell sorting to measure cell-specific microvesicles in familial hypercholesterolemia patients with and without Achilles tendon xanthomas, and their association with atherogenic lipid profile.
Tue Dec 13 00:00:00 EST 2016
Platelets confound the measurement of extracellular miRNA in archived plasma.
Scientific reports
Mitchell AJ,Gray WD,Hayek SS,Ko YA,Thomas S,Rooney K,Awad M,Roback JD,Quyyumi A,Searles CD
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Tue Sep 13 00:00:00 EDT 2016
LECT2 drives haematopoietic stem cell expansion and mobilization via regulating the macrophages and osteolineage cells.
Nature communications
Lu XJ,Chen Q,Rong YJ,Yang GJ,Li CH,Xu NY,Yu CH,Wang HY,Zhang S,Shi YH,Chen J
17-9987 was used in Flow cytometry/Cell sorting to investigate the mechanisms by which cytokines in the blood affect HSC homeostasis, showing that LECT2 drives HSC expansion and mobilisation.
Tue Sep 06 00:00:00 EDT 2016
Genetic Variability Overrides the Impact of Parental Cell Type and Determines iPSC Differentiation Potential.
Stem cell reports
Kyttälä A,Moraghebi R,Valensisi C,Kettunen J,Andrus C,Pasumarthy KK,Nakanishi M,Nishimura K,Ohtaka M,Weltner J,Van Handel B,Parkkonen O,Sinisalo J,Jalanko A,Hawkins RD,Woods NB,Otonkoski T,Trokovic R
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Tue Feb 09 00:00:00 EST 2016
Targeted Application of Human Genetic Variation Can Improve Red Blood Cell Production from Stem Cells.
Cell stem cell
Giani FC,Fiorini C,Wakabayashi A,Ludwig LS,Salem RM,Jobaliya CD,Regan SN,Ulirsch JC,Liang G,Steinberg-Shemer O,Guo MH,Esko T,Tong W,Brugnara C,Hirschhorn JN,Weiss MJ,Zon LI,Chou ST,French DL,Musunuru K,Sankaran VG
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Thu Jan 07 00:00:00 EST 2016
PPAR-α and glucocorticoid receptor synergize to promote erythroid progenitor self-renewal.
Nature
Lee HY,Gao X,Barrasa MI,Li H,Elmes RR,Peters LL,Lodish HF
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Thu Jun 25 00:00:00 EDT 2015
EDAG positively regulates erythroid differentiation and modifies GATA1 acetylation through recruiting p300.
Stem cells (Dayton, Ohio)
Zheng WW,Dong XM,Yin RH,Xu FF,Ning HM,Zhang MJ,Xu CW,Yang Y,Ding YL,Wang ZD,Zhao WB,Tang LJ,Chen H,Wang XH,Zhan YQ,Yu M,Ge CH,Li CY,Yang XM
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
Altered translation of GATA1 in Diamond-Blackfan anemia.
Nature medicine
Ludwig LS,Gazda HT,Eng JC,Eichhorn SW,Thiru P,Ghazvinian R,George TI,Gotlib JR,Beggs AH,Sieff CA,Lodish HF,Lander ES,Sankaran VG
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Tue Jul 01 00:00:00 EDT 2014
A flow cytometric method for characterization of circulating cell-derived microparticles in plasma.
Journal of extracellular vesicles
Nielsen MH,Beck-Nielsen H,Andersen MN,Handberg A
17-9987 was used in Flow cytometry/Cell sorting to establish a method to analyze microparticle subpopulations above the threshold of detection of a new generation BD FACSAriaTM III digital flow cytometer.
Mon Feb 10 00:00:00 EST 2014
A feedback loop consisting of microRNA 23a/27a and the β-like globin suppressors KLF3 and SP1 regulates globin gene expression.
Molecular and cellular biology
Ma Y,Wang B,Jiang F,Wang D,Liu H,Yan Y,Dong H,Wang F,Gong B,Zhu Y,Dong L,Yin H,Zhang Z,Zhao H,Wu Z,Zhang J,Zhou J,Yu J
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
α-1,6-Fucosyltransferase (FUT8) inhibits hemoglobin production during differentiation of murine and K562 human erythroleukemia cells.
The Journal of biological chemistry
Sasaki H,Toda T,Furukawa T,Mawatari Y,Takaesu R,Shimizu M,Wada R,Kato D,Utsugi T,Ohtsu M,Murakami Y
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Fri Jun 07 00:00:00 EDT 2013
Role of ZBP-89 in human globin gene regulation and erythroid differentiation.
Blood
Woo AJ,Kim J,Xu J,Huang H,Cantor AB
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 17-9987-42) in Flow Cytometry
Thu Sep 29 00:00:00 EDT 2011