Product References
miR‑122/SENP1 axis confers stemness and chemoresistance to liver cancer through Wnt/β‑catenin signaling.
Oncology letters
Dai J,Hao Y,Chen X,Yu Q,Wang B
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Fri Sep 01 00:00:00 EDT 2023
Naringin protects human nucleus pulposus cells against TNF-α-induced inflammation, oxidative stress, and loss of cellular homeostasis by enhancing autophagic flux via AMPK/SIRT1 activation.
Oxidative medicine and cellular longevity
Chen R,Gao S,Guan H,Zhang X,Gao Y,Su Y,Song Y,Jiang Y,Li N
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Thu Mar 24 00:00:00 EDT 2022
Elevated transcription and glycosylation of B3GNT5 promotes breast cancer aggressiveness.
Journal of experimental & clinical cancer research : CR
Miao Z,Cao Q,Liao R,Chen X,Li X,Bai L,Ma C,Deng X,Dai Z,Li J,Dong C
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Sat May 07 00:00:00 EDT 2022
Alterations in T and B cell function persist in convalescent COVID-19 patients.
Med (New York, N.Y.)
Shuwa HA,Shaw TN,Knight SB,Wemyss K,McClure FA,Pearmain L,Prise I,Jagger C,Morgan DJ,Khan S,Brand O,Mann ER,Ustianowski A,Bakerly ND,Dark P,Brightling CE,Brij S,Felton T,Simpson A,Grainger JR,Hussell T,Konkel JE,Menon M
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Fri Jun 11 00:00:00 EDT 2021
A CD10-OGP Membrane Peptolytic Signaling Axis in Fibroblasts Regulates Lipid Metabolism of Cancer Stem Cells via SCD1.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Yu S,Lu Y,Su A,Chen J,Li J,Zhou B,Liu X,Xia Q,Li Y,Li J,Huang M,Ye Y,Zhao Q,Jiang S,Yan X,Wang X,Di C,Pan J,Su S
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
IGF2BP2 regulates DANCR by serving as an N6-methyladenosine reader.
Cell death and differentiation
Hu X,Peng WX,Zhou H,Jiang J,Zhou X,Huang D,Mo YY,Yang L
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2020
Chemotherapeutic Stress Influences Epithelial-Mesenchymal Transition and Stemness in Cancer Stem Cells of Triple-Negative Breast Cancer.
International journal of molecular sciences
Li X,Strietz J,Bleilevens A,Stickeler E,Maurer J
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Wed Jan 08 00:00:00 EST 2020
Obesity-Altered Adipose Stem Cells Promote Radiation Resistance of Estrogen Receptor Positive Breast Cancer through Paracrine Signaling.
International journal of molecular sciences
Sabol RA,Villela VA,Denys A,Freeman BT,Hartono AB,Wise RM,Harrison MAA,Sandler MB,Hossain F,Miele L,Bunnell BA
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Wed Apr 15 00:00:00 EDT 2020
HMGB3 silence inhibits breast cancer cell proliferation and tumor growth by interacting with hypoxia-inducible factor 1α.
Cancer management and research
Gu J,Xu T,Huang QH,Zhang CM,Chen HY
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2020
BRCA1 regulates the cancer stem cell fate of breast cancer cells in the context of hypoxia and histone deacetylase inhibitors.
Scientific reports
Kim H,Lin Q,Yun Z
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Thu Jul 04 00:00:00 EDT 2019
The hypoxic tumor microenvironment in vivo selects the cancer stem cell fate of breast cancer cells.
Breast cancer research : BCR
Kim H,Lin Q,Glazer PM,Yun Z
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Tue Mar 06 00:00:00 EST 2018
Glutamic Pyruvate Transaminase GPT2 Promotes Tumorigenesis of Breast Cancer Cells by Activating Sonic Hedgehog Signaling.
Theranostics
Cao Y,Lin SH,Wang Y,Chin YE,Kang L,Mi J
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Mon Apr 30 00:00:00 EDT 2018
HOXC8 regulates self-renewal, differentiation and transformation of breast cancer stem cells.
Molecular cancer
Shah M,Cardenas R,Wang B,Persson J,Mongan NP,Grabowska A,Allegrucci C
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Thu Feb 16 00:00:00 EST 2017
Stage-specific embryonic antigen-3 (SSEA-3) and β3GalT5 are cancer specific and significant markers for breast cancer stem cells.
Proceedings of the National Academy of Sciences of the United States of America
Cheung SK,Chuang PK,Huang HW,Hwang-Verslues WW,Cho CH,Yang WB,Shen CN,Hsiao M,Hsu TL,Chang CF,Wong CH
14024782 was used in flow cytometry to find that breast cancer stem cells isolated with CD44(+)CD24(-/lo)SSEA-3(+) or ESA(hi)PROCR(hi)SSEA-3(+) markers have higher tumorigenicity than those with conventional markers
Tue Jan 26 00:00:00 EST 2016
The Z-cad dual fluorescent sensor detects dynamic changes between the epithelial and mesenchymal cellular states.
BMC biology
Toneff MJ,Sreekumar A,Tinnirello A,Hollander PD,Habib S,Li S,Ellis MJ,Xin L,Mani SA,Rosen JM
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Fri Jun 17 00:00:00 EDT 2016
The Z-cad dual fluorescent sensor detects dynamic changes between the epithelial and mesenchymal cellular states.
BMC biology
Toneff MJ,Sreekumar A,Tinnirello A,Hollander PD,Habib S,Li S,Ellis MJ,Xin L,Mani SA,Rosen JM
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Fri Jun 17 00:00:00 EDT 2016
Llgl1 prevents metaplastic survival driven by epidermal growth factor dependent migration.
Oncotarget
Greenwood E,Maisel S,Ebertz D,Russ A,Pandey R,Schroeder J
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Tue Sep 20 00:00:00 EDT 2016
Brief Report: IFIH1 Mutation Causes Systemic Lupus Erythematosus With Selective IgA Deficiency.
Arthritis & rheumatology (Hoboken, N.J.)
Van Eyck L,De Somer L,Pombal D,Bornschein S,Frans G,Humblet-Baron S,Moens L,de Zegher F,Bossuyt X,Wouters C,Liston A
14-0247 was used in Flow cytometry/Cell sorting to determine the underlying genetic defect responsible for severe early onset and refractory systemic lupus erythematosus.
Mon Jun 01 00:00:00 EDT 2015
TFAP2C governs the luminal epithelial phenotype in mammary development and carcinogenesis.
Oncogene
Cyr AR,Kulak MV,Park JM,Bogachek MV,Spanheimer PM,Woodfield GW,White-Baer LS,O'Malley YQ,Sugg SL,Olivier AK,Zhang W,Domann FE,Weigel RJ
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Thu Jan 22 00:00:00 EST 2015
Special AT-rich sequence-binding protein-1 participates in the maintenance of breast cancer stem cells through regulation of the Notch signaling pathway and expression of Snail1 and Twist1.
Molecular medicine reports
Sun Z,Zhang C,Zou X,Jiang G,Xu Z,Li W,Xie H
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2015
NDY1/KDM2B functions as a master regulator of polycomb complexes and controls self-renewal of breast cancer stem cells.
Cancer research
Kottakis F,Foltopoulou P,Sanidas I,Keller P,Wronski A,Dake BT,Ezell SA,Shen Z,Naber SP,Hinds PW,McNiel E,Kuperwasser C,Tsichlis PN
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Tue Jul 15 00:00:00 EDT 2014
Regulatory B cells are numerically but not functionally deficient in anti-neutrophil cytoplasm antibody-associated vasculitis.
Rheumatology (Oxford, England)
Todd SK,Pepper RJ,Draibe J,Tanna A,Pusey CD,Mauri C,Salama AD
14-0247 was used in Flow cytometry/Cell sorting to study the frequencies of different B cell subtypes during progression of ANCA-associated vasculitis.
Mon Sep 01 00:00:00 EDT 2014
Evaluation of cancer stem cell markers CD133, CD44, CD24: association with AKT isoforms and radiation resistance in colon cancer cells.
PloS one
Sahlberg SH,Spiegelberg D,Glimelius B,Stenerlöw B,Nestor M
14-0247 was used in Flow cytometry/Cell sorting to assess the connection between expression levels of various immune cell surface markers and radiation sensitivity.
Fri Jan 09 00:00:00 EST 2015
Isolation and characterization of intestinal stem cells based on surface marker combinations and colony-formation assay.
Gastroenterology
Wang F,Scoville D,He XC,Mahe MM,Box A,Perry JM,Smith NR,Lei NY,Davies PS,Fuller MK,Haug JS,McClain M,Gracz AD,Ding S,Stelzner M,Dunn JC,Magness ST,Wong MH,Martin MG,Helmrath M,Li L
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Thu Aug 01 00:00:00 EDT 2013
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
Increased invasion and tumorigenicity capacity of CD44+/CD24- breast cancer MCF7 cells in vitro and in nude mice.
Cancer cell international
Yan W,Chen Y,Yao Y,Zhang H,Wang T
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Immunofluorescence
Mon Jun 24 00:00:00 EDT 2013
Increased invasion and tumorigenicity capacity of CD44+/CD24- breast cancer MCF7 cells in vitro and in nude mice.
Cancer cell international
Yan W,Chen Y,Yao Y,Zhang H,Wang T
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Immunofluorescence
Mon Jun 24 00:00:00 EDT 2013
Expression of Six1 in luminal breast cancers predicts poor prognosis and promotes increases in tumor initiating cells by activation of extracellular signal-regulated kinase and transforming growth factor-beta signaling pathways.
Breast cancer research : BCR
Iwanaga R,Wang CA,Micalizzi DS,Harrell JC,Jedlicka P,Sartorius CA,Kabos P,Farabaugh SM,Bradford AP,Ford HL
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Thu Jul 05 00:00:00 EDT 2012
Expression of stem cell markers in the human fetal kidney.
PloS one
Metsuyanim S,Harari-Steinberg O,Buzhor E,Omer D,Pode-Shakked N,Ben-Hur H,Halperin R,Schneider D,Dekel B
Published figure using CD24 monoclonal antibody (Product # 14-0247-82) in Flow Cytometry
Fri Aug 21 00:00:00 EDT 2009