Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PD-L1 (CD274) Antibodies

          Invitrogen

          CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), eBioscience™

          Advanced Verification
          65 Published Figures
          126 References
          View all (250) PD-L1 (CD274) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (65)
          • Advanced Verification (1)
          CD274 (PD-L1, B7-H1) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CD274 (PD-L1, B7-H1) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 69

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD274 (PD-L1, B7-H1) Antibody (14-5982-82) in WB

          Western blot was performed using CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), eBioscience™ (Product # 14-5982-82) and a ~ 55 kDa band corresponding to CD274 (PD-L1) was observed in rat lymph node which is reported to be positive but not in rat brain. Tissue extracts (30 µg lysate) of rat axillary lymph node (Lane 1) and rat brain (Lane 2) were electrophoresed using NuPAGE™ 10% Bis-Tris Protein Gel (Prod... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD274 (PD-L1, B7-H1) Antibody in Western Blot (WB)
          CD274 (PD-L1, B7-H1) Antibody in Western Blot (WB)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Western Blot (WB)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Immunocytochemistry (ICC/IF)
          CD274 (PD-L1, B7-H1) Antibody in Immunocytochemistry (ICC/IF)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Immunohistochemistry (IHC)
          CD274 (PD-L1, B7-H1) Antibody in Western Blot (WB)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Flow Cytometry (Flow)
          CD274 (PD-L1, B7-H1) Antibody in Neutralization (Neu)
          CD274 (PD-L1, B7-H1) Antibody in Neutralization (Neu)
          CD274 (PD-L1, B7-H1) Antibody in Neutralization (Neu)
          CD274 (PD-L1, B7-H1) Antibody in Neutralization (Neu)
          CD274 (PD-L1, B7-H1) Antibody in Neutralization (Neu)
          CD274 (PD-L1, B7-H1) Antibody in Functional Assay (Functional)
          CD274 (PD-L1, B7-H1) Antibody

          Product Details

          14-5982-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 4 publications 4 publications

          Immunohistochemistry (IHC)

          -
          View 18 publications 18 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 5 publications 5 publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 78 publications 78 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Neutralization (Neu)

          Assay-Dependent
          View 12 publications 12 publications

          Functional Assay (Functional)

          Assay-Dependent
          View 3 publications 3 publications

          Inhibition Assays (Inhibition)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, lambda

          Class

          Monoclonal

          Type

          Antibody

          Clone

          MIH5

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 660 
          • APC 
          • Biotin 
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 650
          • Brilliant Violet 786
          • Functional Grade
          • NovaFluor Red 700
          • NovaFluor Red 755
          • NovaFluor Yellow 690
          • NovaFluor Yellow 700
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 780
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_467781

          Product Specific Information

          Description: The MIH5 monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH5 is reported to be a blocking antibody.

          Applications Reported: The MIH5 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections. It has also been reported in blocking in in vitro and in vivo functional assays. (Please use Functional Grade Purified MIH5, Product # 16-5982, in functional assays. Fluorochrome-conjugated MIH5 is recommended for use in flow cytometry).

          Applications Tested: This MIH5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: B7 homolog 1; B7-H1; CD274; PD-L1; PDCD1 ligand 1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; sCD274; soluble CD274; soluble PD L1; sPD L1

          View more View less

          Gene Aliases: A530045L16Rik; B7h1; Cd274; Pdcd1l1; Pdcd1lg1; Pdl1; RGD1566211

          View more View less

          UniProt ID: (Mouse) Q9EP73

          View more View less

          Entrez Gene ID: (Mouse) 60533, (Rat) 499342

          View more View less

          Function(s)
          protein binding immunoglobulin receptor superfamily
          Process(es)
          immune response signal transduction cell surface receptor signaling pathway positive regulation of cell migration T cell costimulation negative regulation of interferon-gamma production negative regulation of interleukin-10 production positive regulation of T cell proliferation negative regulation of T cell proliferation negative regulation of activated T cell proliferation toxin transport negative regulation of tumor necrosis factor superfamily cytokine production positive regulation of interleukin-10 secretion regulation of activated T cell proliferation regulation of T cell apoptotic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0