Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
0.5 µg/test | View 2 publications 2 publications |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Published species |
Mouse |
Host/Isotype |
Rat / IgG2b, kappa |
Recommended Isotype Control |
Rat IgG2b kappa Isotype Control (eB149/10H5), APC, eBioscience™ |
Class |
Monoclonal |
Type |
Antibody |
Clone |
7E.17G9 |
Conjugate |
APC
|
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Concentration |
0.2 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.2 |
Contains |
0.09% sodium azide |
Storage conditions |
4° C, store in dark, DO NOT FREEZE! |
RRID |
AB_2716948 |
Description: The 7E.17G9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells.
Applications Reported: This 7E.17G9 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This 7E.17G9 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
CD278 belongs to the CD28 and CTLA-4 cell-surface receptor family and is highly expressed in activated T-cells. It is essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. It enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: Activation-inducible lymphocyte immunomediatory molecule; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; inducible T-cell co-stimulator; Inducible T-cell costimulator
Gene Aliases: AILIM; CCLP; CRP-1; H4; Icos; Ly115
UniProt ID: (Mouse) Q9WVS0
Entrez Gene ID: (Mouse) 54167
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support