Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ICOS (CD278) Antibodies

          Invitrogen

          CD278 (ICOS) Monoclonal Antibody (C398.4A), APC, eBioscience™

          12 Published Figures
          12 References
          View all (92) ICOS (CD278) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD278 (ICOS) Monoclonal Antibody (C398.4A), APC, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (12)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD278 (ICOS) Antibody (17-9949-82) in Flow

          Staining of 3-day Con A-stimulated C57Bl/6 splenocytes with 0.06 µg of Armenian Hamster IgG Isotype Control APC (Product # 17-4888-82) (blue histogram) or 0.06 µg of Anti-Mouse/Rat CD278 (ICOS) APC (purple histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)
          CD278 (ICOS) Antibody in Flow Cytometry (Flow)

          Product Details

          17-9949-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.125 µg/test
          View 12 publications 12 publications
          Product Specifications

          Species Reactivity

          Mouse, Rhesus monkey, Rat

          Published species

          C. elegans, Mouse

          Host/Isotype

          Armenian hamster / IgG

          Recommended Isotype Control

          Armenian Hamster IgG Isotype Control (eBio299Arm), APC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          C398.4A

          Conjugate

          APC APC APC
          • Unconjugated
          • APC-eFluor 780
          • Biotin 
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • Super Bright 436
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11149880

          Product Specific Information

          Description: The C398.4A monoclonal antibody reacts with ICOS (Inducible Costimulatory molecule), also known as H4/CRP-1/AILIM, and cross-reacts with mouse and rat ICOS. ICOS is a T cell specific activation molecule and a third member of the CD28/CTLA-4 family. A dimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1, is thought to play a protective role in inflammatory autoimmune diseases and be involved in the development of Th2 cells. C398.4A is reported to display strong costimulation of proliferation of T cell clones, intermediate costimulation on activated T cells, and weak costimulation on fresh resting T cells. It is a good costimulator of IL-10 production especially from pre-activated cells that express high level of H4/ICOS. The epitope recognized by C398.4A is different from that bound by the ICOSL (i.e., C398.4A does not block binding of ICOSL).

          Applications Reported: This C398.4A antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This C398.4A antibody has been tested by flow cytometric analysis of Con A-stimulated mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: Activation-inducible lymphocyte immunomediatory molecule; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; inducible T-cell co-stimulator; Inducible T-cell costimulator

          View more View less

          Gene Aliases: AILIM; CCLP; CRP-1; H4; Icos; Ly115

          View more View less

          UniProt ID: (Rat) Q9R1T7, (Mouse) Q9WVS0

          View more View less

          Entrez Gene ID: (Rhesus monkey) 705788, (Rat) 64545, (Mouse) 54167

          View more View less

          Function(s)
          molecular_function protein binding
          Process(es)
          T cell tolerance induction single organismal cell-cell adhesion T cell costimulation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7988999796-cn442:80/100.66.76.23:80.
          git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.39.1-2025.11.99-1.0