Product References
Ultrasound combined with nanobubbles promotes systemic anticancer immunity and augments anti-PD1 efficacy.
Journal for immunotherapy of cancer
Hu J,He J,Wang Y,Zhao Y,Fang K,Dong Y,Chen Y,Zhang Y,Zhang C,Wang H,Tan J,Wang J,Zi R,Liu C,Liang H,Guo Y,Ou J
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2022
CD153/CD30 signaling promotes age-dependent tertiary lymphoid tissue expansion and kidney injury.
The Journal of clinical investigation
Sato Y,Oguchi A,Fukushima Y,Masuda K,Toriu N,Taniguchi K,Yoshikawa T,Cui X,Kondo M,Hosoi T,Komidori S,Shimizu Y,Fujita H,Jiang L,Kong Y,Yamanashi T,Seita J,Yamamoto T,Toyokuni S,Hamazaki Y,Hattori M,Yoshikai Y,Boor P,Floege J,Kawamoto H,Murakawa Y,Minato N,Yanagita M
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Jan 18 00:00:00 EST 2022
Graded RhoA GTPase Expression in Treg Cells Distinguishes Tumor Immunity From Autoimmunity.
Frontiers in immunology
Kalim KW,Yang JQ,Modur V,Nguyen P,Li Y,Zheng Y,Guo F
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Jan 11 00:00:00 EST 2022
Histone deacetylase 3 represses cholesterol efflux during CD4+ T-cell activation.
eLife
Wilfahrt D,Philips RL,Lama J,Kizerwetter M,Shapiro MJ,McCue SA,Kennedy MM,Rajcula MJ,Zeng H,Shapiro VS
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Thu Dec 02 00:00:00 EST 2021
PTENα functions as an immune suppressor and promotes immune resistance in PTEN-mutant cancer.
Nature communications
Sun Y,Lu D,Yin Y,Song J,Liu Y,Hao W,Qi F,Zhang G,Zhang X,Liu L,Lin Z,Liang H,Zhao X,Jin Y,Yin Y
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Thu Aug 26 00:00:00 EDT 2021
Ileal immune tonus is a prognosis marker of proximal colon cancer in mice and patients.
Cell death and differentiation
Picard M,Yonekura S,Slowicka K,Petta I,Rauber C,Routy B,Richard C,Iebba V,Tidjani Alou M,Becharef S,Ly P,Pizzato E,Lehmann CHK,Amon L,Klein C,Opolon P,Gomperts Boneca I,Scoazec JY,Hollebecque A,Malka D,Ghiringhelli F,Dudziak D,Berx G,Vereecke L,van Loo G,Kroemer G,Zitvogel L,Roberti MP
17-9985 was used in Flow cytometry/Cell sorting to suggest that chemotherapy inversely shapes the immune profile of the ileum-tumor axis, influencing clinical outcome.
Sat May 01 00:00:00 EDT 2021
The ubiquitin ligase Peli1 inhibits ICOS and thereby Tfh-mediated immunity.
Cellular & molecular immunology
Huang X,Hao S,Liu J,Huang Y,Liu M,Xiao C,Wang Y,Pei S,Yu T,Xu J,Wang H,Dai D,Su X,Xiao Y
17-9985-82 was used in Flow Cytometry to elucidate the molecular mechanisms regulating T follicular helper cells.
Thu Apr 01 00:00:00 EDT 2021
5-Azacytidine Potentiates Anti-tumor Immunity in a Model of Pancreatic Ductal Adenocarcinoma.
Frontiers in immunology
Ebelt ND,Zuniga E,Johnson BL,Diamond DJ,Manuel ER
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Fri Mar 26 00:00:00 EDT 2021
NF-κB-inducing kinase maintains T cell metabolic fitness in antitumor immunity.
Nature immunology
Gu M,Zhou X,Sohn JH,Zhu L,Jie Z,Yang JY,Zheng X,Xie X,Yang J,Shi Y,Brightbill HD,Kim JB,Wang J,Cheng X,Sun SC
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Ketogenic diet and ketone bodies enhance the anticancer effects of PD-1 blockade.
JCI insight
Ferrere G,Tidjani Alou M,Liu P,Goubet AG,Fidelle M,Kepp O,Durand S,Iebba V,Fluckiger A,Daillère R,Thelemaque C,Grajeda-Iglesias C,Alves Costa Silva C,Aprahamian F,Lefevre D,Zhao L,Ryffel B,Colomba E,Arnedos M,Drubay D,Rauber C,Raoult D,Asnicar F,Spector T,Segata N,Derosa L,Kroemer G,Zitvogel L
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Mon Jan 25 00:00:00 EST 2021
Therapeutic efficacy of combined vaccination against tumor pericyte-associated antigens DLK1 and DLK2 in mice.
Oncoimmunology
Fabian KP,Chi-Sabins N,Taylor JL,Fecek R,Weinstein A,Storkus WJ
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
The stress kinase GCN2 does not mediate suppression of antitumor T cell responses by tryptophan catabolism in experimental melanomas.
Oncoimmunology
Sonner JK,Deumelandt K,Ott M,Thomé CM,Rauschenbach KJ,Schulz S,Munteanu B,Mohapatra S,Adam I,Hofer AC,Feuerer M,Opitz CA,Hopf C,Wick W,Platten M
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Growth and metastasis of lung adenocarcinoma is potentiated by BMP4-mediated immunosuppression.
Oncoimmunology
Chen L,Yi X,Goswami S,Ahn YH,Roybal JD,Yang Y,Diao L,Peng D,Peng D,Fradette JJ,Wang J,Byers LA,Kurie JM,Ullrich SE,Qin FX,Gibbons DL
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Stearoyl-CoA Desaturase-Mediated Monounsaturated Fatty Acid Availability Supports Humoral Immunity.
Cell reports
Zhou X,Zhu X,Li C,Li Y,Ye Z,Shapiro VS,Copland JA,Hitosugi T,Bernlohr DA,Sun J,Zeng H
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Jan 05 00:00:00 EST 2021
Serum HBsAg clearance has minimal impact on CD8+ T cell responses in mouse models of HBV infection.
The Journal of experimental medicine
Fumagalli V,Di Lucia P,Venzin V,Bono EB,Jordan R,Frey CR,Delaney W,Chisari FV,Guidotti LG,Iannacone M
17-9985 was used in Flow cytometry/Cell sorting to reveal that circulating HBsAg clearance does not improve HBV-specific CD8+ T cell responses in vivo and may have important implications for the treatment of chronic HBV infection.
Mon Nov 02 00:00:00 EST 2020
Serum HBsAg clearance has minimal impact on CD8+ T cell responses in mouse models of HBV infection.
The Journal of experimental medicine
Fumagalli V,Di Lucia P,Venzin V,Bono EB,Jordan R,Frey CR,Delaney W,Chisari FV,Guidotti LG,Iannacone M
17-9985 was used in Flow cytometry/Cell sorting to reveal that circulating HBsAg clearance does not improve HBV-specific CD8+ T cell responses in vivo and may have important implications for the treatment of chronic HBV infection.
Mon Nov 02 00:00:00 EST 2020
Lysosomal Acid Lipase Is Required for Donor T Cells to Induce Graft-versus-Host Disease.
Cell reports
Nguyen HD,Ticer T,Bastian D,Kuril S,Li H,Du H,Yan C,Yu XZ
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Oct 27 00:00:00 EDT 2020
The Selenoprotein MsrB1 Instructs Dendritic Cells to Induce T-Helper 1 Immune Responses.
Antioxidants (Basel, Switzerland)
Lee HJ,Park JS,Yoo HJ,Lee HM,Lee BC,Kim JH
17-9985-82 was used in Flow Cytometry to show methionine sulfoxide reductase B1 (MsrB1) acts in dendritic cells to regulate the phosphorylation of signal transducer and activator of transcription-6 (STAT6).
Tue Oct 20 00:00:00 EDT 2020
The Selenoprotein MsrB1 Instructs Dendritic Cells to Induce T-Helper 1 Immune Responses.
Antioxidants (Basel, Switzerland)
Lee HJ,Park JS,Yoo HJ,Lee HM,Lee BC,Kim JH
17-9985-82 was used in Flow Cytometry to show methionine sulfoxide reductase B1 (MsrB1) acts in dendritic cells to regulate the phosphorylation of signal transducer and activator of transcription-6 (STAT6).
Tue Oct 20 00:00:00 EDT 2020
Gene silencing of indoleamine 2,3-dioxygenase 1 inhibits lung cancer growth by suppressing T-cell exhaustion.
Oncology letters
Shang K,Wang Z,Hu Y,Huang Y,Yuan K,Yu Y
17-9985 was used in Flow cytometry/Cell sorting to study the association between Indoleamine 2,3-dioxygenase 1 (IDO1) and T-cell exhaustion during tumor progression.
Mon Jun 01 00:00:00 EDT 2020
Gene silencing of indoleamine 2,3-dioxygenase 1 inhibits lung cancer growth by suppressing T-cell exhaustion.
Oncology letters
Shang K,Wang Z,Hu Y,Huang Y,Yuan K,Yu Y
17-9985 was used in Flow cytometry/Cell sorting to study the association between Indoleamine 2,3-dioxygenase 1 (IDO1) and T-cell exhaustion during tumor progression.
Mon Jun 01 00:00:00 EDT 2020
Dynamics and genomic landscape of CD8+ T cells undergoing hepatic priming.
Nature
Bénéchet AP,De Simone G,Di Lucia P,Cilenti F,Barbiera G,Le Bert N,Fumagalli V,Lusito E,Moalli F,Bianchessi V,Andreata F,Zordan P,Bono E,Giustini L,Bonilla WV,Bleriot C,Kunasegaran K,Gonzalez-Aseguinolaza G,Pinschewer DD,Kennedy PTF,Naldini L,Kuka M,Ginhoux F,Cantore A,Bertoletti A,Ostuni R,Guidotti LG,Iannacone M
17-9985 was used in Flow cytometry/Cell sorting to show that priming by Kupffer cells, which are not natural targets of hepatitis B, leads to differentiation of CD8+ T cells into effector cells that form dense, extravascular clusters of immotile cells scattered throughout the liver.
Tue Oct 01 00:00:00 EDT 2019
CXCR5+PD-1+ follicular helper CD8 T cells control B cell tolerance.
Nature communications
Chen Y,Yu M,Zheng Y,Fu G,Xin G,Zhu W,Luo L,Burns R,Li QZ,Dent AL,Zhu N,Cui W,Malherbe L,Wen R,Wang D
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Fri Sep 27 00:00:00 EDT 2019
Orphan Nuclear Receptor NR2F6 Suppresses T Follicular Helper Cell Accumulation through Regulation of IL-21.
Cell reports
Olson WJ,Jakic B,Labi V,Schoeler K,Kind M,Klepsch V,Baier G,Hermann-Kleiter N
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Sep 10 00:00:00 EDT 2019
Combination PD-1 and PD-L1 Blockade Promotes Durable Neoantigen-Specific T Cell-Mediated Immunity in Pancreatic Ductal Adenocarcinoma.
Cell reports
Burrack AL,Spartz EJ,Raynor JF,Wang I,Olson M,Stromnes IM
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Aug 20 00:00:00 EDT 2019
Orally administered heat-killed Lactobacillus paracasei MCC1849 enhances antigen-specific IgA secretion and induces follicular helper T cells in mice.
PloS one
Arai S,Iwabuchi N,Takahashi S,Xiao JZ,Abe F,Hachimura S
17-9985 was used in Flow cytometry/Cell sorting to investigate the effects of orally administered heat-killed MCC1849 on IgA production in the intestine and on Tfh cell induction in vivo.
Wed Jan 02 00:00:00 EST 2019
PD-1 Blockade and CD27 Stimulation Activate Distinct Transcriptional Programs That Synergize for CD8+ T-Cell-Driven Antitumor Immunity.
Clinical cancer research : an official journal of the American Association for Cancer Research
Buchan SL,Fallatah M,Thirdborough SM,Taraban VY,Rogel A,Thomas LJ,Penfold CA,He LZ,Curran MA,Keler T,Al-Shamkhani A
17-9985 was used in Flow cytometry/Cell sorting to show that suboptimal T-cell invigoration in cancer patients undergoing treatment with PD-1 checkpoint blockers will be improved by dual PD-1 blockade and CD27 agonism.
Tue May 15 00:00:00 EDT 2018
Intravenous Labeling and Analysis of the Content of Thymic Perivascular Spaces.
Bio-protocol
Ruscher R,Hogquist KA
17-9985 was used in Flow cytometry/Cell sorting to investigate hematopoietic cell subsets within the perivascular spaces of the thymus.
Mon Mar 05 00:00:00 EST 2018
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Thu Jan 25 00:00:00 EST 2018
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
17-9985 was used in Flow cytometry/Cell sorting to support that Stat3- and BCL6-dependent extrafollicular CD4+ T and B interactions play critical functions in the pathogenesis of chronic graft-versus-host disease.
Tue Oct 17 00:00:00 EDT 2017
Oncolytic Adenovirus and Tumor-Targeting Immune Modulatory Therapy Improve Autologous Cancer Vaccination.
Cancer research
Jiang H,Rivera-Molina Y,Gomez-Manzano C,Clise-Dwyer K,Bover L,Vence LM,Yuan Y,Lang FF,Toniatti C,Hossain MB,Fueyo J
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Sat Jul 15 00:00:00 EDT 2017
Cblb-deficient T cells are less susceptible to PD-L1-mediated inhibition.
Oncotarget
Peer S,Baier G,Gruber T
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Jun 27 00:00:00 EDT 2017
Intratumoral modulation of the inducible co-stimulator ICOS by recombinant oncolytic virus promotes systemic anti-tumour immunity.
Nature communications
Zamarin D,Holmgaard RB,Ricca J,Plitt T,Palese P,Sharma P,Merghoub T,Wolchok JD,Allison JP
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2017
Metastasis is regulated via microRNA-200/ZEB1 axis control of tumour cell PD-L1 expression and intratumoral immunosuppression.
Nature communications
Chen L,Gibbons DL,Goswami S,Cortez MA,Ahn YH,Byers LA,Zhang X,Yi X,Dwyer D,Lin W,Diao L,Wang J,Roybal J,Patel M,Ungewiss C,Peng D,Antonia S,Mediavilla-Varela M,Robertson G,Suraokar M,Welsh JW,Erez B,Wistuba II,Chen L,Peng D,Wang S,Ullrich SE,Heymach JV,Kurie JM,Qin FX
Published figure using CD279 (PD-1) monoclonal antibody (Product # 17-9985-82) in Flow Cytometry
Tue Oct 28 00:00:00 EDT 2014
The programmed death-1 (PD-1) pathway regulates autoimmune diabetes in nonobese diabetic (NOD) mice.
The Journal of experimental medicine
Ansari MJ,Salama AD,Chitnis T,Smith RN,Yagita H,Akiba H,Yamazaki T,Azuma M,Iwai H,Khoury SJ,Auchincloss H,Sayegh MH
Mon Jul 07 00:00:00 EDT 2003
The programmed death-1 (PD-1) pathway regulates autoimmune diabetes in nonobese diabetic (NOD) mice.
The Journal of experimental medicine
Ansari MJ,Salama AD,Chitnis T,Smith RN,Yagita H,Akiba H,Yamazaki T,Azuma M,Iwai H,Khoury SJ,Auchincloss H,Sayegh MH
Mon Jul 07 00:00:00 EDT 2003
The programmed death-1 (PD-1) pathway regulates autoimmune diabetes in nonobese diabetic (NOD) mice.
The Journal of experimental medicine
Ansari MJ,Salama AD,Chitnis T,Smith RN,Yagita H,Akiba H,Yamazaki T,Azuma M,Iwai H,Khoury SJ,Auchincloss H,Sayegh MH
Mon Jul 07 00:00:00 EDT 2003