Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
Assay-dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Bovine |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
CC219 |
Immunogen |
Plasmid DNA encoding the bovine CD28 gene |
Conjugate |
Unconjugated
|
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS |
Contains |
0.09% sodium azide |
Storage conditions |
4°C or -20°C if preferred |
RRID |
AB_2745224 |
Mouse anti Bovine CD28, clone CC219, recognizes the bovine homologue of human CD28, a co-stimulatory cell surface molecule expressed by T cell subsets.
CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary's Disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen CD28; CD28; contains partial extracellular domain; MGC138290; sCD28; soluble CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T44
Gene Aliases: CD28
UniProt ID: (Bovine) Q28071
Entrez Gene ID: (Bovine) 281050
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support