Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For blocking studies in vitro, dilutions have to be made according to the amounts of MGL (CD301) to be inactivated. The antigen is glutaraldehyde (0.05%) resistant.
Endotoxin <24 EU/mg.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 10 member A; M-ASGP-BP-1; Macrophage asialoglycoprotein-binding protein 1; macrophage galactose N-acetyl-galactosamine specific lectin 1; Macrophage galactose/N-acetylgalactosamine-specific lectin; MMGL
Gene Aliases: CD301a; Clec10a; M-ASGP-BP-1; Mgl; Mgl1
UniProt ID: (Mouse) P49300
Entrez Gene ID: (Mouse) 17312
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support