Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EpCAM (CD326) Antibodies

          Invitrogen

          CD326 (EpCAM) Monoclonal Antibody (G8.8), FITC, eBioscience™

          Advanced Verification
          94 Published Figures
          113 References
          View all (302) EpCAM (CD326) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD326 (EpCAM) Monoclonal Antibody (G8.8), FITC, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (94)
          • Advanced Verification (1)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 96

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD326 (EpCAM) Antibody (11-5791-82) in Flow

          Staining of the TE-71 cell line with 0.25 µg of Rat IgG2a K Isotype Control FITC (Product # 11-4321-42) (blue histogram) or 0.25 µg of Anti-Mouse CD326 (EpCAM) FITC (purple histogram). Total viable cells, as determined by Fixable Viability Dye eFluor® 450, were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Flow Cytometry (Flow)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunocytochemistry (ICC/IF)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody in Immunohistochemistry (IHC)
          CD326 (EpCAM) Antibody

          Product Details

          11-5791-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 15 publications 15 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 16 publications 16 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 77 publications 77 publications

          Affinity Purification (AP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), FITC, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          G8.8

          Conjugate

          FITC FITC FITC
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          498/517 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11151709

          Product Specific Information

          Description: The G8.8 monoclonal antibody reacts with the 40 kDa protein mouse EpCAM (epithelial cellular adhesion molecule), also known as EGP40 (epithelial glycoprotein 40), 17-1A antigen, TACSTD1 (tumor-associated calcium signal transducer 1), and CD326. The immunogen used to generate the G8.8 antibody was the TE-71 thymic epithelial cell line. CD326 is expressed on the majority of epithelial cells, and is considered a pan-carcinoma antigen. CD326 mediates calcium-independent, homophilic, cell-cell adhesion and may function as a growth factor receptor. The antigen is being used as a target for immunotherapy treatment of human carcinomas. CD326 binds LAIR-1 (CD305) and LAIR-2 (CD306) to inhibit cellular activation and inflammation. This epithelial glycoprotein is now recognized as having an important role in tumor biology.

          Applications Reported: This G8.8 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This G8.8 antibody has been tested by flow cytometric analysis of the TE-71 cell line. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Ep-CAM (epithelial adhesion molecule, epithelial specific antigen, ESA) is a transmembrane glycoprotein expressed in the epithelium with a molecular weight of approximately 40 kDa, which functions as an epithelial cell adhesion molecule. Ep-CAM functions as a homotypic calcium-independent cell adhesion molecule, and has a direct impact on cell cycle, proliferation and metabolism of epithelial cells and fibroblasts due to its ability to rapidly induce the proto-oncogene c-myc and the cell cycle regulating genes cyclin A and E. Ep-CAM mediates Ca2+-independent homotypic interactions. Formation of Ep-CAM-mediated adhesions have a negative regulatory effect on adhesions mediated by classic cadherins, which may have strong effects on the differentiation and growth of epithelial cells. Ep-CAM overexpression was suggested to be associated with enhanced epithelial proliferation. Ep-CAM is highly expressed in human carcinomas, and is a marker for tumors of epithelial lineage. Ep-CAM is expressed on baso-lateral cell surface in most simple epithelia and many carcinoma types. Also, Ep-CAM reportedly distinguishes adenocarcinomas from pleural mesotheliomas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD326; EGP314; Ep-CAM; Epithelial cell adhesion molecule; Epithelial glycoprotein 314; hEGP314; lymphocyte antigen 74; mEGP314; panepithelial glycoprotein 314; Protein 289A; Trop-1 protein; Tumor-associated calcium signal transducer 1

          View more View less

          Gene Aliases: CD326; EGP; EGP-2; Egp314; Ep-CAM; Epcam; EpCAM1; GA733-2; gp40; Ly74; Tacsd1; Tacstd1; TROP1

          View more View less

          UniProt ID: (Mouse) Q99JW5

          View more View less

          Entrez Gene ID: (Mouse) 17075

          View more View less

          Function(s)
          protein complex binding cadherin binding involved in cell-cell adhesion
          Process(es)
          positive regulation of cell proliferation signal transduction involved in regulation of gene expression negative regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter stem cell differentiation cell-cell adhesion negative regulation of cell-cell adhesion mediated by cadherin positive regulation of cell motility positive regulation of stem cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline