MA5-13567 targets CD37 in FACS, IF, and IHC (F) applications and shows reactivity with Human samples.
The MA5-13567 immunogen is spleen cells of a patient with hairy cell leukemia.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD37; cell differentiation antigen 37; Leukocyte antigen CD37; leukocyte surface antigen CD37; Tetraspanin-26; Tspan-26
Gene Aliases: CD37; GP52-40; TSPAN26
UniProt ID: (Human) P11049
Entrez Gene ID: (Human) 951
Molecular Function:
cell adhesion molecule
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support