Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200-1:1,000 | View 1 publication 1 publication |
Immunohistochemistry (IHC) |
- | View 1 publication 1 publication |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:20-1:200 | - |
Immunocytochemistry (ICC/IF) |
1:20-1:200 | - |
Flow Cytometry (Flow) |
0.5 µg/1x10^6 cells | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Mouse, Rat |
Published species |
Human |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
CD39/ENTPD1 Fusion Protein Ag5429 (1-340 aa encoded by BC047664) |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.17 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
Immunogen sequence: MESEELADR VLDVVERSLS NYPFDFQGAR IITGQEEGAY GWITINYLLG KFSQKTRWFS IVPYETNNQE TFGALDLGGA STQVTFVPQN QTIESPDNAL QFRLYGKDYN VYTHSFLCYG KDQALWQKLA KDIQVASNEI LRDPCFHPGY KKVVNVSDLY KTPCTKRFEM TLPFQQFEIQ GIGNYQQCHQ SILELFNTSY CPYSQCAFNG IFLPPLQGDF GAFSAFYFVM KFLNLTSEKV SQEKVTEMMK KFCAQPWEEI KTSYAGVKEK YLSEYCFSGT YILSLLLQGY HFTADSWEHI HFIGKIQGSD AGWTLGYMLN LTNMIPAEQP LSTPLSHSTY V (1-340 aa encoded by BC047664)
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPDase; CD39; CD39 antigen; DKFZp686D194; DKFZp686I093; Ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; Ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; FLJ40921; FLJ40959; Lymphoid cell activation antigen; NTPDase 1
Gene Aliases: 2610206B08Rik; AA408691; ATPDase; CD39; ENTPD1; NTPDase-1; SPG64
UniProt ID: (Human) P49961, (Mouse) P55772
Entrez Gene ID: (Human) 953, (Mouse) 12495, (Rat) 64519
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support