Product References
LRPPRC facilitates tumor progression and immune evasion through upregulation of m6A modification of PD-L1 mRNA in hepatocellular carcinoma.
Frontiers in immunology
Wang H,Tang A,Cui Y,Gong H,Li H
Tue Apr 18 00:00:00 EDT 2023
Dose-Dependent Effects in Plasma Oncotherapy: Critical In Vivo Immune Responses Missed by In Vitro Studies.
Biomolecules
He Y,Gong F,Jin T,Liu Q,Fang H,Chen Y,Wang G,Chu PK,Wu Z,Ostrikov KK
Fri Apr 21 00:00:00 EDT 2023
Murine cytomegalovirus localization and uveitic cell infiltration might both contribute to trabecular meshwork impairment in Posner-Schlossman syndrome: Evidence from an open-angle rat model.
Experimental eye research
Sheng Q,Sun Y,Zhai R,Fan X,Ying Y,Liu Z,Kong X
Thu Jun 01 00:00:00 EDT 2023
PYCR1 promotes the malignant progression of lung cancer through the JAK-STAT3 signaling pathway via PRODH-dependent glutamine synthesize.
Translational oncology
Zhang L,Zhao X,Wang E,Yang Y,Hu L,Xu H,Zhang B
Thu Jun 01 00:00:00 EDT 2023
Combination of novel oncolytic herpesvirus with paclitaxel as an efficient strategy for breast cancer therapy.
Journal of medical virology
Deng X,Shen Y,Yi M,Zhang C,Zhao B,Zhong G,Xue D,Leng Q,Ding J,Zhao R,Jia W,Dong C,Dai Z
Mon May 01 00:00:00 EDT 2023
Inhibition of phosphoglycerate kinase 1 attenuates autoimmune myocarditis by reprogramming CD4+ T cell metabolism.
Cardiovascular research
Lu Y,Zhao N,Wu Y,Yang S,Wu Q,Dong Q,Du Y
Tue Jun 13 00:00:00 EDT 2023
Apoptotic tumor cell-derived microparticles loading Napabucasin inhibit CSCs and synergistic immune therapy.
Journal of nanobiotechnology
Jing B,Guo F,An R,Gao Y,Li Y,Xie Y,Wang J,Chen Y,Li H,Gao T,Jin Q,Zhang L,Xie M
Thu Feb 02 00:00:00 EST 2023
Hepatocellular carcinoma subtypes based on metabolic pathways reveals potential therapeutic targets.
Frontiers in oncology
He Z,Chen Q,He W,Cao J,Yao S,Huang Q,Zheng Y
Tue Mar 21 00:00:00 EDT 2023
Combinational Deletions of MGF360-9L and MGF505-7R Attenuated Highly Virulent African Swine Fever Virus and Conferred Protection against Homologous Challenge.
Journal of virology
Ding M,Dang W,Liu H,Xu F,Huang H,Sunkang Y,Li T,Pei J,Liu X,Zhang Y,Zheng H
Wed Jul 27 00:00:00 EDT 2022
Identification of a novel immune gene panel in tongue squamous cell carcinoma.
American journal of translational research
Sun J,Guo F,Tang Q,Chen G,Peng J,Shen Y,Zhang J,Hu J,Yang C
Sat Jul 16 00:00:00 EDT 2022
CLEC1B is a Promising Prognostic Biomarker and Correlated with Immune Infiltration in Hepatocellular Carcinoma.
International journal of general medicine
Liang X,Song F,Fang W,Zhang Y,Feng Z,Chen Z,Han L,Chen Z
Sat Jul 16 00:00:00 EDT 2022
Increased co-expression of MEST and BRCA1 is associated with worse prognosis and immune infiltration in ovarian cancer.
Gynecologic oncology
Zhang J,Yu S,Li Q,Wang Q,Zhang J
Tue Mar 01 00:00:00 EST 2022
The regulatory roles of T helper cells in distinct extracellular matrix characterization in breast cancer.
Frontiers in immunology
Tian Q,Gao H,Ma Y,Zhu L,Zhou Y,Shen Y,Wang B
Wed Sep 28 00:00:00 EDT 2022
Characteristics of the immunogenicity and tumor immune microenvironment in HER2-amplified lung adenocarcinoma.
Frontiers in immunology
Wang Q,Mao Z,Li W,Wang S,Wang L,Chen L,Yang Z,Fu X,Jiang P,Bai Y,Xu L,Zhang S,Hou Y,Jia X,Jiang L,Liu M,Zhang G,Jiang Y,Guo H
Thu Jan 12 00:00:00 EST 2023
The Immunological Contribution of a Novel Metabolism-Related Signature to the Prognosis and Anti-Tumor Immunity in Cervical Cancer.
Cancers
Yu S,Li X,Ma M,Yang R,Zhang J,Wu S
Fri May 13 00:00:00 EDT 2022
The impact of neoadjuvant chemotherapy on the tumor microenvironment in advanced high-grade serous carcinoma.
Oncogenesis
Shen Y,Ren Y,Chen K,Cen Y,Zhang B,Lu W,Xu J
Sat Jul 30 00:00:00 EDT 2022
Immunity and Extracellular Matrix Characteristics of Breast Cancer Subtypes Based on Identification by T Helper Cells Profiling.
Frontiers in immunology
Zhou Y,Tian Q,Gao H,Zhu L,Zhang Y,Zhang C,Yang J,Wang B
Fri Jul 08 00:00:00 EDT 2022
Single-cell RNA sequencing reveals distinct immunology profiles in human keloid.
Frontiers in immunology
Feng C,Shan M,Xia Y,Zheng Z,He K,Wei Y,Song K,Meng T,Liu H,Hao Y,Liang Z,Wang Y,Huang Y
Tue Aug 23 00:00:00 EDT 2022
Chronic stress-induced depression requires the recruitment of peripheral Th17 cells into the brain.
Journal of neuroinflammation
Peng Z,Peng S,Lin K,Zhao B,Wei L,Tuo Q,Liao D,Yuan T,Shi Z
Thu Jul 14 00:00:00 EDT 2022
Hyperbaric oxygen treatment on keloid tumor immune gene expression.
Chinese medical journal
Wang CH,Shan MJ,Liu H,Hao Y,Song KX,Wu HW,Meng T,Feng C,Qi Z,Wang Z,Wang YB
Mon Sep 13 00:00:00 EDT 2021
A Novel Strategy Conjugating PD-L1 Polypeptide With Doxorubicin Alleviates Chemotherapeutic Resistance and Enhances Immune Response in Colon Cancer.
Frontiers in oncology
Wang M,Shu XS,Li M,Zhang Y,Yao Y,Huang X,Li J,Wei P,He Z,Lu J,Ying Y
Thu Apr 28 00:00:00 EDT 2022