Product References
Repositioning liothyronine for cancer immunotherapy by blocking the interaction of immune checkpoint TIGIT/PVR.
Cell communication and signaling : CCS
Zhou X,Du J,Wang H,Chen C,Jiao L,Cheng X,Zhou X,Chen S,Gou S,Zhao W,Zhai W,Chen J,Gao Y
45-0048 was used in Flow cytometry/Cell sorting to discover how liothyronine could serve as a potential candidate for cancer immunotherapy by blocking the immune checkpoint TIGIT/PVR.
Mon Sep 07 00:00:00 EDT 2020
The Potential of Nanobody-Targeted Photodynamic Therapy to Trigger Immune Responses.
Cancers
Beltrán Hernández I,Angelier ML,Del Buono D'Ondes T,Di Maggio A,Yu Y,Oliveira S
45-0048 was used in Flow cytometry/Cell sorting to explore the potential of NB-PDT to stimulate the immune system.
Wed Apr 15 00:00:00 EDT 2020
Human Fc Receptor-like 3 Inhibits Regulatory T Cell Function and Binds Secretory IgA.
Cell reports
Agarwal S,Kraus Z,Dement-Brown J,Alabi O,Starost K,Tolnay M
45-0048 was used in Flow cytometry/Cell sorting to study the functional regulation of T cells by human Fc receptor-like 3.
Tue Feb 04 00:00:00 EST 2020
Schistosoma mansoni rSm29 Antigen Induces a Regulatory Phenotype on Dendritic Cells and Lymphocytes From Patients With Cutaneous Leishmaniasis.
Frontiers in immunology
Lopes DM,Oliveira SC,Page B,Carvalho LP,Carvalho EM,Cardoso LS
45-0048 was used in Flow cytometry/Cell sorting to evaluate the potential of the S.mansoni Sm29 antigen to change aspects of profiles of monocyte derived dendritic cells and lymphocytes from subjects with cutaneous leishmaniasis.
Tue Oct 15 00:00:00 EDT 2019
Efficient gene transfer into T lymphocytes by fiber-modified human adenovirus 5.
BMC biotechnology
Lv Y,Xiao FJ,Wang Y,Zou XH,Wang H,Wang HY,Wang LS,Lu ZZ
45-0048 was used in Flow cytometry/Cell sorting to show the HAdV-11p fiber pseudotyped HAdV-5 could effectively transduce human T cells when human EF1a promoter was used to control the expression of transgene, suggesting its possible application in T cell immunocellular therapy.
Wed Apr 24 00:00:00 EDT 2019
Creation of an immunodeficient HLA-transgenic mouse (HUMAMICE) and functional validation of human immunity after transfer of HLA-matched human cells.
PloS one
Zeng Y,Liu B,Rubio MT,Wang X,Ojcius DM,Tang R,Durrbach A,Ru Z,Zhou Y,Lone YC
45-0048 was used in Flow cytometry/Cell sorting to generate a novel immuno-deficient mouse strain expressing human HLA molecules instead of mouse MHC molecules.
Tue Sep 05 00:00:00 EDT 2017
HTLV-1 bZIP Factor Impairs Anti-viral Immunity by Inducing Co-inhibitory Molecule, T Cell Immunoglobulin and ITIM Domain (TIGIT).
PLoS pathogens
Yasuma K,Yasunaga J,Takemoto K,Sugata K,Mitobe Y,Takenouchi N,Nakagawa M,Suzuki Y,Matsuoka M
45-0048 was used in Flow cytometry/Cell sorting to show that HTLV-1 bZIP factor-induced TIGIT plays a pivotal role in attenuating host immune responses and shaping a microenvironment favourable to HTLV-1 in vitro.
Fri Jan 01 00:00:00 EST 2016
A phase I dose-escalation clinical trial of a peptide-based human papillomavirus therapeutic vaccine with Candida skin test reagent as a novel vaccine adjuvant for treating women with biopsy-proven cervical intraepithelial neoplasia 2/3.
Oncoimmunology
Greenfield WW,Stratton SL,Myrick RS,Vaughn R,Donnalley LM,Coleman HN,Mercado M,Moerman-Herzog AM,Spencer HJ,Andrews-Collins NR,Hitt WC,Low GM,Manning NA,McKelvey SS,Smith D,Smith MV,Phillips AM,Quick CM,Jeffus SK,Hutchins LF,Nakagawa M
45-0048 was used in Flow cytometry/Cell sorting to demonstrate the safety of a human papillomavirus therapeutic vaccine, PepCan.
Thu Oct 01 00:00:00 EDT 2015
Dysbiotic bacteria translocate in progressive SIV infection.
Mucosal immunology
Klase Z,Ortiz A,Deleage C,Mudd JC,Quiñones M,Schwartzman E,Klatt NR,Canary L,Estes JD,Brenchley JM
45-0048 was used in Flow cytometry/Cell sorting to examine the communities of bacteria within the gastrointestinal tract and systemic tissues of both healthy and SIV-infected Asian macaques.
Tue Sep 01 00:00:00 EDT 2015
Expression of GM-CSF in T Cells Is Increased in Multiple Sclerosis and Suppressed by IFN-β Therapy.
Journal of immunology (Baltimore, Md. : 1950)
Rasouli J,Ciric B,Imitola J,Gonnella P,Hwang D,Mahajan K,Mari ER,Safavi F,Leist TP,Zhang GX,Rostami A
45-0048 was used in Flow cytometry/Cell sorting to characterise GM-CSF production by T cells of MS patients and to determine the effect of IFN-β therapy on its production.
Mon Jun 01 00:00:00 EDT 2015
The Expression of Functional Vpx during Pathogenic SIVmac Infections of Rhesus Macaques Suppresses SAMHD1 in CD4+ Memory T Cells.
PLoS pathogens
Shingai M,Welbourn S,Brenchley JM,Acharya P,Miyagi E,Plishka RJ,Buckler-White A,Kwong PD,Nishimura Y,Strebel K,Martin MA
45-0048 was used in Flow cytometry/Cell sorting to indicate that autologous NAb responses in mothers and infants do not protect against mother to child transmission.
Fri May 01 00:00:00 EDT 2015
Increased peripheral proinflammatory T helper subsets contribute to cardiovascular complications in diabetic patients.
Mediators of inflammation
Zhao RX,Li WJ,Lu YR,Qin J,Wu CL,Tian M,He TY,Yi SN,Tang DQ,Sun L,Chen L
45-0048 was used in Flow cytometry/Cell sorting to assess the frequencies of various immune cells in diabetic patients with or without coronary atherosclerotic heart disease.
Thu Dec 18 00:00:00 EST 2014
Elevated peripheral frequencies of Th22 cells: a novel potent participant in obesity and type 2 diabetes.
PloS one
Zhao R,Tang D,Yi S,Li W,Wu C,Lu Y,Hou X,Song J,Lin P,Chen L,Sun L
45-0048 was used in Flow cytometry/Cell sorting to investigate the potential association between Th22 and the pathogenesis of obesity and type 2 diabetes.
Thu Sep 18 00:00:00 EDT 2014
Genome-wide analysis of T cell responses during acute and latent simian varicella virus infections in rhesus macaques.
Journal of virology
Haberthur K,Kraft A,Arnold N,Park B,Meyer C,Asquith M,Dewane J,Messaoudi I
45-0048 was used in Flow cytometry/Cell sorting to characterise the specificity of T cell responses during acute and latent simian varicella virus infection.
Fri Nov 01 00:00:00 EDT 2013
Ultradense array CGH and discovery of micro-copy number alterations and gene fusions in the cancer genome.
Methods in molecular biology (Clifton, N.J.)
Przybytkowski E,Aguilar-Mahecha A,Nabavi S,Tonellato PJ,Basik M
45-0048 was used in Immunocytochemistry-immunoflourescence to characterise the protocol steps and analysis required to obtain reliable array comparative array hybridisation results from clinical samples.
Mon Jul 29 00:00:00 EDT 2013
Assessment of immune interference, antagonism, and diversion following human immunization with biallelic blood-stage malaria viral-vectored vaccines and controlled malaria infection.
Journal of immunology (Baltimore, Md. : 1950)
Elias SC,Collins KA,Halstead FD,Choudhary P,Bliss CM,Ewer KJ,Sheehy SH,Duncan CJ,Biswas S,Hill AV,Draper SJ
45-0048 was used in Flow cytometry/Cell sorting to study the effects of both controlled malaria infection, and human immunisation with biallelic blood-stage malaria viral-vectored vaccines, on the immune system.
Fri Feb 01 00:00:00 EST 2013
Memory T cells in latent Mycobacterium tuberculosis infection are directed against three antigenic islands and largely contained in a CXCR3+CCR6+ Th1 subset.
PLoS pathogens
Lindestam Arlehamn CS,Gerasimova A,Mele F,Henderson R,Swann J,Greenbaum JA,Kim Y,Sidney J,James EA,Taplitz R,McKinney DM,Kwok WW,Grey H,Sallusto F,Peters B,Sette A
45-0048 was used in Flow cytometry/Cell sorting to further define the immunological footprint of Mycobacterium tuberculosis CD4 T cell recognition.
Tue Jan 01 00:00:00 EST 2013
Transcriptional profiling of experimental CD8(+) lymphocyte depletion in rhesus macaques infected with simian immunodeficiency virus SIVmac239.
Journal of virology
Bosinger SE,Jochems SP,Folkner KA,Hayes TL,Klatt NR,Silvestri G
45-0048 was used in Flow cytometry/Cell sorting to identify antiviral pathways perturbed by CD8+ T-cell depletion that may contribute to noncytolytic control of SIV replication.
Tue Jan 01 00:00:00 EST 2013
Reduced frequency of memory T cells and increased Th17 responses in patients with active tuberculosis.
Clinical and vaccine immunology : CVI
Marín ND,París SC,Rojas M,García LF
45-0048 was used in Flow cytometry/Cell sorting to investigate the alterations in Mycobacterium tuberculosis T cell subsets in patients with active tuberculosis, showing reduced frequency of memory T cells and increased Th17 responses.
Mon Oct 01 00:00:00 EDT 2012
IL-17 and IFN-γ expression in lymphocytes from patients with active tuberculosis correlates with the severity of the disease.
Journal of leukocyte biology
Jurado JO,Pasquinelli V,Alvarez IB,Peña D,Rovetta AI,Tateosian NL,Romeo HE,Musella RM,Palmero D,Chuluyán HE,García VE
45-0048 was used in Flow cytometry/Cell sorting to investigate the induction and regulation of IFN-γ and IL-17 during active human Mycobacterium tuberculosis infection.
Fri Jun 01 00:00:00 EDT 2012
CD4+CD8+ T cells represent a significant portion of the anti-HIV T cell response to acute HIV infection.
Journal of immunology (Baltimore, Md. : 1950)
Frahm MA,Picking RA,Kuruc JD,McGee KS,Gay CL,Eron JJ,Hicks CB,Tomaras GD,Ferrari G
45-0048 was used in Flow cytometry/Cell sorting to demonstrate that double-positive T-cells represent a highly reactive cell population during acute HIV infection.
Tue May 01 00:00:00 EDT 2012
SIV infection of rhesus macaques results in dysfunctional T- and B-cell responses to neo and recall Leishmania major vaccination.
Blood
Klatt NR,Vinton CL,Lynch RM,Canary LA,Ho J,Darrah PA,Estes JD,Seder RA,Moir SL,Brenchley JM
45-0048 was used in Flow cytometry/Cell sorting to show that SIV infection results in dysfunctional T-cell responses to neo and recall vaccinations.
Thu Nov 24 00:00:00 EST 2011
Human CD8⁺ and CD4⁺ T cell memory to lymphocytic choriomeningitis virus infection.
Journal of virology
Kotturi MF,Swann JA,Peters B,Arlehamn CL,Sidney J,Kolla RV,James EA,Akondy RS,Ahmed R,Kwok WW,Buchmeier MJ,Sette A
45-0048 was used in Flow cytometry/Cell sorting to analyse the T cell response to acute lymphocytic choriomeningitis virus infection.
Tue Nov 01 00:00:00 EDT 2011
Mechanisms underlying γδ T-cell subset perturbations in SIV-infected Asian rhesus macaques.
Blood
Harris LD,Klatt NR,Vinton C,Briant JA,Tabb B,Ladell K,Lifson J,Estes JD,Price DA,Hirsch VM,Brenchley JM
45-0048 was used in Flow cytometry/Cell sorting to study differing subsets of γδ T cells in HIV and Simian immunodeficiency virus-infected individuals.
Thu Nov 18 00:00:00 EST 2010
Disruption of intestinal CD4+ T cell homeostasis is a key marker of systemic CD4+ T cell activation in HIV-infected individuals.
Journal of immunology (Baltimore, Md. : 1950)
Gordon SN,Cervasi B,Odorizzi P,Silverman R,Aberra F,Ginsberg G,Estes JD,Paiardini M,Frank I,Silvestri G
45-0048 was used in Flow cytometry/Cell sorting to reveal an association between the destruction of intestinal CD4+ T-cell homeostasis and the level of systemic CD4+ T-cell activation.
Mon Nov 01 00:00:00 EDT 2010
Compromised gastrointestinal integrity in pigtail macaques is associated with increased microbial translocation, immune activation, and IL-17 production in the absence of SIV infection.
Mucosal immunology
Klatt NR,Harris LD,Vinton CL,Sung H,Briant JA,Tabb B,Morcock D,McGinty JW,Lifson JD,Lafont BA,Martin MA,Levine AD,Estes JD,Brenchley JM
45-0048 was used in Flow cytometry/Cell sorting to assess whether high basal levels of immune activation and microbial translocation exist in pigtail macaques.
Thu Jul 01 00:00:00 EDT 2010