Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
CD41 (platelet glycoprotein IIb, ITGA2B) is composed of two subunits -120 kDa a, alpha and 23 kDa b, beta- that interact with CD61 in the presence of calcium to form a functional adhesive protein receptor. CD41 is also involved in blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to a variety of proteins including von Willebrand factor, fibrinogen, fibronectin and vitronectin. CD41 is mainly expressed on megakaryocyte-platelet lineage, but generally belongs to the antigens that are expressed during the early stages of hematopoietic differentiation. Diseases associated with CD41 dysfunction include Glansmann Thrombasthenia, and Platelet type-16 bleeding disorder.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha IIb; CD41; form 1; form 2; GP IIb; GPalpha IIb; GPIIb; Integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; platelet glycoprotein IIb of IIb/II Ia complex; Platelet membrane glycoprotein IIb
Gene Aliases: AI172977; alphaIIb; CD41; CD41B; GpIIb; Itga2b
UniProt ID: (Mouse) Q9QUM0
Entrez Gene ID: (Mouse) 16399
Molecular Function: integrin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support