Product References
Myeloid Cell CK2 Regulates Inflammation and Resistance to Bacterial Infection.
Frontiers in immunology
Larson SR,Bortell N,Illies A,Crisler WJ,Matsuda JL,Lenz LL
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue Jul 27 00:00:00 EDT 2021
γ-Tocotrienol-Loaded Liposomes for Radioprotection from Hematopoietic Side Effects Caused by Radiotherapeutic Drugs.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Lee SG,Kalidindi TM,Lou H,Gangangari K,Punzalan B,Bitton A,Lee CJ,Vargas HA,Park S,Bodei L,Kharas MG,Singh VK,Kishore Pillarsetty NV,Larson SM
15-0452-83 was used in Flow Cytometry to report a novel radioprotector drug, a liposomal formulation of gamma-tocotrienol (GT3), or GT3-Nano for short, to mitigate bone marrow radiation damage during targeted radionuclide therapy (TRT).
Thu Apr 01 00:00:00 EDT 2021
The Interleukin (IL)-1R1 pathway is a critical negative regulator of PyMT-mediated mammary tumorigenesis and pulmonary metastasis.
Oncoimmunology
Dagenais M,Dupaul-Chicoine J,Douglas T,Champagne C,Morizot A,Saleh M
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Tpl2 Protects Against Fulminant Hepatitis Through Mobilization of Myeloid-Derived Suppressor Cells.
Frontiers in immunology
Xu J,Pei S,Wang Y,Liu J,Qian Y,Huang M,Zhang Y,Xiao Y
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Wed Oct 14 00:00:00 EDT 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
DNA-PKcs phosphorylation at the T2609 cluster alters the repair pathway choice during immunoglobulin class switch recombination.
Proceedings of the National Academy of Sciences of the United States of America
Crowe JL,Wang XS,Shao Z,Lee BJ,Estes VM,Zha S
15-0452 was used in Flow cytometry/Cell sorting to uncover a role of DNA-PKcs T2609 phosphorylation in promoting cNHEJ repair pathway choice during CSR.
Tue Sep 15 00:00:00 EDT 2020
HyperTRIBE uncovers increased MUSASHI-2 RNA binding activity and differential regulation in leukemic stem cells.
Nature communications
Nguyen DTT,Lu Y,Chu KL,Yang X,Park SM,Choo ZN,Chin CR,Prieto C,Schurer A,Barin E,Savino AM,Gourkanti S,Patel P,Vu LP,Leslie CS,Kharas MG
15-0452 was used in Flow cytometry/Cell sorting to globally map the mRNA targets of the RBP MSI2 in mammalian adult normal and malignant stem cells.
Fri Apr 24 00:00:00 EDT 2020
Conditional antibody expression to avoid central B cell deletion in humanized HIV-1 vaccine mouse models.
Proceedings of the National Academy of Sciences of the United States of America
Tian M,McGovern K,Cheng HL,Waddicor P,Rieble L,Dao M,Chen Y,Kimble MT,Cantor E,Manfredonia N,Judson R,Chapdelaine-Williams A,Cain DW,Haynes BF,Alt FW
15-0452 was used in Flow cytometry/Cell sorting to develop a new approach to test a vaccine which circumvents B cell developmental blocks.
Tue Apr 07 00:00:00 EDT 2020
Dysregulated TRAF3 and BCL2 Expression Promotes Multiple Classes of Mature Non-hodgkin B Cell Lymphoma in Mice.
Frontiers in immunology
Perez-Chacon G,Adrados M,Vallejo-Cremades MT,Lefebvre S,Reed JC,Zapata JM
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Fri Oct 25 00:00:00 EDT 2019
ATF3 Sustains IL-22-Induced STAT3 Phosphorylation to Maintain Mucosal Immunity Through Inhibiting Phosphatases.
Frontiers in immunology
Glal D,Sudhakar JN,Lu HH,Liu MC,Chiang HY,Liu YC,Cheng CF,Shui JW
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu Sep 26 00:00:00 EDT 2019
Phosphorylation at S2053 in Murine (S2056 in Human) DNA-PKcs Is Dispensable for Lymphocyte Development and Class Switch Recombination.
Journal of immunology (Baltimore, Md. : 1950)
Jiang W,Estes VM,Wang XS,Shao Z,Lee BJ,Lin X,Crowe JL,Zha S
15-0452-82 was used in Flow Cytometry to show that radiation can induce phosphorylation of murine DNA-PKcs at the corresponding S2053.
Mon Jul 01 00:00:00 EDT 2019
Bacteroides fragilis polysaccharide A induces IL-10 secreting B and T cells that prevent viral encephalitis.
Nature communications
Ramakrishna C,Kujawski M,Chu H,Li L,Mazmanian SK,Cantin EM
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue May 14 00:00:00 EDT 2019
Proapoptotic BIM Impacts B Lymphoid Homeostasis by Limiting the Survival of Mature B Cells in a Cell-Autonomous Manner.
Frontiers in immunology
Liu R,King A,Bouillet P,Tarlinton DM,Strasser A,Heierhorst J
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2019
The Hematopoietic Oxidase NOX2 Regulates Self-Renewal of Leukemic Stem Cells.
Cell reports
Adane B,Ye H,Khan N,Pei S,Minhajuddin M,Stevens BM,Jones CL,D'Alessandro A,Reisz JA,Zaberezhnyy V,Gasparetto M,Ho TC,Kelly KK,Myers JR,Ashton JM,Siegenthaler J,Kume T,Campbell EL,Pollyea DA,Becker MW,Jordan CT
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue Apr 02 00:00:00 EDT 2019
Nidogen-1 Contributes to the Interaction Network Involved in Pro-B Cell Retention in the Peri-sinusoidal Hematopoietic Stem Cell Niche.
Cell reports
Balzano M,De Grandis M,Vu Manh TP,Chasson L,Bardin F,Farina A,Sergé A,Bidaut G,Charbord P,Hérault L,Bailly AL,Cartier-Michaud A,Boned A,Dalod M,Duprez E,Genever P,Coles M,Bajenoff M,Xerri L,Aurrand-Lions M,Schiff C,Mancini SJC
15-0452 was used in Flow cytometry/Cell sorting to study pro-B cells in their support of stroll cells and their interaction network allowing their retention.
Tue Mar 19 00:00:00 EDT 2019
miR-143/145 differentially regulate hematopoietic stem and progenitor activity through suppression of canonical TGFβ signaling.
Nature communications
Lam J,van den Bosch M,Wegrzyn J,Parker J,Ibrahim R,Slowski K,Chang L,Martinez-Høyer S,Condorelli G,Boldin M,Deng Y,Umlandt P,Fuller M,Karsan A
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Wed Jun 20 00:00:00 EDT 2018
Adrenergic nerve degeneration in bone marrow drives aging of the hematopoietic stem cell niche.
Nature medicine
Maryanovich M,Zahalka AH,Pierce H,Pinho S,Nakahara F,Asada N,Wei Q,Wang X,Ciero P,Xu J,Leftin A,Frenette PS
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Retinoid-Sensitive Epigenetic Regulation of the Hoxb Cluster Maintains Normal Hematopoiesis and Inhibits Leukemogenesis.
Cell stem cell
Qian P,De Kumar B,He XC,Nolte C,Gogol M,Ahn Y,Chen S,Li Z,Xu H,Perry JM,Hu D,Tao F,Zhao M,Han Y,Hall K,Peak A,Paulson A,Zhao C,Venkatraman A,Box A,Perera A,Haug JS,Parmely T,Li H,Krumlauf R,Li L
15-0452 was used in Flow cytometry/Cell sorting to demonstrate roles for retinoid signaling and the DERARE in maintaining HSCs and preventing leukemogenesis by coordinate regulation of Hoxb genes.
Thu May 03 00:00:00 EDT 2018
Inactivation of mTORC1 Signaling in Osterix-Expressing Cells Impairs B-cell Differentiation.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Wang Y,Xiao M,Tao C,Chen J,Wang Z,Yang J,Chen Z,Zou Z,Liu A,Cai D,Jiang Y,Ding C,Li M,Bai X
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Host Tumor Suppressor p18INK4c Functions as a Potent Cell-Intrinsic Inhibitor of Murine Gammaherpesvirus 68 Reactivation and Pathogenesis.
Journal of virology
Niemeyer BF,Oko LM,Medina EM,Oldenburg DG,White DW,Cool CD,Clambey ET,van Dyk LF
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu Mar 15 00:00:00 EDT 2018
Tanshinone IIA and Astragaloside IV promote the angiogenesis of mesenchymal stem cell-derived endothelial cell-like cells via upregulation of Cx37, Cx40 and Cx43.
Experimental and therapeutic medicine
Li Z,Zhang S,Cao L,Li W,Ye YC,Shi ZX,Wang ZR,Sun LX,Wang JW,Jia LT,Wang W
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu Feb 01 00:00:00 EST 2018
DNA-binding of the Tet-transactivator curtails antigen-induced lymphocyte activation in mice.
Nature communications
Ottina E,Peperzak V,Schoeler K,Carrington E,Sgonc R,Pellegrini M,Preston S,Herold MJ,Strasser A,Villunger A
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Wed Oct 18 00:00:00 EDT 2017
EZH2 enables germinal centre formation through epigenetic silencing of CDKN1A and an Rb-E2F1 feedback loop.
Nature communications
Béguelin W,Rivas MA,Calvo Fernández MT,Teater M,Purwada A,Redmond D,Shen H,Challman MF,Elemento O,Singh A,Melnick AM
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu Oct 12 00:00:00 EDT 2017
Direct Activation of BAX by BTSA1 Overcomes Apoptosis Resistance in Acute Myeloid Leukemia.
Cancer cell
Reyna DE,Garner TP,Lopez A,Kopp F,Choudhary GS,Sridharan A,Narayanagari SR,Mitchell K,Dong B,Bartholdy BA,Walensky LD,Verma A,Steidl U,Gavathiotis E
15-0452 was used in Flow cytometry/Cell sorting to identify BTSA1, a pharmacologically optimised BAX activator that binds with high affinity and specificity to the N-terminal activation site and induces conformational changes to BAX leading to BAX-mediated apoptosis.
Mon Oct 09 00:00:00 EDT 2017
Multiple functional therapeutic effects of TnP: A small stable synthetic peptide derived from fish venom in a mouse model of multiple sclerosis.
PloS one
Komegae EN,Souza TA,Grund LZ,Lima C,Lopes-Ferreira M
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu Aug 17 00:00:00 EDT 2017
Irgm1 coordinately regulates autoimmunity and host defense at select mucosal surfaces.
JCI insight
Azzam KM,Madenspacher JH,Cain DW,Lai L,Gowdy KM,Rai P,Janardhan K,Clayton N,Cunningham W,Jensen H,Patel PS,Kearney JF,Taylor GA,Fessler MB
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu Aug 17 00:00:00 EDT 2017
Alkylator-Induced and Patient-Derived Xenograft Mouse Models of Therapy-Related Myeloid Neoplasms Model Clinical Disease and Suggest the Presence of Multiple Cell Subpopulations with Leukemia Stem Cell Activity.
PloS one
Jonas BA,Johnson C,Gratzinger D,Majeti R
15-0452 was used in Flow cytometry/Cell sorting to report several new t-AML/MDS mouse models that could potentially be used to further define disease pathogenesis and test novel therapeutics.
Wed Jul 26 00:00:00 EDT 2017
Epithelial Fli1 deficiency drives systemic autoimmunity and fibrosis: Possible roles in scleroderma.
The Journal of experimental medicine
Takahashi T,Asano Y,Sugawara K,Yamashita T,Nakamura K,Saigusa R,Ichimura Y,Toyama T,Taniguchi T,Akamata K,Noda S,Yoshizaki A,Tsuruta D,Trojanowska M,Sato S
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Mon Apr 03 00:00:00 EDT 2017
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Clec11a/osteolectin is an osteogenic growth factor that promotes the maintenance of the adult skeleton.
eLife
Yue R,Shen B,Morrison SJ
15-0452 was used in Flow cytometry/Cell sorting to study how Clec11a/Osteolectin maintains the adult skeleton by promoting the differentiation of mesenchymal progenitors into mature osteoblasts.
Tue Dec 13 00:00:00 EST 2016
Isolation and immunomodulatory activity of bursal peptide, a novel bursal peptide from the chicken bursa of Fabricius.
Journal of veterinary science
Liu XD,Qian Y,Jung YS,Chen PY
15-0452 was used in Flow cytometry/Cell sorting to provide novel insights into the use of a potential candidate reagent for B cell development and future immuno-pharmacological use.
Thu Sep 22 00:00:00 EDT 2016
Chk1 inhibition significantly potentiates activity of nucleoside analogs in TP53-mutated B-lymphoid cells.
Oncotarget
Zemanova J,Hylse O,Collakova J,Vesely P,Oltova A,Borsky M,Zaprazna K,Kasparkova M,Janovska P,Verner J,Kohoutek J,Dzimkova M,Bryja V,Jaskova Z,Brychtova Y,Paruch K,Trbusek M
15-0452 was used in Flow cytometry/Cell sorting to compare the effects of adding a Chk1 inhibitor to nucleoside analogs in p53-proficient and p53-defective cells.
Tue Sep 20 00:00:00 EDT 2016
Resident T Cells Are Unable To Control Herpes Simplex Virus-1 Activity in the Brain Ependymal Region during Latency.
Journal of immunology (Baltimore, Md. : 1950)
Menendez CM,Jinkins JK,Carr DJ
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Mon Aug 15 00:00:00 EDT 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
IgE-mediated enhancement of CD4(+) T cell responses requires antigen presentation by CD8α(-) conventional dendritic cells.
Scientific reports
Ding Z,Dahlin JS,Xu H,Heyman B
15-0452 was used in Flow cytometry/Cell sorting to elucidate the dendritic cell population involved in the presentation of IgE-Ag complexes during enhanced specific antibody and CD4+ T cell responses.
Thu Jun 16 00:00:00 EDT 2016
Aging-associated inflammation promotes selection for adaptive oncogenic events in B cell progenitors.
The Journal of clinical investigation
Henry CJ,Casás-Selves M,Kim J,Zaberezhnyy V,Aghili L,Daniel AE,Jimenez L,Azam T,McNamee EN,Clambey ET,Klawitter J,Serkova NJ,Tan AC,Dinarello CA,DeGregori J
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
Endothelial Gata5 transcription factor regulates blood pressure.
Nature communications
Messaoudi S,He Y,Gutsol A,Wight A,Hébert RL,Vilmundarson RO,Makrigiannis AP,Chalmers J,Hamet P,Tremblay J,McPherson R,Stewart AFR,Touyz RM,Nemer M
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Mon Nov 30 00:00:00 EST 2015
Sleep disruption impairs haematopoietic stem cell transplantation in mice.
Nature communications
Rolls A,Pang WW,Ibarra I,Colas D,Bonnavion P,Korin B,Heller HC,Weissman IL,de Lecea L
15-0452 was used in Flow cytometry/Cell sorting to provide insights into cellular and molecular mechanisms underlying the effects of sleep deprivation on HSCs.
Wed Oct 14 00:00:00 EDT 2015
Mitochondrial ATP transporter Ant2 depletion impairs erythropoiesis and B lymphopoiesis.
Cell death and differentiation
Cho J,Seo J,Lim CH,Yang L,Shiratsuchi T,Lee MH,Chowdhury RR,Kasahara H,Kim JS,Oh SP,Lee YJ,Terada N
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Overexpression of Bovine FcRn in Mice Enhances T-Dependent Immune Responses by Amplifying T Helper Cell Frequency and Germinal Center Enlargement in the Spleen.
Frontiers in immunology
Schneider Z,Jani PK,Szikora B,Végh A,Kövesdi D,Iliás A,Cervenak J,Balogh P,Kurucz I,Kacskovics I
15-0452 was used in Flow cytometry/Cell sorting to demonstrate that Tg mice that overexpress bFcRn developed efficient immune response against very low dose of antigen.
Mon Aug 10 00:00:00 EDT 2015
Disruption of p21-activated kinase 1 gene diminishes atherosclerosis in apolipoprotein E-deficient mice.
Nature communications
Singh NK,Kotla S,Dyukova E,Traylor JG,Orr AW,Chernoff J,Marion TN,Rao GN
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Wed Jun 24 00:00:00 EDT 2015
An anti-silencer- and SATB1-dependent chromatin hub regulates Rag1 and Rag2 gene expression during thymocyte development.
The Journal of experimental medicine
Hao B,Naik AK,Watanabe A,Tanaka H,Chen L,Richards HW,Kondo M,Taniuchi I,Kohwi Y,Kohwi-Shigematsu T,Krangel MS
15-0452 was used in Flow cytometry/Cell sorting to provide a novel framework for understanding ASE function for gene expression in double-positive thymocytes.
Mon May 04 00:00:00 EDT 2015
Chitin recognition via chitotriosidase promotes pathologic type-2 helper T cell responses to cryptococcal infection.
PLoS pathogens
Wiesner DL,Specht CA,Lee CK,Smith KD,Mukaremera L,Lee ST,Lee CG,Elias JA,Nielsen JN,Boulware DR,Bohjanen PR,Jenkins MK,Levitz SM,Nielsen K
15-0452 was used in Flow cytometry/Cell sorting to investigate Th2 cell responses to pulmonary fungal infection, showing that chitin recognition via chitotriosidase promotes pathologic type-2 helper T cell responses to cryptococcal infection.
Sun Mar 01 00:00:00 EST 2015
B-cell-intrinsic hepatitis C virus expression leads to B-cell-lymphomagenesis and induction of NF-κB signalling.
PloS one
Kasama Y,Mizukami T,Kusunoki H,Peveling-Oberhag J,Nishito Y,Ozawa M,Kohara M,Mizuochi T,Tsukiyama-Kohara K
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Mon Dec 22 00:00:00 EST 2014
Mouse mast cell protease-6 and MHC are involved in the development of experimental asthma.
Journal of immunology (Baltimore, Md. : 1950)
Cui Y,Dahlin JS,Feinstein R,Bankova LG,Xing W,Shin K,Gurish MF,Hallgren J
15-0452 was used in Flow cytometry/Cell sorting to compare the immune responses of BALB/c mice to mMCP-6(-/-) and mMCP-6(-/-)/H-2bc mice in a mouse model of experimental asthma.
Sat Nov 15 00:00:00 EST 2014
Transcription elongation factor ELL2 drives Ig secretory-specific mRNA production and the unfolded protein response.
Journal of immunology (Baltimore, Md. : 1950)
Park KS,Bayles I,Szlachta-McGinn A,Paul J,Boiko J,Santos P,Liu J,Wang Z,Borghesi L,Milcarek C
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Sat Nov 01 00:00:00 EDT 2014
Rhof promotes murine marginal zone B cell development.
Nagoya journal of medical science
Kishimoto M,Matsuda T,Yanase S,Katsumi A,Suzuki N,Ikejiri M,Takagi A,Ikawa M,Kojima T,Kunishima S,Kiyoi H,Naoe T,Matsushita T,Maruyama M
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
Infection mobilizes hematopoietic stem cells through cooperative NOD-like receptor and Toll-like receptor signaling.
Cell host & microbe
Burberry A,Zeng MY,Ding L,Wicks I,Inohara N,Morrison SJ,Núñez G
15-0452 was used in Flow cytometry/Cell sorting to investigate how infection mobilises hematopoietic stem cells through cooperative NOD-like receptor and Toll-like receptor signalling.
Wed Jun 11 00:00:00 EDT 2014
IκBε is a key regulator of B cell expansion by providing negative feedback on cRel and RelA in a stimulus-specific manner.
Journal of immunology (Baltimore, Md. : 1950)
Alves BN,Tsui R,Almaden J,Shokhirev MN,Davis-Turak J,Fujimoto J,Birnbaum H,Ponomarenko J,Hoffmann A
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue Apr 01 00:00:00 EDT 2014
The miR-126-VEGFR2 axis controls the innate response to pathogen-associated nucleic acids.
Nature immunology
Agudo J,Ruzo A,Tung N,Salmon H,Leboeuf M,Hashimoto D,Becker C,Garrett-Sinha LA,Baccarini A,Merad M,Brown BD
15-0452 was used in Western Blotting to identify the miR-126-VEGFR2 axis as an important regulator of the innate response that operates through multiscale control of pDCs.
Wed Jan 01 00:00:00 EST 2014
The miR-126-VEGFR2 axis controls the innate response to pathogen-associated nucleic acids.
Nature immunology
Agudo J,Ruzo A,Tung N,Salmon H,Leboeuf M,Hashimoto D,Becker C,Garrett-Sinha LA,Baccarini A,Merad M,Brown BD
15-0452 was used in Western Blotting to identify the miR-126-VEGFR2 axis as an important regulator of the innate response that operates through multiscale control of pDCs.
Wed Jan 01 00:00:00 EST 2014
Usp16 contributes to somatic stem-cell defects in Down's syndrome.
Nature
Adorno M,Sikandar S,Mitra SS,Kuo A,Nicolis Di Robilant B,Haro-Acosta V,Ouadah Y,Quarta M,Rodriguez J,Qian D,Reddy VM,Cheshier S,Garner CC,Clarke MF
15-0452 was used in Flow cytometry/Cell sorting to demonstrate a role for USP16 in antagonising the self-renewal and senescence pathways in Down's syndrome.
Thu Sep 19 00:00:00 EDT 2013
Partial depletion of regulatory T cells does not influence the inflammation caused by high dose hemi-body irradiation.
PloS one
Ma S,Richardson JA,Bitmansour A,Solberg TD,Pidikiti R,Song K,Stojadinovic S,Vitetta ES,Meyer JJ
15-0452 was used in Flow cytometry/Cell sorting to investigate the effects of partial depletion of regulatory T cells alongside ionizing radiation on inflammation and tissue injury in mice.
Mon Jul 29 00:00:00 EDT 2013
Reciprocal effects of rab7 deletion in activated and neglected T cells.
Autophagy
Roy SG,Stevens MW,So L,Edinger AL
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2013
Both NK cell-intrinsic and -extrinsic STAT1 signaling are required for NK cell response against vaccinia virus.
Journal of immunology (Baltimore, Md. : 1950)
Fortin C,Huang X,Yang Y
15-0452 was used in Flow cytometry/Cell sorting to explore the role of STAT1 in natural killer cell responses to vaccinia virus infection in vivo.
Mon Jul 01 00:00:00 EDT 2013
TDP2-dependent non-homologous end-joining protects against topoisomerase II-induced DNA breaks and genome instability in cells and in vivo.
PLoS genetics
Gómez-Herreros F,Romero-Granados R,Zeng Z,Alvarez-Quilón A,Quintero C,Ju L,Umans L,Vermeire L,Huylebroeck D,Caldecott KW,Cortés-Ledesma F
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu Jun 13 00:00:00 EDT 2013
FOXO3A directs a protective autophagy program in haematopoietic stem cells.
Nature
Warr MR,Binnewies M,Flach J,Reynaud D,Garg T,Malhotra R,Debnath J,Passegué E
15-0452 was used in Flow cytometry/Cell sorting to show how FOXO3A directs a protective autophagy program in haematopoietic stem cells.
Thu Feb 21 00:00:00 EST 2013
Coactivator-associated arginine methyltransferase 1 regulates fetal hematopoiesis and thymocyte development.
Journal of immunology (Baltimore, Md. : 1950)
Li J,Zhao Z,Carter C,Ehrlich LI,Bedford MT,Richie ER
15-0452 was used in Flow cytometry/Cell sorting to demonstrate how CARM1 is a key epigenetic regulator of hematopoiesis that affects multiple lineages at various stages of differentiation.
Tue Jan 15 00:00:00 EST 2013
Flow cytometry analysis of murine hematopoietic stem cells.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Mayle A,Luo M,Jeong M,Goodell MA
15-0452 was used in Flow cytometry/Cell sorting to discuss methods for rapid flow cytometric analysis of peripheral blood cell types and novel strategies for working with rare cell populations such as hematopoietic stem cells in the analysis of cell cycle status by BrdU, Ki-67, and Pyronin Y staining.
Tue Jan 01 00:00:00 EST 2013
Critical role of B cell lymphoma 10 in BAFF-regulated NF-κB activation and survival of anergic B cells.
Journal of immunology (Baltimore, Md. : 1950)
Yu M,Chen Y,He Y,Podd A,Fu G,Wright JA,Kleiman E,Khan WN,Wen R,Wang D
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2012
CD11c+ cells are required for antigen-induced increase of mast cells in the lung.
Journal of immunology (Baltimore, Md. : 1950)
Dahlin JS,Feinstein R,Cui Y,Heyman B,Hallgren J
15-0452 was used in Flow cytometry/Cell sorting to determine the contribution of Ag-stimulated CD11c(+) cells to the allergic response in asthmatics.
Mon Oct 15 00:00:00 EDT 2012
CD5-dependent CK2 activation pathway regulates threshold for T cell anergy.
Journal of immunology (Baltimore, Md. : 1950)
Sestero CM,McGuire DJ,De Sarno P,Brantley EC,Soldevila G,Axtell RC,Raman C
15-0452 was used in Flow cytometry/Cell sorting to determine the physiological significance of CD5-dependent casein kinase 2 activation in T cells.
Sat Sep 15 00:00:00 EDT 2012
Activation-induced cytidine deaminase-initiated off-target DNA breaks are detected and resolved during S phase.
Journal of immunology (Baltimore, Md. : 1950)
Hasham MG,Snow KJ,Donghia NM,Branca JA,Lessard MD,Stavnezer J,Shopland LS,Mills KD
15-0452 was used in Flow cytometry/Cell sorting to elucidate when off-target DNA breaks initiated by AID are detected and resolved.
Sat Sep 01 00:00:00 EDT 2012
Ymer acts as a multifunctional regulator in nuclear factor-κB and Fas signaling pathways.
Molecular medicine (Cambridge, Mass.)
Tsukiyama T,Matsuda-Tsukiyama M,Bohgaki M,Terai S,Tanaka S,Hatakeyama S
15-0452 was used in Flow cytometry/Cell sorting to explore the role of Ymer as a positive or negative regulator in several signalling pathways in mice.
Wed May 09 00:00:00 EDT 2012
Overexpression of Snai3 suppresses lymphoid- and enhances myeloid-cell differentiation.
European journal of immunology
Dahlem T,Cho S,Spangrude GJ,Weis JJ,Weis JH
15-0452 was used in Flow cytometry/Cell sorting to investigate the role of the transcriptional repressor Snai3 protein in the derivation of cells of the haematopoietic system.
Sun Apr 01 00:00:00 EDT 2012
PAD4-mediated neutrophil extracellular trap formation is not required for immunity against influenza infection.
PloS one
Hemmers S,Teijaro JR,Arandjelovic S,Mowen KA
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Tue Nov 15 00:00:00 EST 2011
An information theoretic, microfluidic-based single cell analysis permits identification of subpopulations among putatively homogeneous stem cells.
PloS one
Glotzbach JP,Januszyk M,Vial IN,Wong VW,Gelbard A,Kalisky T,Thangarajah H,Longaker MT,Quake SR,Chu G,Gurtner GC
15-0452 was used in Flow cytometry/Cell sorting to develop a novel method to characterize and predict transcriptional programs across individual homogeneous stem cells.
Wed Nov 02 00:00:00 EDT 2011
Requirement for complement in antibody responses is not explained by the classic pathway activator IgM.
Proceedings of the National Academy of Sciences of the United States of America
Rutemark C,Alicot E,Bergman A,Ma M,Getahun A,Ellmerich S,Carroll MC,Heyman B
15-0452 was used in Flow cytometry/Cell sorting to investigate the role of the classical pathway activator IgM in complement activated antibody responses.
Tue Oct 25 00:00:00 EDT 2011
Type I interferon drives dendritic cell apoptosis via multiple BH3-only proteins following activation by PolyIC in vivo.
PloS one
Fuertes Marraco SA,Scott CL,Bouillet P,Ives A,Masina S,Vremec D,Jansen ES,O'Reilly LA,Schneider P,Fasel N,Shortman K,Strasser A,Acha-Orbea H
15-0452 was used in Flow cytometry/Cell sorting to elucidate the possible apoptotic mechanisms that control activated dendritic cell lifespan in response to PAMPs.
Fri Sep 30 00:00:00 EDT 2011
Langerhans cells regulate cutaneous injury by licensing CD8 effector cells recruited to the skin.
Blood
Bennett CL,Fallah-Arani F,Conlan T,Trouillet C,Goold H,Chorro L,Flutter B,Means TK,Geissmann F,Chakraverty R
15-0452 was used in Flow cytometry/Cell sorting to explore the role of Langerhans cells in the development of tissue injury using a tractable model of cutaneous graft-versus-host disease.
Thu Jun 30 00:00:00 EDT 2011
Nuclear export of the NF-κB inhibitor IκBα is required for proper B cell and secondary lymphoid tissue formation.
Immunity
Wuerzberger-Davis SM,Chen Y,Yang DT,Kearns JD,Bates PW,Lynch C,Ladell NC,Yu M,Podd A,Zeng H,Huang TT,Wen R,Hoffmann A,Wang D,Miyamoto S
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Fri Feb 25 00:00:00 EST 2011
Nonhematopoietic antigen blocks memory programming of alloreactive CD8+ T cells and drives their eventual exhaustion in mouse models of bone marrow transplantation.
The Journal of clinical investigation
Flutter B,Edwards N,Fallah-Arani F,Henderson S,Chai JG,Sivakumaran S,Ghorashian S,Bennett CL,Freeman GJ,Sykes M,Chakraverty R
15-0452 was used in Flow cytometry/Cell sorting to explore the role of recipient antigen expression by nonhaematopoietic cells in the failure to sustain effective cytotoxic T lymphocyte immunity following blood transplantation.
Mon Nov 01 00:00:00 EDT 2010
Additive and global functions of HoxA cluster genes in mesoderm derivatives.
Developmental biology
Di-Poï N,Koch U,Radtke F,Duboule D
15-0452 was used in Flow cytometry/Cell sorting to report that mutant mice lacking the entire HoxA cluster in mesodermal lineages display the expected spectrum of postnatal respiratory, cardiac and urogenital defects, previously reported for single gene mutations.
Sat May 15 00:00:00 EDT 2010
Gas6 deficiency in recipient mice of allogeneic transplantation alleviates hepatic graft-versus-host disease.
Blood
Burnier L,Saller F,Kadi L,Brisset AC,Sugamele R,Baudino L,Bono F,Herbert JM,Carmeliet P,Schapira M,Izui S,Angelillo-Scherrer A
15-0452 was used in Flow cytometry/Cell sorting to study the effects of growth arrest-specific gene 6 deficiency on hepatic graft-versus-host disease in recipient mice of allogeneic transplantation.
Thu Apr 22 00:00:00 EDT 2010
In vivo requirement for Atg5 in antigen presentation by dendritic cells.
Immunity
Lee HK,Mattei LM,Steinberg BE,Alberts P,Lee YH,Chervonsky A,Mizushima N,Grinstein S,Iwasaki A
15-0452 was used in Flow cytometry/Cell sorting to investigate how dendritic cells utilise autophagic machinery to optimally present extracellular microbial antigens for MHC II molecules.
Fri Feb 26 00:00:00 EST 2010
A critical role of TAK1 in B-cell receptor-mediated nuclear factor kappaB activation.
Blood
Schuman J,Chen Y,Podd A,Yu M,Liu HH,Wen R,Chen ZJ,Wang D
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Thu May 07 00:00:00 EDT 2009
Flow-cytometric phosphoprotein analysis reveals agonist and temporal differences in responses of murine hematopoietic stem/progenitor cells.
PloS one
Kalaitzidis D,Neel BG
15-0452 was used in Flow cytometry/Cell sorting to develop a method to study intracellular signaling in murine HSC/progenitor cell (HPC)-enriched populations, showing agonist and temporal differences in responses of HSC/HPC.
Tue Feb 10 00:00:00 EST 2009
Stable form of JAB1 enhances proliferation and maintenance of hematopoietic progenitors.
The Journal of biological chemistry
Mori M,Yoneda-Kato N,Yoshida A,Kato JY
15-0452 was used in Flow cytometry/Cell sorting to investigate how the expression and function of JAB1 are critical for the proliferation and maintenance of hematopoietic progenitors.
Fri Oct 24 00:00:00 EDT 2008
IL-7 specifies B cell fate at the common lymphoid progenitor to pre-proB transition stage by maintaining early B cell factor expression.
Journal of immunology (Baltimore, Md. : 1950)
Kikuchi K,Kasai H,Watanabe A,Lai AY,Kondo M
15-0452 was used in Flow cytometry/Cell sorting to investigate when IL-7 stimulation is necessary in hemato-/lymphopoiesis in adult mice, showing that IL-7 specifies B cell fate at the common lymphoid progenitor to pre-proB transition stage by maintaining early B cell factor expression.
Tue Jul 01 00:00:00 EDT 2008
p27 deficiency cooperates with Bcl-2 but not Bax to promote T-cell lymphoma.
PloS one
Cheng N,van de Wetering CI,Knudson CM
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Wed Apr 02 00:00:00 EDT 2008
Phospholipase Cgamma2 contributes to light-chain gene activation and receptor editing.
Molecular and cellular biology
Bai L,Chen Y,He Y,Dai X,Lin X,Wen R,Wang D
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2007
Phospholipase Cgamma2 contributes to light-chain gene activation and receptor editing.
Molecular and cellular biology
Bai L,Chen Y,He Y,Dai X,Lin X,Wen R,Wang D
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2007
miR-150, a microRNA expressed in mature B and T cells, blocks early B cell development when expressed prematurely.
Proceedings of the National Academy of Sciences of the United States of America
Zhou B,Wang S,Mayr C,Bartel DP,Lodish HF
15-0452 was used in Immunofluorescence to study how miR-150 can down-regulate mRNAs important for B cell function, and how it can block further B cell development when expressed prematurely.
Tue Apr 24 00:00:00 EDT 2007
miR-150, a microRNA expressed in mature B and T cells, blocks early B cell development when expressed prematurely.
Proceedings of the National Academy of Sciences of the United States of America
Zhou B,Wang S,Mayr C,Bartel DP,Lodish HF
15-0452 was used in Immunofluorescence to study how miR-150 can down-regulate mRNAs important for B cell function, and how it can block further B cell development when expressed prematurely.
Tue Apr 24 00:00:00 EDT 2007
The impact of altered p53 dosage on hematopoietic stem cell dynamics during aging.
Blood
Dumble M,Moore L,Chambers SM,Geiger H,Van Zant G,Goodell MA,Donehower LA
15-0452 was used in Flow cytometry/Cell sorting to study how altering p53 dosage during ageing impacts hematopoietic stem cell dynamics.
Thu Feb 15 00:00:00 EST 2007
Essential role of phospholipase C gamma 2 in early B-cell development and Myc-mediated lymphomagenesis.
Molecular and cellular biology
Wen R,Chen Y,Bai L,Fu G,Schuman J,Dai X,Zeng H,Yang C,Stephan RP,Cleveland JL,Wang D
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2006
Developmental switch of mouse hematopoietic stem cells from fetal to adult type occurs in bone marrow after birth.
Proceedings of the National Academy of Sciences of the United States of America
Kikuchi K,Kondo M
15-0452 was used in Flow cytometry/Cell sorting to demonstrate how foetal haematopoietic stem cells may acquire adult characteristics between 1 and 2 weeks after birth in mouse bone marrow.
Tue Nov 21 00:00:00 EST 2006
Hypersensitivity and loss of disease site targeting caused by antibody responses to PEGylated liposomes.
Molecular therapy : the journal of the American Society of Gene Therapy
Judge A,McClintock K,Phelps JR,Maclachlan I
15-0452 was used in Flow cytometry/Cell sorting to report that the in vivo efficacy and safety of PEGylated liposomes carrying therapeutic plasmid DNA can be severely compromised following repeat administration.
Wed Feb 01 00:00:00 EST 2006
Distinct roles of phosphoinositide-3 kinase and phospholipase Cgamma2 in B-cell receptor-mediated signal transduction.
Molecular and cellular biology
Dai X,Chen Y,Schuman J,Hua Z,Adamson JW,Wen R,Wang D
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2006
A role for brain-derived neurotrophic factor in B cell development.
Journal of neuroimmunology
Schuhmann B,Dietrich A,Sel S,Hahn C,Klingenspor M,Lommatzsch M,Gudermann T,Braun A,Renz H,Nockher WA
Wed Jun 01 00:00:00 EDT 2005
Beta-catenin is dispensable for hematopoiesis and lymphopoiesis.
The Journal of experimental medicine
Cobas M,Wilson A,Ernst B,Mancini SJ,MacDonald HR,Kemler R,Radtke F
15-0452 was used in Flow cytometry/Cell sorting to investigate an essential role for beta-catenin during hematopoiesis and lymphopoiesis.
Mon Jan 19 00:00:00 EST 2004
Impaired immune responses and B-cell proliferation in mice lacking the Id3 gene.
Molecular and cellular biology
Pan L,Sato S,Frederick JP,Sun XH,Zhuang Y
Published figure using CD45R (B220) monoclonal antibody (Product # 15-0452-82) in Flow Cytometry
Wed Sep 01 00:00:00 EDT 1999