Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (Paraffin) (IHC (P)) |
0.25-0.5 µg/mL | - |
Immunocytochemistry (ICC/IF) |
1-2 µg/mL | - |
Flow Cytometry (Flow) |
1-2 µg/10^6 cells | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Mouse / IgG2a, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
T200, 797 |
Immunogen |
Human T-lymphocytes |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
200 µg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.4, with 0.05% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
4° C |
Immunohistochemistry (PFA fixed): incubate antibody for 30 min at RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.
CD45RO (UCHL1, ubiquitin carboxyl terminal esterase L1) is the shortest isoform of a receptor-type protein tyrosine phosphatase, CD45 glycoprotein. CD45 is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases, promotes cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45RO is expressed on macrophages, CD8+ T cells, activated T cells and myeloma cells. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD45; CD45 antigen; L-CA; Leukocyte common antigen; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen
Gene Aliases: B220; CD45; CD45R; GP180; L-CA; LCA; LY5; PTPRC; T200
UniProt ID: (Human) P08575
Entrez Gene ID: (Human) 5788
Molecular Function:
protein phosphatase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support