Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Flow Cytometry (Flow) |
0.125 µg/test | View 21 publications 21 publications |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Published species |
Human, Mouse |
Host/Isotype |
Armenian hamster / IgG |
Recommended Isotype Control |
Armenian Hamster IgG Isotype Control (eBio299Arm), APC, eBioscience™ |
Class |
Monoclonal |
Type |
Antibody |
Clone |
HM48-1 |
Conjugate |
APC
|
Excitation/Emission Profile |
View spectra |
Form |
Liquid |
Concentration |
0.2 mg/mL |
Purification |
Affinity chromatography |
Storage buffer |
PBS, pH 7.2 |
Contains |
0.09% sodium azide |
Storage conditions |
4° C, store in dark, DO NOT FREEZE! |
RRID |
AB_469408 |
Description: The HM48-1 monoclonal antibody reacts with the mouse CD48 antigen; also known as BCM1, Blast-1 (human), and OX-45 (rat). CD48, a member of the SLAM family and Ig superfamily, is a 45 kDa GPI-linked glycoprotein expressed on the majority of hematopoietic cells. Recent publications have reported differential expression of members of the SLAM family including CD48, CD150, and CD244 among functionally distinct bone marrow hematopoietic progenitors providing a useful tool for prediction of the primitiveness of hematopoietic progenitors based on the expression of these SLAM family members. Hematopoietic stem cells (HSC) are highly purified as CD150(+)CD244(-)CD48(-) cells while non-self-renewing multipotent hematopoietic progenitors (MPP) are CD244(+)CD150(-)CD48(-) and the most restricted progenitors are CD48(+)CD244(+)CD150(-). CD48 plays a critical role in adhesion and T cell activation. In the mouse, the primary counter-receptors for CD48 are CD2 and CD244. HM48-1 is reported to modulate in vitro and in vivo CD48 functions including blocking the CD48/CD2 and CD48/CD244 interactions, inhibiting the proliferative response of mitogen-activated spleen cells, providing a costimulation signal for T cells activated in vitro through their TCR, and prolonging cardiac allograft survival in vivo.
Applications Reported: This HM48-1 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This HM48-1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Filtration: 0.2 µm post-manufacturing filtered.
CD48 (Blast-1) belongs to the CD2 subset of the Ig superfamily that includes CD2, CD2F-10, CD58, CD84, CD150, CD229, CD244 and others. These molecules bind to the same or another members of the Ig family, thus mediating homotypic or heterotypic adhesion. CD48 is a GPI-anchored protein broadly expressed on hematopoietic cells and serves as a high affinity ligand for 2B4 and low affinity ligand for CD2. 2B4-CD48 interaction among NK cells and NK-T cells regulates cell proliferation. Further, signaling through CD48 results in eosinophil activation and CD48 expression is increased in several infectious diseases. CD48 is strongly expressed on lymphocytes and monocytes, weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of marker for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2. 2B4-CD48 interaction may be involved in the pathology of allergic airway diseases and provide a pharmacological target for the treatment of diseases such as allergic rhinitis and asthma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: B-cell activation marker; BCM1 surface antigen; BLAST-1; CD48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48d; HM48-1; MRC OX-45 surface antigen; sCD 48; sCD48; sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48
Gene Aliases: AI449234; AW610730; Bcm-1; BCM1; BLAST; BLAST-1; BLAST1; Cd48; MEM-102; Sgp-60; SLAMF2
UniProt ID: (Mouse) P18181
Entrez Gene ID: (Mouse) 12506
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support