Applications | Tested Dilution | Publications |
---|---|---|
Immunohistochemistry (Frozen) (IHC (F)) |
Assay-dependent | - |
Immunocytochemistry (ICC/IF) |
Assay-dependent | View 1 publication 1 publication |
Flow Cytometry (Flow) |
1:40-1:80 | - |
Immunoprecipitation (IP) |
Assay-dependent | - |
Miscellaneous PubMed (Misc) |
- | View 1 publication 1 publication |
Product Specifications | |
---|---|
Species Reactivity |
Chicken, Human |
Published species |
Human |
Host/Isotype |
Mouse / IgG1 |
Class |
Monoclonal |
Type |
Antibody |
Clone |
23C6 |
Immunogen |
Osteoclasts from osteoclastomas |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS |
Contains |
0.09% sodium azide |
Storage conditions |
4°C or -20°C if preferred |
RRID |
AB_2537984 |
This antibody does not react with bovine, pig or rabbit.
The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
CD51 and CD61 molecules (alpha V and beta 3 integrins) form a complex. This complex binds vitronection at the RGD sequence and can also bind fibrinogen, von Willebrand factor, thrombospondin, fibronectin, osteopontin and collagen. Reacts with osteoclasts, placenta, melanoma cell lines and weakly with platelets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen CD61; antigen identified by monoclonal antibody L230; CD51; CD61; GPIIIa; integrin alpha V; integrin alpha-5 beta-3; Integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; Integrin beta; integrin beta 3; Integrin beta-3; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); Platelet membrane glycoprotein IIIa; Vitronectin receptor; vitronectin receptor alpha subunit; Vitronectin receptor subunit alpha
Gene Aliases: BDPLT16; BDPLT2; CD51; CD61; GP3A; GPIIIa; GT; integrin; ITGAV; ITGB3; MSK8; VNRA; VTNR
UniProt ID: (Human) P06756, (Chicken) P26008, (Human) P05106
Entrez Gene ID: (Human) 3685, (Chicken) 396420, (Human) 3690, (Chicken) 374209
Molecular Function:
integrin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support