Product References
Usefulness of the octreotide test in Japanese patients for predicting the presence/absence of somatostatin receptor 2 expression in insulinomas.
Endocrine journal
Nakamura A,Mitsuhashi T,Takano Y,Miyoshi H,Kameda H,Nomoto H,Nagai S,Hatanaka Y,Shimizu C,Terauchi Y,Atsumi T
MA135249 was used in immunohistochemistry - paraffin section to assess the relationship between the results of the octreotide test and somatostatin receptor 2 expression in insulinoma patients to predict insulinomas in Japanese patients
Fri Dec 23 00:00:00 EST 2016
Variable Expression of Neural Cell Adhesion Molecule Isoforms in Renal Tissue: Possible Role in Incipient Renal Fibrosis.
PloS one
Marković-Lipkovski J,Životić M,Müller CA,Tampe B,Ćirović S,Vještica J,Tomanović N,Zeisberg M,Müller GA
MA1-35249 was used in immunohistochemistry - paraffin section to learn the possible role of incipient renal fibrosis by variable expression of neural cell adhesion molecule isoforms in renal tissue
Wed May 18 00:00:00 EDT 2016
Role of cd56 and e-cadherin expression in the differential diagnosis of papillary thyroid carcinoma and suspected follicular-patterned lesions of the thyroid: the prognostic importance of e-cadherin.
International journal of clinical and experimental pathology
Ceyran AB,Şenol S,Şimşek BÇ,Sağıroğlu J,Aydın A
MA1-35249 was used in immunohistochemistry - paraffin section to discuss how to distinguish follicular variant of PTC from other follicular-patterned lesions of the thyroid.
Thu Mar 24 00:00:00 EDT 2016
CD30+ extranodal natural killer/T-cell lymphoma mimicking phlegmonous myositis: A case report.
Oncology letters
Yang YJ,Li YX,Liu YB,Yang M,Liu K
MA1-35249 was used in immunohistochemistry (paraffin) to report on a case of CD30-positive extranodal natural killer/T-cell lymphoma.
Thu May 01 00:00:00 EDT 2014
A Case of Distal Epithelioid Sarcoma of the Thumb Expressing Podoplanin, TLE1 and Ca 125.
Case reports in pathology
Karagkounis G,Argyrakos T,Charkiolakis G,Castana O,Rontogianni D
MA1-35249 was used in immunohistochemistry to characterize the case of a distal epithelioid sarcoma arising in the thumb of a 14-year-old girl
Wed May 22 00:00:00 EDT 2013
Serous effusion cytology of extranodal natural killer/T-cell lymphoma.
Cytopathology : official journal of the British Society for Clinical Cytology
Su XY,Huang J,Jiang Y,Tang Y,Li GD,Liu WP
MA1-35249 was used in flow cytometry to characterize extranodal natural killer/T-cell lymphoma, nasal type using patient samples.
Sun Apr 01 00:00:00 EDT 2012
Extraosseous (extramedullary) plasmacytomas: a clinicopathologic and immunophenotypic study of 32 Chinese cases.
Diagnostic pathology
Zuo Z,Tang Y,Bi CF,Zhang WY,Zhao S,Wang XQ,Yang QP,Zou LQ,Liu WP
MA1-35249 was used in immunohistochemistry - paraffin section to determine the clinicopathologic features and diagnosis of extraosseous plasmacytomas
Mon Dec 19 00:00:00 EST 2011
Subcutaneous panniculitis-like T-cell lymphoma: a clinical and pathologic study of 14 korean patients.
Annals of dermatology
Lee DW,Yang JH,Lee SM,Won CH,Chang S,Lee MW,Choi JH,Moon KC
MA1-35249 was used in immunohistochemistry to study subcutaneous panniculitis-like T-cell lymphomas in Korean patients
Mon Aug 01 00:00:00 EDT 2011
Goblet cell carcinoid of the appendix: a clinicopathological and immunohistochemical study of 26 cases from southwest china.
International journal of surgical pathology
MA1-35249 was used in immunohistochemistry - paraffin section to report the clinicopathological and immunohistochemical features of goblet cell carcinoid
Wed Dec 01 00:00:00 EST 2010
Primary central nervous system peripheral T-cell lymphoma in a child.
Fetal and pediatric pathology
Gualco G,Wludarski S,Hayashi-Silva L,Medeiros Filho P,Veras G,Bacchi CE
MA1-35249 was used in immunohistochemistry to report on a case of primary central nervous system peripheral T-cell lymphoma in a child
Tue Sep 28 00:00:00 EDT 2010
Atypical carcinoid with prominent mucinous stroma: a hitherto unreported variant of pulmonary neuroendocrine tumor.
Endocrine pathology
Nannini N,Bertolini F,Cavazza A,Casali C,Mengoli MC,Rossi G
MA1-35249 was used in immunohistochemistry to report on a case of atypical carcinoid with prominent mucinous stroma
Tue Jun 01 00:00:00 EDT 2010
Primary plasmacytoma of the cervix in a 21-year-old female patient.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists
Schor AP,Moraes MP,Bisson FW,Bisson MA,Luiz OM,Bacchi CE
MA1-35249 was used in immunohistochemistry to report on a case of primary plasmacytoma of the cervix
Sat May 01 00:00:00 EDT 2010
EGFR and K-ras mutations along the spectrum of pulmonary epithelial tumors of the lung and elaboration of a combined clinicopathologic and molecular scoring system to predict clinical responsiveness to EGFR inhibitors.
American journal of clinical pathology
Sartori G,Cavazza A,Sgambato A,Marchioni A,Barbieri F,Longo L,Bavieri M,Murer B,Meschiari E,Tamberi S,Cadioli A,Luppi F,Migaldi M,Rossi G
MA1-35249 was used in immunohistochemistry to develop a combined clinicopathologic and molecular scoring system for assessing the tumor responses to EGFR inhibitors
Wed Apr 01 00:00:00 EDT 2009
Composite lymphoma: EBV-positive classic Hodgkin lymphoma and peripheral T-cell lymphoma: a case report.
Applied immunohistochemistry & molecular morphology : AIMM
Gualco G,Chioato L,Van Den Berg A,Weiss LM,Bacchi CE
MA1-35249 was used in immunohistochemistry to report on a case of composite EBV-positive classic Hodgkin lymphoma and peripheral T-cell lymphoma
Thu Jan 01 00:00:00 EST 2009
TTF-1, cytokeratin 7, 34betaE12, and CD56/NCAM immunostaining in the subclassification of large cell carcinomas of the lung.
American journal of clinical pathology
Rossi G,Marchioni A,Milani M,Scotti R,Foroni M,Cesinaro A,Longo L,Migaldi M,Cavazza A
MA1-35249 was used in immunohistochemistry to study the use of a number of immunohistochemical markers for the subclassification of large cell carcinomas of the lung
Wed Dec 01 00:00:00 EST 2004
The prognostic role of c-kit protein expression in resected large cell neuroendocrine carcinoma of the lung.
The Annals of thoracic surgery
Casali C,Stefani A,Rossi G,Migaldi M,Bettelli S,Parise A,Morandi U
MA1-35249 was used in immunohistochemistry to study the prognostic value of c-kit protein expression in resected large cell neuroendocrine carcinoma
Thu Jan 01 00:00:00 EST 2004
Desmoplastic and spindle cell melanomas express protein markers of the neural crest but not of later committed stages of Schwann cell differentiation.
Journal of cutaneous pathology
Huttenbach Y,Prieto VG,Reed JA
MA1-35249 was used in immunohistochemistry to investigate the expression of unique protein markers in desmoplastic and spindle cell melanomas
Tue Oct 01 00:00:00 EDT 2002