Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
0.01 µg/mL | - |
Immunohistochemistry (Paraffin) (IHC (P)) |
5 µg/mL | - |
Flow Cytometry (Flow) |
10 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Host/Isotype |
Goat / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide with sequence C-KASWTRPEKQETLD. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.5 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
TBS, pH 7.3, with 0.5% BSA |
Contains |
0.02% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
Additional Information: This antibody is expected to recognize reported isoform 2 (NP_851996.2) only.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
CD56 (NCAM, neural cell adhesion molecule) is a transmembrane glycoprotein of the immunoglobulin family that serves as an adhesive molecule and is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa. CD56 is found on T cells and NK cells, and is involved in cell migration, axonal growth, pathfinding and synaptic plasticity. Polysialic modification results in reduction of CD56-mediated cell adhesion. Through its extracellular region, CD56 mediates homophilic and heterophilic interactions by binding extracellular matrix components such as laminin and integrins. CD56 is expressed on most neuroectodermal derived cell lines, tissues and neoplasms such as retinoblastoma, medulloblastoma, astrocytomas and neuroblastoma. Further, CD56 is a widely used neuroendocrine marker with a high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen recognized by monoclonal antibody 5.1H11; CD-56; CD56; E NCAM; N CAM1; N-CAM-1; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM
Gene Aliases: CD56; MSK39; NCAM; NCAM1
UniProt ID: (Human) P13591
Entrez Gene ID: (Human) 4684
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support