Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store product undiluted at 4ºC in the dark.
For FACS analysis, use 20 µL of the working dilution to label 1x10^6 cells in 100 µL.
Mouse anti Human CD58 antibody, clone MEM-63 recognizes human CD58, also known as LFA-3. CD58 is a membrane glycoprotein of approximately 55-70 kDa.
This gene encodes a member of the immunoglobulin superfamily. The encoded protein is a ligand of the T lymphocyte CD2 protein, and functions in adhesion and activation of T lymphocytes. The protein is localized to the plasma membrane. Alternatively spliced transcript variants have been described. [provided by RefSeq, Jan 2009]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: Ag3; CD305; CD58; CD58 antigen, (lymphocyte function-associated antigen 3); FLJ23181; FLJ43722; LAIR-1; LAIR1; Lymphocyte function-associated antigen 3; RP5-1086K13.2; Surface glycoprotein LFA-3
Gene Aliases: ag3; CD58; LFA-3; LFA3
UniProt ID: (Human) P19256
Entrez Gene ID: (Human) 965
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support