Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD66a (CEACAM1) Antibodies

          Invitrogen

          CD66a (CEACAM1) Monoclonal Antibody (CC1), eBioscience™

          Advanced Verification
          9 Published Figures
          6 References
          View all (67) CD66a (CEACAM1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD66a (CEACAM1) Monoclonal Antibody (CC1), eBioscience™

          • Antibody Testing Data (2)
          • Published Figures (9)
          • Advanced Verification (2)
          CD66a (CEACAM1) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CD66a (CEACAM1) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD66a (CEACAM1) Antibody (14-0661-82) in WB

          Western blot was performed using Anti-CD66a (CEACAM1) Monoclonal Antibody (Product # 14-0661-82) and a 110 kDa band corresponding to CEACAM1 was observed across tissues tested except Mouse Lung. A 25 kDa band corresponding to circulating IgG was observed as well. Tissue extracts (30ug lysate) of Mouse Liver (Lane 1), Mouse Small Intestine (Lane 2) and Mouse Lung (Lane 3) were electrophoresed using NuPAGE® 4-12 % Bis-Tris gel (Product # NP0322BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # IB23001) by... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD66a (CEACAM1) Antibody in Western Blot (WB)
          CD66a (CEACAM1) Antibody in Western Blot (WB)
          CD66a (CEACAM1) Antibody in Western Blot (WB)
          CD66a (CEACAM1) Antibody in Flow Cytometry (Flow)
          CD66a (CEACAM1) Antibody in Flow Cytometry (Flow)
          CD66a (CEACAM1) Antibody in Flow Cytometry (Flow)
          CD66a (CEACAM1) Antibody in Flow Cytometry (Flow)
          CD66a (CEACAM1) Antibody in Flow Cytometry (Flow)
          CD66a (CEACAM1) Antibody in Flow Cytometry (Flow)
          CD66a (CEACAM1) Antibody in Flow Cytometry (Flow)
          CD66a (CEACAM1) Antibody in Flow Cytometry (Flow)
          CD66a (CEACAM1) Antibody
          CD66a (CEACAM1) Antibody

          Product Details

          14-0661-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          -

          Flow Cytometry (Flow)

          0.25 µg/test
          View 4 publications 4 publications

          Functional Assay (Functional)

          Assay-Dependent
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          CC1

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • FITC
          • PE 
          • PE-Cyanine7
          • Super Bright 436
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1210503

          Product Specific Information

          Description: The monoclonal antibody CC1 recognizes CD66a, also known as carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1), biliary glycoprotein, and BPG. Expression of CD66a is found on brush borders, epithelial, and endothelial cells. In hematopoietic cells expression is found abundantly on B cells, as well as some NKs, monocytes, DCs, and granulocytes. Although low levels of mRNA have been identified in T cells in humans, resting mouse T lymphocytes are not reported to express CD66a, as confirmed by lack of staining with CC1 antibody. In humans, expression levels of CD66a have been used to identify malignancies. CD66a plays a key role as a regulator of BCR activation of B lymphocytes.

          An alternate allele, CEACAM1b, is expressed in SJL mice; therefore, CC1 does not stain SJL tissue. The monoclonal CC1 has been shown to block viral infection and also enhance B cell proliferation when combined with IgM crosslinking.

          Applications Reported: This CC1 antibody has been reported for use in flow cytometric analysis, and immunohistology staining of paraffin embedded tissue sections.

          Applications Tested: This CC1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CEACAM1 is a member of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily. Two subgroups of the CEA family, the CEA cell adhesion molecules and the pregnancy-specific glycoproteins, are located within a 1.2 Mb cluster on the long arm of chromosome 19. Eleven pseudogenes of the CEA cell adhesion molecule subgroup are also found in the cluster. The encoded protein was originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. The encoded protein mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. Multiple cellular activities have been attributed to the encoded protein, including roles in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses. Multiple transcript variants encoding different isoforms have been reported, but the full-length nature of only two has been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: BGP-1; Biliary glycoprotein 1; Biliary glycoprotein D; carcinoembryonic antigen 1; carcinoembryonic antigen 7; Carcinoembryonic antigen-related cell adhesion molecule 1; CD66a; CEA cell adhesion molecule 1; CEA-related cell adhesion molecule 1; Cell adhesion molecule CEACAM1; hepatitis virus (MHV-4) susceptibility; hepatitis virus receptor; MHV-R; MHVR1; Murine hepatitis virus receptor

          View more View less

          Gene Aliases: bb-1; Bgp; Bgp1; C-CAM; Cc1; CD66a; Cea-1; Cea-7; Cea1; Cea7; Ceacam1; Hv-2; Hv2; mCEA1; Mhv-1; MHVR; MHVR1; mmCGM1; mmCGM1a; mmCGM2

          View more View less

          UniProt ID: (Mouse) P31809

          View more View less

          Entrez Gene ID: (Mouse) 26365

          View more View less

          Function(s)
          virus receptor activity protein binding protein kinase binding protein homodimerization activity virion binding immunoglobulin receptor superfamily
          Process(es)
          negative regulation of cytokine production homophilic cell adhesion via plasma membrane adhesion molecules viral process calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules negative regulation of interleukin-2 production positive regulation of T cell proliferation negative regulation of T cell proliferation positive regulation of MAP kinase activity negative regulation by host of viral process positive regulation by host of viral process negative regulation by host of viral genome replication negative regulation of JNK cascade positive regulation of JNK cascade viral entry into host cell Peyer's patch development positive regulation of immunoglobulin secretion positive regulation of NFAT protein import into nucleus positive regulation of activation-induced cell death of T cells positive regulation of CD4-positive, alpha-beta T cell activation positive regulation of CD4-positive, alpha-beta T cell proliferation positive regulation of CD8-positive, alpha-beta T cell activation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
          git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.39.1-2025.11.99-1.0