Product References
Temporal Characterization of Microglia/Macrophage Phenotypes in a Mouse Model of Neonatal Hypoxic-Ischemic Brain Injury.
Frontiers in cellular neuroscience
Hellström Erkenstam N,Smith PL,Fleiss B,Nair S,Svedin P,Wang W,Boström M,Gressens P,Hagberg H,Brown KL,Sävman K,Mallard C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Sat Jan 29 00:00:00 EST 2022
MiR-103 protects from recurrent spontaneous abortion via inhibiting STAT1 mediated M1 macrophage polarization.
International journal of biological sciences
Zhu X,Liu H,Zhang Z,Wei R,Zhou X,Wang Z,Zhao L,Guo Q,Zhang Y,Chu C,Wang L,Li X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Fri Nov 12 00:00:00 EST 2021
A bilirubin-derived nanomedicine attenuates the pathological cascade of pulmonary fibrosis.
Biomaterials
Keum H,Kim D,Kim J,Kim TW,Whang CH,Jung W,Jon S
25-0862-82 was used in Flow Cytometry to report the therapeutic potential of nanoparticles derived from the endogenous antioxidant and anti-inflammatory bile acid, bilirubin (BRNPs), in treating pulmonary fibrosis in a bleomycin-induced mouse model of the disease.
Sun Aug 01 00:00:00 EDT 2021
Platelet-Like Fusogenic Liposome-Mediated Targeting Delivery of miR-21 Improves Myocardial Remodeling by Reprogramming Macrophages Post Myocardial Ischemia-Reperfusion Injury.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Tan H,Song Y,Chen J,Zhang N,Wang Q,Li Q,Gao J,Yang H,Dong Z,Weng X,Wang Z,Sun D,Yakufu W,Pang Z,Huang Z,Ge J
25-0862-82 was used in Flow Cytometry to construct a platelet-like fusogenic liposome (PLPs) to deliver contents to the cytoplasm of target cells.
Sun Aug 01 00:00:00 EDT 2021
Adrenomedullin 2 improves bone regeneration in type 1 diabetic rats by restoring imbalanced macrophage polarization and impaired osteogenesis.
Stem cell research & therapy
Wang F,Kong L,Wang W,Shi L,Wang M,Chai Y,Xu J,Kang Q
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Thu May 13 00:00:00 EDT 2021
Salmonella-Based Therapy Targeting Indoleamine 2,3-Dioxygenase Restructures the Immune Contexture to Improve Checkpoint Blockade Efficacy.
Biomedicines
Ebelt ND,Zuniga E,Marzagalli M,Zamloot V,Blazar BR,Salgia R,Manuel ER
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Wed Dec 16 00:00:00 EST 2020
Mesenchymal stem cell-secreted extracellular vesicles carrying TGF-β1 up-regulate miR-132 and promote mouse M2 macrophage polarization.
Journal of cellular and molecular medicine
Wang Y,Han B,Wang Y,Wang C,Zhang H,Xue J,Wang X,Niu T,Niu Z,Chen Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2020
Mesenchymal stem cell-secreted extracellular vesicles carrying TGF-β1 up-regulate miR-132 and promote mouse M2 macrophage polarization.
Journal of cellular and molecular medicine
Wang Y,Han B,Wang Y,Wang C,Zhang H,Xue J,Wang X,Niu T,Niu Z,Chen Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2020
Mutation of an L-Type Calcium Channel Gene Leads to T Lymphocyte Dysfunction.
Frontiers in immunology
Fenninger F,Han J,Stanwood SR,Nohara LL,Arora H,Choi KB,Munro L,Pfeifer CG,Shanina I,Horwitz MS,Jefferies WA
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Thu Oct 29 00:00:00 EDT 2020
Ginsenoside Rg3 Mitigates Atherosclerosis Progression in Diabetic apoE-/- Mice by Skewing Macrophages to the M2 Phenotype.
Frontiers in pharmacology
Guo M,Xiao J,Sheng X,Zhang X,Tie Y,Wang L,Zhao L,Ji X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Senolytics prevent mt-DNA-induced inflammation and promote the survival of aged organs following transplantation.
Nature communications
Iske J,Seyda M,Heinbokel T,Maenosono R,Minami K,Nian Y,Quante M,Falk CS,Azuma H,Martin F,Passos JF,Niemann CU,Tchkonia T,Kirkland JL,Elkhal A,Tullius SG
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Thu Aug 27 00:00:00 EDT 2020
Excessive Costimulation Leads to Dysfunction of Adoptively Transferred T Cells.
Cancer immunology research
Wijewarnasuriya D,Bebernitz C,Lopez AV,Rafiq S,Brentjens RJ
25-0862 was used in Flow cytometry/Cell sorting to show excessive CD28 and 4-1BB co-stimulation can drive chimeric antigen receptor T-cell dysfunction.
Mon Jun 01 00:00:00 EDT 2020
MicroRNA-9 Fine-Tunes Dendritic Cell Function by Suppressing Negative Regulators in a Cell-Type-Specific Manner.
Cell reports
Cordeiro B,Jeon P,Boukhaled GM,Corrado M,Lapohos O,Roy DG,Williams K,Jones RG,Gruenheid S,Sagan SM,Krawczyk CM
25-0862 was used in Flow cytometry/Cell sorting to demonstrate that miR-9 facilitates the transition of DCs from steady state to mature state by regulating the expression of several negative regulators of DC function in a cell-type-specific manner.
Tue May 05 00:00:00 EDT 2020
FcεRI γ-Chain Negatively Modulates Dectin-1 Responses in Dendritic Cells.
Frontiers in immunology
Pan YG,Yu YL,Lin CC,Lanier LL,Chu CL
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Wed Nov 20 00:00:00 EST 2019
Sestrin2 Suppresses Classically Activated Macrophages-Mediated Inflammatory Response in Myocardial Infarction through Inhibition of mTORC1 Signaling.
Frontiers in immunology
Yang K,Xu C,Zhang Y,He S,Li D
25-0862 was used in Flow cytometry/Cell sorting to indicate the importance of Sestrin2 for suppression of M1 macrophage-mediated cardiac inflammation after myocardial infarction.
Wed Nov 20 00:00:00 EST 2019
Doxorubicin-polyglycerol-nanodiamond conjugate is a cytostatic agent that evades chemoresistance and reverses cancer-induced immunosuppression in triple-negative breast cancer.
Journal of nanobiotechnology
Yuan SJ,Xu YH,Wang C,An HC,Xu HZ,Li K,Komatsu N,Zhao L,Chen X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Thu Oct 17 00:00:00 EDT 2019
Arp2/3 complex-driven spatial patterning of the BCR enhances immune synapse formation, BCR signaling and B cell activation.
eLife
Bolger-Munro M,Choi K,Scurll JM,Abraham L,Chappell RS,Sheen D,Dang-Lawson M,Wu X,Priatel JJ,Coombs D,Hammer JA,Gold MR
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Mon Jun 03 00:00:00 EDT 2019
Leptin produced by obesity-altered adipose stem cells promotes metastasis but not tumorigenesis of triple-negative breast cancer in orthotopic xenograft and patient-derived xenograft models.
Breast cancer research : BCR
Sabol RA,Bowles AC,Côté A,Wise R,O'Donnell B,Matossian MD,Hossain FM,Burks HE,Del Valle L,Miele L,Collins-Burow BM,Burow ME,Bunnell BA
25-0862 was used in Flow cytometry/Cell sorting to study the mechanisms by which obesity promotes tumour progression.
Wed May 22 00:00:00 EDT 2019
Low-Dose Apatinib Optimizes Tumor Microenvironment and Potentiates Antitumor Effect of PD-1/PD-L1 Blockade in Lung Cancer.
Cancer immunology research
Zhao S,Ren S,Jiang T,Zhu B,Li X,Zhao C,Jia Y,Shi J,Zhang L,Liu X,Qiao M,Chen X,Su C,Yu H,Zhou C,Zhang J,Camidge DR,Hirsch FR
25-0862 was used in Flow cytometry/Cell sorting to analyze the impact of apatinib, a smallmolecule VEGFR2-TKI, on the TME in murine Lewis lung carcinoma (LLC) models at different doses alone and in combination with an anti–PD-L1.
Mon Apr 01 00:00:00 EDT 2019
cGAS/STING/TBK1/IRF3 Signaling Pathway Activates BMDCs Maturation Following Mycobacterium bovis Infection.
International journal of molecular sciences
Li Q,Liu C,Yue R,El-Ashram S,Wang J,He X,Zhao D,Zhou X,Xu L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Tue Feb 19 00:00:00 EST 2019
Efficient oral vaccination by bioengineering virus-like particles with protozoan surface proteins.
Nature communications
Serradell MC,Rupil LL,Martino RA,Prucca CG,Carranza PG,Saura A,Fernández EA,Gargantini PR,Tenaglia AH,Petiti JP,Tonelli RR,Reinoso-Vizcaino N,Echenique J,Berod L,Piaggio E,Bellier B,Sparwasser T,Klatzmann D,Luján HD
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Mon Jan 21 00:00:00 EST 2019
Immunological Properties of Neural Crest Cells Derived from Human Induced Pluripotent Stem Cells.
Stem cells and development
Fujii S,Yoshida S,Inagaki E,Hatou S,Tsubota K,Takahashi M,Shimmura S,Sugita S
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2019
STAT3 Inhibition Combined with CpG Immunostimulation Activates Antitumor Immunity to Eradicate Genetically Distinct Castration-Resistant Prostate Cancers.
Clinical cancer research : an official journal of the American Association for Cancer Research
Moreira D,Adamus T,Zhao X,Su YL,Zhang Z,White SV,Swiderski P,Lu X,DePinho RA,Pal SK,Kortylewski M
25-0862 was used in Flow cytometry/Cell sorting to show a novel strategy combining STAT3 inhibition with Toll-like Receptor 9 (TLR9) stimulation to unleash immune response against prostate cancers regardless of the genetic background.
Sat Dec 01 00:00:00 EST 2018
Targeting macrophage and microglia activation with colony stimulating factor 1 receptor inhibitor is an effective strategy to treat injury-triggered neuropathic pain.
Molecular pain
Lee S,Shi XQ,Fan A,West B,Zhang J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Mon Oct 22 00:00:00 EDT 2018
T cell microvilli constitute immunological synaptosomes that carry messages to antigen-presenting cells.
Nature communications
Kim HR,Mun Y,Lee KS,Park YJ,Park JS,Park JH,Jeon BN,Kim CH,Jun Y,Hyun YM,Kim M,Lee SM,Park CS,Im SH,Jun CD
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Fri Sep 07 00:00:00 EDT 2018
Discrimination of the heterogeneity of bone marrow‑derived dendritic cells.
Molecular medicine reports
Wang J,Dai X,Hsu C,Ming C,He Y,Zhang J,Wei L,Zhou P,Wang CY,Yang J,Gong N
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Wed Nov 01 00:00:00 EDT 2017
Calreticulin is an effective immunologic adjuvant to tumor-associated antigens.
Experimental and therapeutic medicine
Wang J,Gao ZP,Qin S,Liu CB,Zou LL
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Sun Oct 01 00:00:00 EDT 2017
IRF6 Regulates Alternative Activation by Suppressing PPARγ in Male Murine Macrophages.
Endocrinology
Li C,Ying W,Huang Z,Brehm T,Morin A,Vella AT,Zhou B
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Fri Sep 01 00:00:00 EDT 2017
Sec61 blockade by mycolactone inhibits antigen cross-presentation independently of endosome-to-cytosol export.
Proceedings of the National Academy of Sciences of the United States of America
Grotzke JE,Kozik P,Morel JD,Impens F,Pietrosemoli N,Cresswell P,Amigorena S,Demangel C
25-0862 was used in Flow cytometry/Cell sorting to analyse Sec61's contribution to antigen cross-presentation, ER-associated degradation, and transport of internalised antigens into the cytosol using a newly identified Sec61 blocker.
Tue Jul 18 00:00:00 EDT 2017
Metformin Inhibits Advanced Glycation End Products-Induced Inflammatory Response in Murine Macrophages Partly through AMPK Activation and RAGE/NFκB Pathway Suppression.
Journal of diabetes research
Zhou Z,Tang Y,Jin X,Chen C,Lu Y,Liu L,Shen C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Tue Jun 06 00:00:00 EDT 2017
Repulsive guidance molecule a blockade exerts the immunoregulatory function in DCs stimulated with ABP and LPS.
Human vaccines & immunotherapeutics
Xu X,Gao Y,Zhai Z,Zhang S,Shan F,Feng J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Tue Aug 02 00:00:00 EDT 2016
D-Alanylation of Teichoic Acids and Loss of Poly-N-Acetyl Glucosamine in Staphylococcus aureus during Exponential Growth Phase Enhance IL-12 Production in Murine Dendritic Cells.
PloS one
Lund LD,Ingmer H,Frøkiær H
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Thu Jul 14 00:00:00 EDT 2016
CD80 and CD86 knockdown in dendritic cells regulates Th1/Th2 cytokine production in asthmatic mice.
Experimental and therapeutic medicine
Li JG,DU YM,Yan ZD,Yan J,Zhuansun YX,Chen R,Zhang W,Feng SL,Ran PX
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
Towards programming immune tolerance through geometric manipulation of phosphatidylserine.
Biomaterials
Roberts RA,Eitas TK,Byrne JD,Johnson BM,Short PJ,McKinnon KP,Reisdorf S,Luft JC,DeSimone JM,Ting JP
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2015
Reducing progression of experimental lupus nephritis via inhibition of the B7/CD28 signaling pathway.
Molecular medicine reports
Huang L,Kong Y,Wang J,Sun J,Shi Q,Qiu YH
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Injectable cryogel-based whole-cell cancer vaccines.
Nature communications
Bencherif SA,Warren Sands R,Ali OA,Li WA,Lewin SA,Braschler TM,Shih TY,Verbeke CS,Bhatta D,Dranoff G,Mooney DJ
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Wed Aug 12 00:00:00 EDT 2015
Nizatidine, a small molecular compound, enhances killed H5N1 vaccine cell-mediated responses and protects mice from lethal viral challenge.
Human vaccines & immunotherapeutics
Wang S,Wu B,Xue J,Wang M,Chen R,Wang B
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Mon Jan 05 00:00:00 EST 2015
Injectable, spontaneously assembling, inorganic scaffolds modulate immune cells in vivo and increase vaccine efficacy.
Nature biotechnology
Kim J,Li WA,Choi Y,Lewin SA,Verbeke CS,Dranoff G,Mooney DJ
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2015
Lung dendritic cells undergo maturation and polarization towards a T helper type 2-stimulating phenotype in a mouse model of asthma: Role of nerve growth factor.
Experimental and therapeutic medicine
Qin Q,Wang Z,Pan P,Cao Z,Xia Q,Tan H,Hu C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Sat Nov 01 00:00:00 EDT 2014
BCG vaccine mediated reduction in the MHC-II expression of macrophages and dendritic cells is reversed by activation of Toll-like receptors 7 and 9.
Cellular immunology
Bakhru P,Sirisaengtaksin N,Soudani E,Mukherjee S,Khan A,Jagannath C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
Histone deacetylase inhibitors suppress immune activation in primary mouse microglia.
Journal of neuroscience research
Kannan V,Brouwer N,Hanisch UK,Regen T,Eggen BJ,Boddeke HW
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Sun Sep 01 00:00:00 EDT 2013
Neutralization of TSLP inhibits airway remodeling in a murine model of allergic asthma induced by chronic exposure to house dust mite.
PloS one
Chen ZG,Zhang TT,Li HT,Chen FH,Zou XL,Ji JZ,Chen H
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Tue Aug 20 00:00:00 EDT 2013
Orientia tsutsugamushi subverts dendritic cell functions by escaping from autophagy and impairing their migration.
PLoS neglected tropical diseases
Choi JH,Cheong TC,Ha NY,Ko Y,Cho CH,Jeon JH,So I,Kim IK,Choi MS,Kim IS,Cho NH
25-0862 was used in Flow cytometry/Cell sorting to investigate how Orientia tsutsugamushi can exploit dendritic cells as replication reservoirs and impair functional maturation of these cells during infection in mammals.
Wed Jun 19 00:00:00 EDT 2013
Immunogenicity and protective capacity of a virosomal respiratory syncytial virus vaccine adjuvanted with monophosphoryl lipid A in mice.
PloS one
Kamphuis T,Meijerhof T,Stegmann T,Lederhofer J,Wilschut J,de Haan A
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Wed Sep 19 00:00:00 EDT 2012
Diabetic mice display a delayed adaptive immune response to Mycobacterium tuberculosis.
Journal of immunology (Baltimore, Md. : 1950)
Vallerskog T,Martens GW,Kornfeld H
25-0862 was used in Flow cytometry/Cell sorting to conclude that TB increased susceptibility in DM results from a delayed innate immune response to the presence of M. tuberculosis-infected alveolar macrophages.
Tue Jun 01 00:00:00 EDT 2010
Identification of regulatory functions for 4-1BB and 4-1BBL in myelopoiesis and the development of dendritic cells.
Nature immunology
Lee SW,Park Y,So T,Kwon BS,Cheroutre H,Mittler RS,Croft M
Published figure using CD86 (B7-2) monoclonal antibody (Product # 25-0862-82) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2008