Product References
Immunogenicity of undifferentiated and differentiated allogeneic mouse mesenchymal stem cells.
Journal of tissue engineering
Mukonoweshuro B,Brown CJ,Fisher J,Ingham E
MA1-70062 was used in flow cytometry to study the immunogenicity and immunomodulatory properties of bone marrow-derived mesenchymal stem cells using an allogeneic mouse model.
Fri May 09 00:00:00 EDT 2014
Accumulation of effector CD4 T cells during type 2 immune responses is negatively regulated by Stat6.
Journal of immunology (Baltimore, Md. : 1950)
King SB,Knorn AM,Ohnmacht C,Voehringer D
MA1-70062 was used in flow cytometry to investigate the effect of Stat6 in T cell expansion, survival, and recruitment to the lung
Tue Jan 15 00:00:00 EST 2008
B-lymphocyte depletion ameliorates Sjögren's syndrome in Id3 knockout mice.
Immunology
Hayakawa I,Tedder TF,Zhuang Y
MA1-70062 was used in flow cytometry to investigate the role of B cells in Sjogren's syndrome in a mouse model
Sat Sep 01 00:00:00 EDT 2007
Influenza A virus elevates active cathepsin B in primary murine DC.
International immunology
Burster T,Giffon T,Dahl ME,Björck P,Bogyo M,Weber E,Mahmood K,Lewis DB,Mellins ED
MA1-70062 was used in flow cytometry to suggest that the influenza A virus affects the MHC class II antigen-processing pathway
Tue May 01 00:00:00 EDT 2007
Stepwise development of committed progenitors in the bone marrow that generate functional T cells in the absence of the thymus.
Journal of immunology (Baltimore, Md. : 1950)
García-Ojeda ME,Dejbakhsh-Jones S,Chatterjea-Matthes D,Mukhopadhyay A,BitMansour A,Weissman IL,Brown JM,Strober S
MA1-70062 was used in flow cytometry to conclude that bone marrow committed T cell progenitors can generate functional T cells via an extrathymic pathway
Sat Oct 01 00:00:00 EDT 2005
Development and functional analysis of eosinophils from murine embryonic stem cells.
British journal of haematology
Hamaguchi-Tsuru E,Nobumoto A,Hirose N,Kataoka S,Fujikawa-Adachi K,Furuya M,Tominaga A
MA1-70062 was used in flow cytometry to generate and characterize eosinophils from murine embryonic stem cells.
Mon Mar 01 00:00:00 EST 2004
Development of thymic microenvironments in vitro is oxygen-dependent and requires permanent presence of T-cell progenitors.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Germeraad WT,Kawamoto H,Itoi M,Jiang Y,Amagai T,Katsura Y,van Ewijk W
MA1-70062 was used in flow cytometry to investigate factors that mediate induction and maintenance of the 3D configuration of the epithelial network in fetal thymic lobes in vitro.
Mon Sep 01 00:00:00 EDT 2003
Early defect prethymic in bone marrow T cell progenitors in athymic nu/nu mice.
Journal of immunology (Baltimore, Md. : 1950)
Chatterjea-Matthes D,García-Ojeda ME,Dejbakhsh-Jones S,Jerabek L,Manz MG,Weissman IL,Strober S
MA1-70062 was used in flow cytometry to examine early T cell progenitor development in the nu/nu bone marrow.
Fri Aug 01 00:00:00 EDT 2003
Donor-type CD4(+)CD25(+) regulatory T cells suppress lethal acute graft-versus-host disease after allogeneic bone marrow transplantation.
The Journal of experimental medicine
Hoffmann P,Ermann J,Edinger M,Fathman CG,Strober S
MA1-70062 was used in flow cytometry to report that the balance of donor-type CD4(+)CD25(+) T(reg) and conventional CD4(+)CD25(-) T cells determines the outcome of acute graft-versus-host disease.
Mon Aug 05 00:00:00 EDT 2002
Predominance of NK1.1+TCR alpha beta+ or DX5+TCR alpha beta+ T cells in mice conditioned with fractionated lymphoid irradiation protects against graft-versus-host disease: "natural suppressor" cells.
Journal of immunology (Baltimore, Md. : 1950)
Lan F,Zeng D,Higuchi M,Huie P,Higgins JP,Strober S
MA1-70062 was used in flow cytometry to develop a nonmyeloablative host conditioning regimen for transplantation.
Wed Aug 15 00:00:00 EDT 2001
Clonable progenitors committed to the T lymphocyte lineage in the mouse bone marrow; use of an extrathymic pathway.
Proceedings of the National Academy of Sciences of the United States of America
Dejbakhsh-Jones S,Garcia-Ojeda ME,Chatterjea-Matthes D,Zeng D,Strober S
MA1-70062 was used in flow cytometry to identify progenitors that can result in T cells via an extrathymic pathway.
Tue Jun 19 00:00:00 EDT 2001
Allogeneic bone marrow cells that facilitate complete chimerism and eliminate tumor cells express both CD8 and T-cell antigen receptor-alphabeta.
Blood
Lan F,Zeng D,Huie P,Higgins JP,Strober S
MA1-70062 was used in flow cytometry to test if specific subsets of cells in allogeneic bone marrow transplants can treat the BCL(1) B-cell lymphoma without inducing graft verses host disease.
Fri Jun 01 00:00:00 EDT 2001
Cryptomonad algal phycobiliproteins as fluorochromes for extracellular and intracellular antigen detection by flow cytometry.
Cytometry
Telford WG,Moss MW,Morseman JP,Allnutt FC
MA1-70062 was used in flow cytometry to assess cryptomonad-derived phycobiliproteins for their applicability to flow cytometry.
Tue May 01 00:00:00 EDT 2001
Immature multipotent hemopoietic progenitors lacking long-term bone marrow-reconstituting activity in the aorta-gonad-mesonephros region of murine day 10 fetuses.
Journal of immunology (Baltimore, Md. : 1950)
Ohmura K,Kawamoto H,Lu M,Ikawa T,Ozaki S,Nakao K,Katsura Y
MA1-70062 was used in flow cytometry to examine multipotent progenitors from early fetuses.
Thu Mar 01 00:00:00 EST 2001
Developmental dissociation of thymic dendritic cell and thymocyte lineages revealed in growth factor receptor mutant mice.
Proceedings of the National Academy of Sciences of the United States of America
Rodewald HR,Brocker T,Haller C
MA1-70062 was used in flow cytometry to test if thymocytes and thymic dendritic cell develop from a common intrathymic progenitor.
Tue Dec 21 00:00:00 EST 1999
Identification of an early T cell progenitor for a pathway of T cell maturation in the bone marrow.
Proceedings of the National Academy of Sciences of the United States of America
Dejbakhsh-Jones S,Strober S
MA1-70062 was used in flow cytometry to identify a rare population of cells in the adult mouse bone marrow that generates CD4(+) and CD8(+) TCRalphabeta(+) T cells.
Tue Dec 07 00:00:00 EST 1999
Commitment of common T/Natural killer (NK) progenitors to unipotent T and NK progenitors in the murine fetal thymus revealed by a single progenitor assay.
The Journal of experimental medicine
Ikawa T,Kawamoto H,Fujimoto S,Katsura Y
MA1-70062 was used in flow cytometry to describe a clonal system that supports the development of T and natural killer cells evenly.
Mon Dec 06 00:00:00 EST 1999
Emergence of T, B, and myeloid lineage-committed as well as multipotent hemopoietic progenitors in the aorta-gonad-mesonephros region of day 10 fetuses of the mouse.
Journal of immunology (Baltimore, Md. : 1950)
Ohmura K,Kawamoto H,Fujimoto S,Ozaki S,Nakao K,Katsura Y
MA1-70062 was used in flow cytometry to investigate the aorta-gonad-mesonephros region as a source of hemopoietic progenitors.
Mon Nov 01 00:00:00 EST 1999
Emergence of T cell progenitors without B cell or myeloid differentiation potential at the earliest stage of hematopoiesis in the murine fetal liver.
Journal of immunology (Baltimore, Md. : 1950)
Kawamoto H,Ohmura K,Fujimoto S,Katsura Y
MA1-70062 was used in flow cytometry to assess if progenitors colonizing the thymus are T cell lineage restricted.
Mon Mar 01 00:00:00 EST 1999
Surface antigen expression in spleen cells of C57B1/6 mice during ageing: influence of sex and parity.
Clinical and experimental immunology
Barrat F,Lesourd BM,Louise A,Boulouis HJ,Vincent-Naulleau S,Thibault D,Sanaa M,Neway T,Pilet CH
MA1-70062 was used in flow cytometry to test if pregnancies and gender influence the cellar changes observed during ageing in mice.
Sat Mar 01 00:00:00 EST 1997
Isolation of the most immature population of murine fetal thymocytes that includes progenitors capable of generating T, B, and myeloid cells.
The Journal of experimental medicine
Hattori N,Kawamoto H,Katsura Y
MA1-70062 was used in flow cytometry to report that FcR is a useful marker to isolate the most immature population of murine fetal thymic cells.
Fri Nov 01 00:00:00 EST 1996
Exclusive development of T cell neoplasms in mice transplanted with bone marrow expressing activated Notch alleles.
The Journal of experimental medicine
Pear WS,Aster JC,Scott ML,Hasserjian RP,Soffer B,Sklar J,Baltimore D
MA1-70062 was used in flow cytometry to report that TAN1 is an oncoprotein.
Wed May 01 00:00:00 EDT 1996
Effects of the lpr mutation on elimination and inactivation of self-reactive B cells.
Journal of immunology (Baltimore, Md. : 1950)
Rathmell JC,Goodnow CC
MA1-70062 was used in flow cytometry to investigate the fate of autoreactive B cells in lpr/lpr mice.
Thu Sep 15 00:00:00 EDT 1994