Sequence of this protein is as follows: MGSAKSVPVT PARPPPHNKH LARVADPRSP SAGILRTPIQ VESSPQPGLP AGEQLEGLKH AQDSDPRSPT LGIARTPMKT SSGDPPSPLV KQLSEVFETE DSKSNLPPEP VLPPEAPLSS ELDLPLGTQL SVEEQMPPWN QTEFPSKQVF SKEEARQPTE TPVASQSSDK PSRDPETPRS SGSMRNRWKP NSSKVLGRSP LTILQDDNSP GTLTLRQGKR PSPLSENVSE LKEGAILGTG RLLKTGGRAW EQGQDHDKEN QHFPLVES
F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDCA 3; Cell division cycle-associated protein 3; gene rich cluster, C8; Gene-rich cluster protein C8; GRCC 8; MGC2577; TOME 1; TOME-1; trigger of mitotic entry 1; Trigger of mitotic entry protein 1
Gene Aliases: C8; CDCA3; GRCC8; TOME-1; TOME1
UniProt ID: (Human) Q99618
Entrez Gene ID: (Human) 83461
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support