Sequence of this protein is as follows: TGQPLWPGKS DVDQLYLIIR TLGKLIPRHQ SIFKSNGFFH GISIPEPEDM ETLEEKFSDV HPVALNFMKG CLKMNPDDRL TCSQLLESSY FDSFQEAQIK
CDKL4 (Cyclin Dependent Kinase Like 4) is a Protein Coding gene. Diseases associated with CDKL4 include Orofaciodigital Syndrome Vi. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is CDKL1. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclin-dependent kinase-like 4; EC 2.7.11.22
Gene Aliases: CDKL4
UniProt ID: (Human) Q5MAI5
Entrez Gene ID: (Human) 344387
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support