Sequence of this protein is as follows: MLELRHRGSC PGPREAVSPP HREGEAAGGD HETESTSDKE TDIDDRYGDL DSRTDSDIPE IPPSSDRTPE ILKKALSGLS SRWKNWWIRG ILTLTMIS
Breakdown products of phosphoinositides are ubiquitous second messengers that function downstream of many G protein-coupled receptors and tyrosine kinases regulating cell growth, calcium metabolism, and protein kinase C activity. This gene encodes an enzyme which regulates the amount of phosphatidylinositol available for signaling by catalyzing the conversion of phosphatidic acid to CDP-diacylglycerol. This enzyme is an integral membrane protein localized to two subcellular domains, the matrix side of the inner mitochondrial membrane where it is thought to be involved in the synthesis of phosphatidylglycerol and cardiolipin and the cytoplasmic side of the endoplasmic reticulum where it functions in phosphatidylinositol biosynthesis. Two genes encoding this enzyme have been identified in humans, one mapping to human chromosome 4q21 and a second to 20p13. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDP-DAG synthase 1; CDP-DG synthase 1; CDP-DG synthetase 1; CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 1; CDP-diacylglycerol synthase 1; CDP-diglyceride pyrophosphorylase 1; CDP-diglyceride synthase 1; CDP-diglyceride synthetase 1; CDS 1; CTP:phosphatidate cytidylyltransferase 1; Phosphatidate cytidylyltransferase 1
Gene Aliases: CDS; CDS1
UniProt ID: (Human) Q92903
Entrez Gene ID: (Human) 1040
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support