Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
RNA-binding protein implicated in the regulation of pre-mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of TNNT2 in a muscle-specific splicing enhancer (MSE)-dependent manner. Promotes also exon exclusion of INSR pre-mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bruno -like 6, RNA binding protein; bruno-like 6, RNA binding protein; Bruno-like protein 6; CELF-6; CUG-BP and ETR-3 like factor 6; CUG-BP- and ETR-3-like factor 6; CUGBP Elav-like family member 6; RNA-binding protein BRUNOL-6
Gene Aliases: 6330569O16Rik; BRUNOL6; CELF6
UniProt ID: (Human) Q96J87, (Mouse) Q7TN33
Entrez Gene ID: (Human) 60677, (Mouse) 76183
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support