Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
A novel gene containing SNF2, PHD-finger, RING-finger, helicase, and linker histone domains termed SHPRH, is a possible candidate for a tumor suppressor gene. These domains are characteristic of several DNA repair proteins, transcription factors, and helicases. Mapping of this gene to 6q24, a region reported to contain a tumor suppressor locus suggests a role in tumor suppression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2610103K11Rik; AA450458; AU024614; BC006883; CENP-I; Centromere protein I; D230017O13Rik; E130018M05; Follicle-stimulating hormone primary response protein; FSH primary response 1; FSH primary response protein 1; Interphase centromere complex protein 19; Leucine-rich primary response protein 1; Mis6
Gene Aliases: CENP-I; CENPI; FSHPRH1; ICEN19; LRPR1
UniProt ID: (Human) Q92674
Entrez Gene ID: (Human) 2491
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support