Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells.
This gene encodes a centrosomal protein that is both required for the regulation of centrosome-related events during the cell cycle, and required for ciliogenesis. The encoded protein has an N-terminal leucine-rich repeat (LRR) domain with six consecutive LRR repeats, and a C-terminal coiled-coil domain. It interacts with the N-terminal catalytic domain of polo-like kinase 4 (PLK4) and colocalizes with PLK4 to the distal end of the centriole. Naturally occurring mutations in this gene cause defects in primary cilia that result in retinal degeneration and sensorineural hearing loss which are associated with cone-rod degeneration disease as well as Usher syndrome. Low expression of this gene is associated with poor prognosis of colorectal cancer patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosomal protein 78kDa; Centrosomal protein of 78 kDa; Cep78
Gene Aliases: C9orf81; CEP78
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support