The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine
May play a role during embryogenesis. Its mouse homolog is involved in embryogenesis and craniofacial development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bucentaur; centromere protein 29; Craniofacial development protein 1; phosphoprotein (Bucentaur)
Gene Aliases: BCNT; BUCENTAUR; CENP-29; CFDP1; CP27; p97; SWC5; Yeti
UniProt ID: (Human) Q9UEE9
Entrez Gene ID: (Human) 10428
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support