Chromatin assembly factor 1 (CAF-1) is a member of the histone chaperone family. It is required for inheritance of epigenetically determined chromosomal states, and in vitro, it promotes the assembly of chromatin during DNA replication and DNA repair. CAF-1 is a complex of three subunits; p150, p60, and p48. This complex is localized to the nucleus of human cells. Both the composition and the biochemical activity of CAF-1 are conserved among human, yeast, and Drosophila.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAF-1 subunit B; CAF-I 60 kDa subunit; CAF-I p60; CAF1 p60; Chromatin assembly factor 1 subunit B; Chromatin assembly factor I p60 subunit; human chromatin assembly factor-I p60 subunit; M-phase phosphoprotein 7; p60 CAF1; p60-CAF1
Gene Aliases: CAF-1; CAF-IP60; CAF1; CAF1A; CAF1P60; CHAF1B; MPHOSPH7; MPP7
UniProt ID: (Human) Q13112
Entrez Gene ID: (Human) 8208
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support